Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99675518-99676007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A1
Entrez Gene ID
1295
Gene Name
collagen type VIII alpha 1
Gene Aliases
C3orf7
UniGene
Hs.654548
Gene Chromosome Location
Chr.3: 99638596-99796733 on Build GRCh38
Transcripts
NM_001850.4 NM_020351.3

SNP Detail

SNP ID Chromosome Location
rs567567293 Chr.3: 99675611-99675611 on Build GRCh38
rs1013485450 Chr.3: 99675783-99675783 on Build GRCh38
rs773690042 Chr.3: 99675752-99675752 on Build GRCh38
rs760710100 Chr.3: 99675753-99675753 on Build GRCh38
rs964834619 Chr.3: 99675866-99675866 on Build GRCh38
rs1057054630 Chr.3: 99675762-99675762 on Build GRCh38
rs1035307634 Chr.3: 99675560-99675560 on Build GRCh38
rs977906446 Chr.3: 99675797-99675797 on Build GRCh38
rs572275929 Chr.3: 99675743-99675743 on Build GRCh38
rs934585442 Chr.3: 99675671-99675671 on Build GRCh38
rs936331401 Chr.3: 99675642-99675642 on Build GRCh38
rs760199098 Chr.3: 99675790-99675790 on Build GRCh38
rs183064298 Chr.3: 99675887-99675887 on Build GRCh38
rs562506453 Chr.3: 99675844-99675844 on Build GRCh38
rs537813934 Chr.3: 99675622-99675622 on Build GRCh38
rs1051898109 Chr.3: 99675683-99675683 on Build GRCh38
rs910387094 Chr.3: 99675644-99675644 on Build GRCh38
rs555345625 Chr.3: 99675589-99675589 on Build GRCh38
rs966504416 Chr.3: 99675602-99675603 on Build GRCh38
rs551530412 Chr.3: 99675891-99675891 on Build GRCh38
rs964601932 Chr.3: 99675650-99675650 on Build GRCh38
rs974383127 Chr.3: 99675821-99675821 on Build GRCh38
rs150747167 Chr.3: 99675769-99675769 on Build GRCh38
rs486688 Chr.3: 99675963-99675963 on Build GRCh38
rs545140512 Chr.3: 99675717-99675717 on Build GRCh38
rs960634035 Chr.3: 99675568-99675568 on Build GRCh38
rs527461170 Chr.3: 99675958-99675958 on Build GRCh38
rs534059486 Chr.3: 99675579-99675579 on Build GRCh38
rs190662581 Chr.3: 99675815-99675815 on Build GRCh38
rs955942679 Chr.3: 99675909-99675909 on Build GRCh38
rs538159015 Chr.3: 99675966-99675966 on Build GRCh38
rs938108721 Chr.3: 99675970-99675970 on Build GRCh38
rs1037825215 Chr.3: 99675878-99675878 on Build GRCh38
rs73858891 Chr.3: 99675623-99675623 on Build GRCh38
rs185959588 Chr.3: 99675745-99675745 on Build GRCh38
rs913896694 Chr.3: 99675954-99675954 on Build GRCh38
rs940854255 Chr.3: 99675874-99675874 on Build GRCh38
rs559155125 Chr.3: 99675652-99675652 on Build GRCh38
rs937371734 Chr.3: 99675946-99675946 on Build GRCh38
rs566595204 Chr.3: 99675950-99675950 on Build GRCh38
rs1002399929 Chr.3: 99675755-99675755 on Build GRCh38
rs1024822952 Chr.3: 99675789-99675789 on Build GRCh38
rs113100818 Chr.3: 99675734-99675734 on Build GRCh38
rs1025948978 Chr.3: 99675795-99675795 on Build GRCh38
rs543861614 Chr.3: 99675843-99675843 on Build GRCh38
rs753726016 Chr.3: 99675962-99675962 on Build GRCh38
rs933535048 Chr.3: 99675676-99675676 on Build GRCh38
rs975911117 Chr.3: 99675880-99675880 on Build GRCh38
rs577715005 Chr.3: 99675670-99675670 on Build GRCh38
rs993156078 Chr.3: 99675592-99675592 on Build GRCh38
rs767846793 Chr.3: 99675959-99675959 on Build GRCh38
rs908056595 Chr.3: 99675864-99675864 on Build GRCh38
rs925015937 Chr.3: 99675814-99675814 on Build GRCh38
rs959279513 Chr.3: 99675770-99675770 on Build GRCh38
rs566705771 Chr.3: 99675547-99675547 on Build GRCh38
rs921575386 Chr.3: 99675824-99675824 on Build GRCh38
rs768136463 Chr.3: 99675610-99675610 on Build GRCh38
rs186330144 Chr.3: 99675976-99675976 on Build GRCh38
rs41267901 Chr.3: 99675848-99675848 on Build GRCh38

View Full Product Details