Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 99795613-99796114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL8A1
Entrez Gene ID
1295
Gene Name
collagen type VIII alpha 1
Gene Aliases
C3orf7
UniGene
Hs.654548
Gene Chromosome Location
Chr.3: 99638596-99796733 on Build GRCh38
Transcripts
NM_001850.4 NM_020351.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3945799 c.1875T>A p.P625P Chr.3: 99795776-99795776 on Build GRCh38
6990511 c.1794C>G p.P598P Chr.3: 99795695-99795695 on Build GRCh38
3945801 c.2146C>A p.L716I Chr.3: 99796047-99796047 on Build GRCh38
7181442 c.2047G>A p.E683K Chr.3: 99795948-99795948 on Build GRCh38
5941781 c.1888G>A p.G630R Chr.3: 99795789-99795789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763969341 Chr.3: 99795771-99795771 on Build GRCh38
rs140378684 Chr.3: 99795686-99795686 on Build GRCh38
rs368547664 Chr.3: 99795740-99795740 on Build GRCh38
rs367839810 Chr.3: 99795813-99795813 on Build GRCh38
rs750024152 Chr.3: 99795781-99795781 on Build GRCh38
rs770660787 Chr.3: 99796031-99796031 on Build GRCh38
rs1047151837 Chr.3: 99795936-99795936 on Build GRCh38
rs757946565 Chr.3: 99795983-99795983 on Build GRCh38
rs768709521 Chr.3: 99796011-99796011 on Build GRCh38
rs761529802 Chr.3: 99795850-99795850 on Build GRCh38
rs987871878 Chr.3: 99795670-99795670 on Build GRCh38
rs757041087 Chr.3: 99795776-99795776 on Build GRCh38
rs145535583 Chr.3: 99795692-99795692 on Build GRCh38
rs375160699 Chr.3: 99795767-99795767 on Build GRCh38
rs760912063 Chr.3: 99795825-99795825 on Build GRCh38
rs777703141 Chr.3: 99796013-99796013 on Build GRCh38
rs774531978 Chr.3: 99795745-99795745 on Build GRCh38
rs756412346 Chr.3: 99795999-99795999 on Build GRCh38
rs573385581 Chr.3: 99795964-99795964 on Build GRCh38
rs760433031 Chr.3: 99795920-99795920 on Build GRCh38
rs770795680 Chr.3: 99795863-99795863 on Build GRCh38
rs757707029 Chr.3: 99795991-99795991 on Build GRCh38
rs377127489 Chr.3: 99795683-99795683 on Build GRCh38
rs754507179 Chr.3: 99795862-99795862 on Build GRCh38
rs371441680 Chr.3: 99795757-99795757 on Build GRCh38
rs887289195 Chr.3: 99795743-99795743 on Build GRCh38
rs753331382 Chr.3: 99795855-99795855 on Build GRCh38
rs766141769 Chr.3: 99795972-99795972 on Build GRCh38
rs1049114344 Chr.3: 99795708-99795708 on Build GRCh38
rs746582848 Chr.3: 99795639-99795639 on Build GRCh38
rs758925301 Chr.3: 99795947-99795947 on Build GRCh38
rs552134484 Chr.3: 99795691-99795691 on Build GRCh38
rs759228709 Chr.3: 99796032-99796032 on Build GRCh38
rs140332231 Chr.3: 99795953-99795953 on Build GRCh38
rs775994874 Chr.3: 99795731-99795731 on Build GRCh38
rs777363083 Chr.3: 99795698-99795698 on Build GRCh38
rs150077356 Chr.3: 99795985-99795985 on Build GRCh38
rs899981606 Chr.3: 99795984-99795984 on Build GRCh38
rs776879409 Chr.3: 99795899-99795899 on Build GRCh38
rs762335752 Chr.3: 99796086-99796086 on Build GRCh38
rs371735925 Chr.3: 99795828-99795828 on Build GRCh38
rs774163244 Chr.3: 99795907-99795907 on Build GRCh38
rs754460957 Chr.3: 99795932-99795932 on Build GRCh38
rs746702523 Chr.3: 99795704-99795704 on Build GRCh38
rs143399698 Chr.3: 99795945-99795945 on Build GRCh38
rs751357905 Chr.3: 99795773-99795773 on Build GRCh38
rs571065653 Chr.3: 99795786-99795786 on Build GRCh38
rs370855024 Chr.3: 99795696-99795696 on Build GRCh38
rs753516059 Chr.3: 99795930-99795930 on Build GRCh38
rs375501498 Chr.3: 99795738-99795738 on Build GRCh38
rs762217511 Chr.3: 99795751-99795751 on Build GRCh38
rs746882914 Chr.3: 99796016-99796016 on Build GRCh38
rs568437951 Chr.3: 99795807-99795807 on Build GRCh38
rs563077439 Chr.3: 99795647-99795647 on Build GRCh38
rs763636624 Chr.3: 99796062-99796062 on Build GRCh38
rs375350800 Chr.3: 99795969-99795969 on Build GRCh38
rs773043396 Chr.3: 99795915-99795915 on Build GRCh38
rs575838433 Chr.3: 99795875-99795875 on Build GRCh38
rs750635074 Chr.3: 99795996-99795996 on Build GRCh38
rs190381283 Chr.3: 99795913-99795913 on Build GRCh38
rs950157934 Chr.3: 99795723-99795723 on Build GRCh38
rs147290721 Chr.3: 99795759-99795759 on Build GRCh38
rs773581749 Chr.3: 99795824-99795824 on Build GRCh38
rs755629975 Chr.3: 99795784-99795784 on Build GRCh38
rs752145764 Chr.3: 99796076-99796076 on Build GRCh38
rs764737921 Chr.3: 99795940-99795940 on Build GRCh38
rs750843916 Chr.3: 99796091-99796091 on Build GRCh38
rs781583659 Chr.3: 99795995-99795995 on Build GRCh38
rs749350499 Chr.3: 99795968-99795968 on Build GRCh38
rs761702469 Chr.3: 99795911-99795911 on Build GRCh38
rs763369239 Chr.3: 99795675-99795675 on Build GRCh38
rs770330907 Chr.3: 99795660-99795660 on Build GRCh38
rs201189348 Chr.3: 99795885-99795885 on Build GRCh38
rs143982019 Chr.3: 99795695-99795695 on Build GRCh38
rs944063386 Chr.3: 99795876-99795876 on Build GRCh38
rs762757556 Chr.3: 99795770-99795770 on Build GRCh38
rs748629899 Chr.3: 99795869-99795869 on Build GRCh38
rs375268132 Chr.3: 99795870-99795870 on Build GRCh38
rs746394191 Chr.3: 99795651-99795651 on Build GRCh38
rs376085430 Chr.3: 99795762-99795762 on Build GRCh38
rs567228826 Chr.3: 99796037-99796037 on Build GRCh38
rs757686994 Chr.3: 99795954-99795954 on Build GRCh38
rs62283505 Chr.3: 99795785-99795785 on Build GRCh38
rs144956888 Chr.3: 99795789-99795789 on Build GRCh38
rs760894116 Chr.3: 99795761-99795761 on Build GRCh38
rs764312334 Chr.3: 99795676-99795676 on Build GRCh38
rs866517871 Chr.3: 99795684-99795684 on Build GRCh38
rs115676498 Chr.3: 99795758-99795758 on Build GRCh38
rs767659023 Chr.3: 99795690-99795690 on Build GRCh38
rs767237025 Chr.3: 99795852-99795852 on Build GRCh38
rs762606970 Chr.3: 99796054-99796054 on Build GRCh38
rs753058238 Chr.3: 99795689-99795690 on Build GRCh38
rs752303108 Chr.3: 99795982-99795982 on Build GRCh38
rs200281259 Chr.3: 99796039-99796039 on Build GRCh38
rs766014652 Chr.3: 99795921-99795921 on Build GRCh38
rs775941656 Chr.3: 99795672-99795672 on Build GRCh38
rs778398809 Chr.3: 99795948-99795948 on Build GRCh38
rs760996196 Chr.3: 99795821-99795822 on Build GRCh38
rs780885710 Chr.3: 99795777-99795777 on Build GRCh38
rs139277585 Chr.3: 99795874-99795874 on Build GRCh38
rs764675709 Chr.3: 99795979-99795979 on Build GRCh38
rs759186515 Chr.3: 99795974-99795974 on Build GRCh38
rs770477385 Chr.3: 99795645-99795645 on Build GRCh38
rs769093108 Chr.3: 99795798-99795798 on Build GRCh38
rs540541234 Chr.3: 99795965-99795965 on Build GRCh38
rs868374775 Chr.3: 99795726-99795726 on Build GRCh38
rs758837285 Chr.3: 99795871-99795871 on Build GRCh38
rs201819230 Chr.3: 99795839-99795839 on Build GRCh38
rs372983340 Chr.3: 99796007-99796007 on Build GRCh38
rs145251667 Chr.3: 99795905-99795905 on Build GRCh38
rs148059974 Chr.3: 99795970-99795970 on Build GRCh38
rs779659047 Chr.3: 99795854-99795854 on Build GRCh38
rs775273019 Chr.3: 99796040-99796040 on Build GRCh38
rs748528586 Chr.3: 99795810-99795810 on Build GRCh38
rs767892937 Chr.3: 99796088-99796088 on Build GRCh38
rs763974579 Chr.3: 99795844-99795844 on Build GRCh38
rs967877876 Chr.3: 99795819-99795819 on Build GRCh38
rs781323203 Chr.3: 99795957-99795957 on Build GRCh38
rs776076529 Chr.3: 99795649-99795649 on Build GRCh38
rs776522099 Chr.3: 99795829-99795829 on Build GRCh38
rs751506766 Chr.3: 99795846-99795846 on Build GRCh38
rs750108911 Chr.3: 99795853-99795853 on Build GRCh38
rs369054191 Chr.3: 99795971-99795971 on Build GRCh38
rs181756171 Chr.3: 99795705-99795705 on Build GRCh38
rs769284042 Chr.3: 99795725-99795725 on Build GRCh38

View Full Product Details