Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101224536-101225043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG2
Entrez Gene ID
50939
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, RP56, SPACRCAN, VMD5
UniGene
Hs.209249
Gene Chromosome Location
Chr.3: 101222546-101320575 on Build GRCh38
Transcripts
NM_016247.3

SNP Detail

SNP ID Chromosome Location
rs886815712 Chr.3: 101224694-101224694 on Build GRCh38
rs1055990743 Chr.3: 101224591-101224592 on Build GRCh38
rs369826537 Chr.3: 101224850-101224850 on Build GRCh38
rs187323753 Chr.3: 101224745-101224745 on Build GRCh38
rs561589565 Chr.3: 101224849-101224849 on Build GRCh38
rs958622477 Chr.3: 101224996-101224996 on Build GRCh38
rs987489861 Chr.3: 101224592-101224592 on Build GRCh38
rs1020313948 Chr.3: 101224603-101224603 on Build GRCh38
rs973772805 Chr.3: 101224795-101224795 on Build GRCh38
rs547458849 Chr.3: 101224888-101224888 on Build GRCh38
rs183226725 Chr.3: 101224669-101224669 on Build GRCh38
rs1038005376 Chr.3: 101224722-101224722 on Build GRCh38
rs941015800 Chr.3: 101224720-101224720 on Build GRCh38
rs191772691 Chr.3: 101224874-101224874 on Build GRCh38
rs547795358 Chr.3: 101224598-101224598 on Build GRCh38
rs370293706 Chr.3: 101224875-101224875 on Build GRCh38
rs113855688 Chr.3: 101225015-101225015 on Build GRCh38
rs950156175 Chr.3: 101224623-101224623 on Build GRCh38
rs72930551 Chr.3: 101224691-101224691 on Build GRCh38
rs564899023 Chr.3: 101224816-101224816 on Build GRCh38
rs34365019 Chr.3: 101224890-101224891 on Build GRCh38
rs937586760 Chr.3: 101224590-101224590 on Build GRCh38
rs748729628 Chr.3: 101224584-101224584 on Build GRCh38
rs1037964110 Chr.3: 101224575-101224575 on Build GRCh38
rs925857014 Chr.3: 101224632-101224632 on Build GRCh38
rs937622197 Chr.3: 101224857-101224857 on Build GRCh38
rs774459531 Chr.3: 101224948-101224948 on Build GRCh38
rs1054491765 Chr.3: 101224971-101224971 on Build GRCh38
rs996361310 Chr.3: 101224732-101224732 on Build GRCh38
rs531023160 Chr.3: 101224727-101224727 on Build GRCh38
rs565670029 Chr.3: 101224946-101224946 on Build GRCh38
rs542970653 Chr.3: 101224810-101224810 on Build GRCh38
rs144532789 Chr.3: 101224637-101224637 on Build GRCh38
rs139105267 Chr.3: 101224820-101224820 on Build GRCh38
rs552610520 Chr.3: 101224758-101224758 on Build GRCh38
rs937199887 Chr.3: 101224641-101224641 on Build GRCh38
rs917056712 Chr.3: 101224661-101224661 on Build GRCh38
rs967766128 Chr.3: 101224609-101224609 on Build GRCh38
rs991336369 Chr.3: 101224658-101224658 on Build GRCh38
rs978743039 Chr.3: 101224630-101224630 on Build GRCh38
rs541519478 Chr.3: 101224650-101224650 on Build GRCh38
rs1032974342 Chr.3: 101224700-101224700 on Build GRCh38
rs146169742 Chr.3: 101224790-101224790 on Build GRCh38
rs144012012 Chr.3: 101224949-101224949 on Build GRCh38
rs74941051 Chr.3: 101224565-101224565 on Build GRCh38

View Full Product Details