Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101226870-101227373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IMPG2
Entrez Gene ID
50939
Gene Name
interphotoreceptor matrix proteoglycan 2
Gene Aliases
IPM200, RP56, SPACRCAN, VMD5
UniGene
Hs.209249
Gene Chromosome Location
Chr.3: 101222546-101320575 on Build GRCh38
Transcripts
NM_016247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6192727 c.3716A>G p.E1239G Chr.3: 101226979-101226979 on Build GRCh38
5897440 c.3714G>A p.V1238V Chr.3: 101226981-101226981 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777280206 Chr.3: 101227217-101227217 on Build GRCh38
rs774726948 Chr.3: 101226956-101226956 on Build GRCh38
rs546346736 Chr.3: 101227041-101227041 on Build GRCh38
rs897333132 Chr.3: 101227210-101227210 on Build GRCh38
rs943993289 Chr.3: 101227225-101227225 on Build GRCh38
rs760878163 Chr.3: 101226987-101226987 on Build GRCh38
rs749733874 Chr.3: 101226927-101226927 on Build GRCh38
rs902327128 Chr.3: 101227046-101227046 on Build GRCh38
rs374887946 Chr.3: 101227098-101227101 on Build GRCh38
rs528728124 Chr.3: 101227055-101227055 on Build GRCh38
rs757781992 Chr.3: 101227271-101227271 on Build GRCh38
rs772299291 Chr.3: 101226973-101226973 on Build GRCh38
rs113649855 Chr.3: 101227138-101227138 on Build GRCh38
rs138251963 Chr.3: 101227048-101227048 on Build GRCh38
rs981966662 Chr.3: 101227096-101227096 on Build GRCh38
rs775548050 Chr.3: 101227011-101227011 on Build GRCh38
rs943925290 Chr.3: 101226998-101226998 on Build GRCh38
rs985440565 Chr.3: 101227215-101227215 on Build GRCh38
rs80055627 Chr.3: 101227140-101227140 on Build GRCh38
rs779205718 Chr.3: 101227288-101227288 on Build GRCh38
rs486352 Chr.3: 101227139-101227139 on Build GRCh38
rs369517701 Chr.3: 101226968-101226968 on Build GRCh38
rs952766026 Chr.3: 101227208-101227208 on Build GRCh38
rs368225624 Chr.3: 101227020-101227020 on Build GRCh38
rs1027031074 Chr.3: 101227206-101227206 on Build GRCh38
rs1049678705 Chr.3: 101226944-101226944 on Build GRCh38
rs952902053 Chr.3: 101227025-101227025 on Build GRCh38
rs76704416 Chr.3: 101227080-101227080 on Build GRCh38
rs764084338 Chr.3: 101227021-101227021 on Build GRCh38
rs947755632 Chr.3: 101227309-101227309 on Build GRCh38
rs199541140 Chr.3: 101226952-101226952 on Build GRCh38
rs878960356 Chr.3: 101226900-101226900 on Build GRCh38
rs1035884435 Chr.3: 101227153-101227153 on Build GRCh38
rs552303414 Chr.3: 101227295-101227295 on Build GRCh38
rs529227587 Chr.3: 101227124-101227124 on Build GRCh38
rs142337066 Chr.3: 101227103-101227103 on Build GRCh38
rs1053524829 Chr.3: 101227056-101227056 on Build GRCh38
rs761687323 Chr.3: 101227028-101227028 on Build GRCh38
rs1024040043 Chr.3: 101227070-101227070 on Build GRCh38
rs977061647 Chr.3: 101227243-101227243 on Build GRCh38
rs915034635 Chr.3: 101227135-101227135 on Build GRCh38
rs767727420 Chr.3: 101227105-101227105 on Build GRCh38
rs923795614 Chr.3: 101227100-101227100 on Build GRCh38
rs755707438 Chr.3: 101227211-101227211 on Build GRCh38
rs1041276525 Chr.3: 101227169-101227169 on Build GRCh38
rs776570156 Chr.3: 101227012-101227012 on Build GRCh38
rs1003510712 Chr.3: 101227151-101227151 on Build GRCh38
rs906171088 Chr.3: 101227342-101227342 on Build GRCh38
rs1003153404 Chr.3: 101227348-101227348 on Build GRCh38
rs113265320 Chr.3: 101227139-101227139 on Build GRCh38
rs751353632 Chr.3: 101227017-101227017 on Build GRCh38
rs748442117 Chr.3: 101226958-101226958 on Build GRCh38
rs956499391 Chr.3: 101227107-101227107 on Build GRCh38
rs533275929 Chr.3: 101227163-101227163 on Build GRCh38
rs766341426 Chr.3: 101226995-101226995 on Build GRCh38
rs533992550 Chr.3: 101227011-101227012 on Build GRCh38
rs78143117 Chr.3: 101227326-101227326 on Build GRCh38
rs754725131 Chr.3: 101227079-101227079 on Build GRCh38
rs773413755 Chr.3: 101226979-101226979 on Build GRCh38
rs768510691 Chr.3: 101226922-101226924 on Build GRCh38
rs184993326 Chr.3: 101227012-101227012 on Build GRCh38
rs377140286 Chr.3: 101226924-101226924 on Build GRCh38
rs562077746 Chr.3: 101227121-101227121 on Build GRCh38
rs972883557 Chr.3: 101227194-101227194 on Build GRCh38
rs749766558 Chr.3: 101227269-101227269 on Build GRCh38

View Full Product Details