Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 108550188-108550688 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
UniGene
Hs.591308
Gene Chromosome Location
Chr.3: 108545752-108589644 on Build GRCh38
Transcripts
NM_020890.2 XM_006713716.3 XM_006713717.2 XM_011513056.1 XM_011513057.2

SNP Detail

SNP ID Chromosome Location
rs1016562262 Chr.3: 108550280-108550280 on Build GRCh38
rs879424887 Chr.3: 108550566-108550566 on Build GRCh38
rs886611648 Chr.3: 108550253-108550253 on Build GRCh38
rs190827314 Chr.3: 108550405-108550405 on Build GRCh38
rs779718250 Chr.3: 108550470-108550470 on Build GRCh38
rs937658938 Chr.3: 108550483-108550483 on Build GRCh38
rs996203432 Chr.3: 108550329-108550329 on Build GRCh38
rs1036145788 Chr.3: 108550608-108550608 on Build GRCh38
rs1051373486 Chr.3: 108550212-108550212 on Build GRCh38
rs573975471 Chr.3: 108550353-108550353 on Build GRCh38
rs562759880 Chr.3: 108550400-108550400 on Build GRCh38
rs896258339 Chr.3: 108550224-108550224 on Build GRCh38
rs899305513 Chr.3: 108550258-108550258 on Build GRCh38
rs954789349 Chr.3: 108550378-108550378 on Build GRCh38
rs949175216 Chr.3: 108550234-108550234 on Build GRCh38
rs1015527091 Chr.3: 108550245-108550245 on Build GRCh38
rs868047064 Chr.3: 108550239-108550239 on Build GRCh38
rs186379490 Chr.3: 108550218-108550218 on Build GRCh38
rs995403161 Chr.3: 108550305-108550305 on Build GRCh38
rs940450628 Chr.3: 108550582-108550582 on Build GRCh38
rs9836969 Chr.3: 108550533-108550533 on Build GRCh38
rs989309839 Chr.3: 108550387-108550388 on Build GRCh38
rs79148379 Chr.3: 108550217-108550217 on Build GRCh38
rs1034986280 Chr.3: 108550401-108550401 on Build GRCh38
rs890097919 Chr.3: 108550581-108550581 on Build GRCh38
rs886603011 Chr.3: 108550579-108550579 on Build GRCh38
rs1051381773 Chr.3: 108550534-108550534 on Build GRCh38
rs925231916 Chr.3: 108550457-108550457 on Build GRCh38
rs982143681 Chr.3: 108550383-108550383 on Build GRCh38
rs768787468 Chr.3: 108550367-108550367 on Build GRCh38
rs942207852 Chr.3: 108550586-108550586 on Build GRCh38
rs913069345 Chr.3: 108550471-108550471 on Build GRCh38
rs921890301 Chr.3: 108550501-108550501 on Build GRCh38
rs995055467 Chr.3: 108550657-108550657 on Build GRCh38
rs796705436 Chr.3: 108550451-108550451 on Build GRCh38
rs898892465 Chr.3: 108550637-108550637 on Build GRCh38
rs542649589 Chr.3: 108550375-108550375 on Build GRCh38
rs1047977091 Chr.3: 108550541-108550541 on Build GRCh38
rs182733486 Chr.3: 108550485-108550485 on Build GRCh38
rs1023916857 Chr.3: 108550334-108550334 on Build GRCh38
rs937712923 Chr.3: 108550217-108550218 on Build GRCh38
rs947267704 Chr.3: 108550482-108550482 on Build GRCh38
rs753503847 Chr.3: 108550528-108550528 on Build GRCh38
rs561800250 Chr.3: 108550445-108550445 on Build GRCh38

View Full Product Details