Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 112465613-112466123 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTLA
Entrez Gene ID
151888
Gene Name
B and T lymphocyte associated
Gene Aliases
BTLA1, CD272
UniGene
Hs.445162
Gene Chromosome Location
Chr.3: 112458790-112499756 on Build GRCh38
Transcripts
NM_001085357.1 NM_181780.3 XM_011512447.2 XM_017005748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863422 c.857G>T p.C286F Chr.3: 112466121-112466121 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1040977990 Chr.3: 112466040-112466040 on Build GRCh38
rs1008153335 Chr.3: 112465760-112465760 on Build GRCh38
rs769062837 Chr.3: 112466088-112466088 on Build GRCh38
rs35030966 Chr.3: 112466087-112466088 on Build GRCh38
rs889624089 Chr.3: 112465994-112465994 on Build GRCh38
rs577427058 Chr.3: 112465656-112465656 on Build GRCh38
rs772700223 Chr.3: 112465670-112465670 on Build GRCh38
rs984921779 Chr.3: 112465831-112465831 on Build GRCh38
rs2171513 Chr.3: 112466080-112466080 on Build GRCh38
rs1000689245 Chr.3: 112465720-112465720 on Build GRCh38
rs528362277 Chr.3: 112465797-112465797 on Build GRCh38
rs76307483 Chr.3: 112465952-112465952 on Build GRCh38
rs532865354 Chr.3: 112465846-112465846 on Build GRCh38
rs776798639 Chr.3: 112465745-112465745 on Build GRCh38
rs181693180 Chr.3: 112465700-112465700 on Build GRCh38
rs116209341 Chr.3: 112466090-112466090 on Build GRCh38
rs76786565 Chr.3: 112465804-112465804 on Build GRCh38
rs771303615 Chr.3: 112465692-112465692 on Build GRCh38
rs922012334 Chr.3: 112465993-112465993 on Build GRCh38
rs966089055 Chr.3: 112465773-112465773 on Build GRCh38
rs746883407 Chr.3: 112466082-112466082 on Build GRCh38
rs992425895 Chr.3: 112465864-112465864 on Build GRCh38
rs945075982 Chr.3: 112465920-112465920 on Build GRCh38
rs1019321397 Chr.3: 112465772-112465772 on Build GRCh38
rs917551231 Chr.3: 112465909-112465909 on Build GRCh38
rs748034910 Chr.3: 112466058-112466058 on Build GRCh38
rs1020769228 Chr.3: 112465770-112465770 on Build GRCh38
rs770019001 Chr.3: 112466085-112466085 on Build GRCh38
rs889707431 Chr.3: 112465747-112465747 on Build GRCh38
rs936447938 Chr.3: 112465961-112465961 on Build GRCh38
rs559809634 Chr.3: 112465759-112465759 on Build GRCh38
rs1049583610 Chr.3: 112465964-112465964 on Build GRCh38
rs560622730 Chr.3: 112465825-112465825 on Build GRCh38
rs1052158265 Chr.3: 112465744-112465744 on Build GRCh38
rs138145669 Chr.3: 112465842-112465842 on Build GRCh38
rs747196663 Chr.3: 112465666-112465666 on Build GRCh38
rs1008122213 Chr.3: 112466008-112466008 on Build GRCh38
rs896161972 Chr.3: 112465640-112465640 on Build GRCh38
rs563797174 Chr.3: 112465843-112465843 on Build GRCh38
rs933375738 Chr.3: 112466021-112466021 on Build GRCh38
rs566167309 Chr.3: 112465932-112465932 on Build GRCh38
rs952152972 Chr.3: 112465819-112465819 on Build GRCh38
rs534987081 Chr.3: 112466024-112466024 on Build GRCh38
rs2971205 Chr.3: 112465925-112465925 on Build GRCh38
rs561709388 Chr.3: 112465821-112465821 on Build GRCh38
rs373688286 Chr.3: 112466035-112466035 on Build GRCh38
rs771707369 Chr.3: 112466059-112466059 on Build GRCh38
rs1039134216 Chr.3: 112465663-112465663 on Build GRCh38
rs1028185693 Chr.3: 112465729-112465729 on Build GRCh38
rs1026384428 Chr.3: 112465805-112465805 on Build GRCh38
rs530777263 Chr.3: 112465837-112465837 on Build GRCh38
rs917546275 Chr.3: 112465907-112465907 on Build GRCh38
rs1039184772 Chr.3: 112465962-112465962 on Build GRCh38

View Full Product Details