Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 115663740-115664289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAP43
Entrez Gene ID
2596
Gene Name
growth associated protein 43
Gene Aliases
B-50, PP46
UniGene
Hs.134974
Gene Chromosome Location
Chr.3: 115623304-115721487 on Build GRCh38
Transcripts
NM_001130064.1 NM_002045.3 XM_017006128.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4431364 c.134C>T p.S45L Chr.3: 115663912-115663912 on Build GRCh38
6038655 c.114C>T p.G38G Chr.3: 115663892-115663892 on Build GRCh38
3119589 c.65T>A p.L22* Chr.3: 115663843-115663843 on Build GRCh38
445342 c.129G>T p.W43C Chr.3: 115663907-115663907 on Build GRCh38
4787680 c.58G>A p.G20R Chr.3: 115663836-115663836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187989039 Chr.3: 115663944-115663944 on Build GRCh38
rs192458019 Chr.3: 115663971-115663971 on Build GRCh38
rs749461861 Chr.3: 115663955-115663955 on Build GRCh38
rs576408045 Chr.3: 115663806-115663806 on Build GRCh38
rs980264134 Chr.3: 115664086-115664086 on Build GRCh38
rs777715681 Chr.3: 115663877-115663877 on Build GRCh38
rs183673207 Chr.3: 115663809-115663809 on Build GRCh38
rs763993565 Chr.3: 115663778-115663778 on Build GRCh38
rs962239636 Chr.3: 115664156-115664156 on Build GRCh38
rs906636884 Chr.3: 115664100-115664100 on Build GRCh38
rs984067466 Chr.3: 115664140-115664140 on Build GRCh38
rs1028594160 Chr.3: 115663983-115663983 on Build GRCh38
rs909130489 Chr.3: 115663896-115663896 on Build GRCh38
rs144156729 Chr.3: 115664107-115664107 on Build GRCh38
rs373081251 Chr.3: 115663883-115663883 on Build GRCh38
rs993044381 Chr.3: 115663882-115663882 on Build GRCh38
rs397714398 Chr.3: 115664224-115664224 on Build GRCh38
rs754725718 Chr.3: 115663811-115663811 on Build GRCh38
rs28399378 Chr.3: 115664068-115664068 on Build GRCh38
rs983781934 Chr.3: 115663831-115663831 on Build GRCh38
rs913227530 Chr.3: 115664130-115664130 on Build GRCh38
rs1009767750 Chr.3: 115664070-115664070 on Build GRCh38
rs28399380 Chr.3: 115664222-115664222 on Build GRCh38
rs780984017 Chr.3: 115664128-115664128 on Build GRCh38
rs113612849 Chr.3: 115664061-115664061 on Build GRCh38
rs139252844 Chr.3: 115663975-115663975 on Build GRCh38
rs1033617485 Chr.3: 115663871-115663871 on Build GRCh38
rs1005722966 Chr.3: 115663988-115663988 on Build GRCh38
rs560273819 Chr.3: 115663864-115663864 on Build GRCh38
rs923055539 Chr.3: 115664251-115664251 on Build GRCh38
rs774228599 Chr.3: 115663964-115663964 on Build GRCh38
rs558431830 Chr.3: 115664118-115664118 on Build GRCh38
rs758200430 Chr.3: 115663841-115663841 on Build GRCh38
rs200098020 Chr.3: 115664230-115664230 on Build GRCh38
rs781740844 Chr.3: 115664237-115664237 on Build GRCh38
rs28399377 Chr.3: 115663808-115663808 on Build GRCh38
rs184661560 Chr.3: 115663997-115663997 on Build GRCh38
rs1016162913 Chr.3: 115664147-115664147 on Build GRCh38
rs570326239 Chr.3: 115664129-115664129 on Build GRCh38
rs1033717255 Chr.3: 115664115-115664115 on Build GRCh38
rs555797679 Chr.3: 115664252-115664252 on Build GRCh38
rs1053090570 Chr.3: 115663941-115663941 on Build GRCh38
rs146731442 Chr.3: 115663954-115663954 on Build GRCh38
rs200879857 Chr.3: 115663878-115663878 on Build GRCh38
rs1054081354 Chr.3: 115664013-115664013 on Build GRCh38
rs985915255 Chr.3: 115664262-115664262 on Build GRCh38
rs573299373 Chr.3: 115664030-115664030 on Build GRCh38
rs901045424 Chr.3: 115663787-115663787 on Build GRCh38
rs28399379 Chr.3: 115664204-115664204 on Build GRCh38
rs547710770 Chr.3: 115664055-115664055 on Build GRCh38
rs527459324 Chr.3: 115663938-115663938 on Build GRCh38
rs1025536049 Chr.3: 115664071-115664071 on Build GRCh38
rs574172422 Chr.3: 115663812-115663812 on Build GRCh38
rs972294393 Chr.3: 115663946-115663946 on Build GRCh38
rs531215739 Chr.3: 115663963-115663963 on Build GRCh38
rs768145260 Chr.3: 115663783-115663783 on Build GRCh38
rs778454724 Chr.3: 115664105-115664105 on Build GRCh38
rs980809927 Chr.3: 115663869-115663869 on Build GRCh38
rs940557787 Chr.3: 115663933-115663933 on Build GRCh38
rs867144356 Chr.3: 115663870-115663870 on Build GRCh38
rs888752905 Chr.3: 115663987-115663987 on Build GRCh38
rs566281242 Chr.3: 115664056-115664056 on Build GRCh38
rs542093374 Chr.3: 115663840-115663840 on Build GRCh38
rs961069815 Chr.3: 115664059-115664059 on Build GRCh38
rs28399376 Chr.3: 115663790-115663790 on Build GRCh38
rs376486384 Chr.3: 115664240-115664241 on Build GRCh38

View Full Product Details