Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119365216-119365760 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP31
Entrez Gene ID
57514
Gene Name
Rho GTPase activating protein 31
Gene Aliases
AOS1, CDGAP
UniGene
Hs.668218
Gene Chromosome Location
Chr.3: 119294289-119419476 on Build GRCh38
Transcripts
NM_020754.3 XM_006713714.3 XM_017006955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4889764 c.114G>A p.L38L Chr.3: 119365329-119365329 on Build GRCh38
3391971 c.151G>A p.V51M Chr.3: 119365366-119365366 on Build GRCh38
3664602 c.112T>C p.L38L Chr.3: 119365327-119365327 on Build GRCh38
7196551 c.197G>A p.R66Q Chr.3: 119365412-119365412 on Build GRCh38
4971009 c.177A>C p.G59G Chr.3: 119365392-119365392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530777876 Chr.3: 119365574-119365574 on Build GRCh38
rs774643942 Chr.3: 119365289-119365289 on Build GRCh38
rs544787108 Chr.3: 119365530-119365530 on Build GRCh38
rs116187120 Chr.3: 119365267-119365267 on Build GRCh38
rs772471119 Chr.3: 119365412-119365412 on Build GRCh38
rs529770262 Chr.3: 119365292-119365293 on Build GRCh38
rs991551113 Chr.3: 119365254-119365254 on Build GRCh38
rs757179237 Chr.3: 119365350-119365350 on Build GRCh38
rs138832644 Chr.3: 119365700-119365700 on Build GRCh38
rs770602731 Chr.3: 119365288-119365288 on Build GRCh38
rs774789736 Chr.3: 119365628-119365628 on Build GRCh38
rs763806829 Chr.3: 119365456-119365456 on Build GRCh38
rs963412540 Chr.3: 119365614-119365614 on Build GRCh38
rs770166698 Chr.3: 119365440-119365440 on Build GRCh38
rs752428737 Chr.3: 119365324-119365324 on Build GRCh38
rs777986579 Chr.3: 119365414-119365414 on Build GRCh38
rs867229350 Chr.3: 119365359-119365359 on Build GRCh38
rs751132057 Chr.3: 119365338-119365338 on Build GRCh38
rs1016797430 Chr.3: 119365597-119365597 on Build GRCh38
rs369138946 Chr.3: 119365313-119365313 on Build GRCh38
rs114159352 Chr.3: 119365677-119365677 on Build GRCh38
rs761241753 Chr.3: 119365305-119365305 on Build GRCh38
rs748347343 Chr.3: 119365411-119365411 on Build GRCh38
rs141911639 Chr.3: 119365424-119365424 on Build GRCh38
rs186926261 Chr.3: 119365476-119365476 on Build GRCh38
rs762390921 Chr.3: 119365662-119365662 on Build GRCh38
rs199618631 Chr.3: 119365403-119365403 on Build GRCh38
rs1026238579 Chr.3: 119365663-119365663 on Build GRCh38
rs1034018577 Chr.3: 119365548-119365548 on Build GRCh38
rs942853714 Chr.3: 119365538-119365538 on Build GRCh38
rs943528769 Chr.3: 119365405-119365405 on Build GRCh38
rs757238638 Chr.3: 119365274-119365274 on Build GRCh38
rs868277054 Chr.3: 119365340-119365340 on Build GRCh38
rs561462425 Chr.3: 119365556-119365556 on Build GRCh38
rs772308862 Chr.3: 119365300-119365300 on Build GRCh38
rs370097944 Chr.3: 119365383-119365383 on Build GRCh38
rs191992844 Chr.3: 119365587-119365587 on Build GRCh38
rs777218987 Chr.3: 119365430-119365430 on Build GRCh38
rs768690142 Chr.3: 119365285-119365285 on Build GRCh38
rs17203055 Chr.3: 119365484-119365484 on Build GRCh38
rs543552697 Chr.3: 119365427-119365427 on Build GRCh38
rs113568026 Chr.3: 119365279-119365279 on Build GRCh38
rs775884598 Chr.3: 119365444-119365444 on Build GRCh38
rs909286769 Chr.3: 119365264-119365264 on Build GRCh38
rs748187636 Chr.3: 119365292-119365292 on Build GRCh38
rs910786168 Chr.3: 119365685-119365685 on Build GRCh38
rs758089081 Chr.3: 119365330-119365330 on Build GRCh38
rs781190136 Chr.3: 119365362-119365362 on Build GRCh38
rs986921087 Chr.3: 119365491-119365491 on Build GRCh38
rs1003428798 Chr.3: 119365537-119365537 on Build GRCh38
rs754856287 Chr.3: 119365453-119365454 on Build GRCh38
rs781108896 Chr.3: 119365455-119365456 on Build GRCh38
rs1005791396 Chr.3: 119365559-119365559 on Build GRCh38
rs115297448 Chr.3: 119365487-119365487 on Build GRCh38
rs759961068 Chr.3: 119365320-119365320 on Build GRCh38
rs758280117 Chr.3: 119365277-119365277 on Build GRCh38
rs933544664 Chr.3: 119365291-119365291 on Build GRCh38
rs377483659 Chr.3: 119365436-119365436 on Build GRCh38
rs541152628 Chr.3: 119365410-119365410 on Build GRCh38
rs746919758 Chr.3: 119365270-119365270 on Build GRCh38
rs764749352 Chr.3: 119365321-119365321 on Build GRCh38
rs375072197 Chr.3: 119365365-119365365 on Build GRCh38
rs546354557 Chr.3: 119365705-119365705 on Build GRCh38
rs146089778 Chr.3: 119365664-119365664 on Build GRCh38
rs766966306 Chr.3: 119365310-119365310 on Build GRCh38
rs367872864 Chr.3: 119365594-119365594 on Build GRCh38
rs527930921 Chr.3: 119365333-119365333 on Build GRCh38
rs959477107 Chr.3: 119365247-119365247 on Build GRCh38
rs746915997 Chr.3: 119365311-119365311 on Build GRCh38
rs975548730 Chr.3: 119365539-119365539 on Build GRCh38

View Full Product Details