Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119380824-119381321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP31
Entrez Gene ID
57514
Gene Name
Rho GTPase activating protein 31
Gene Aliases
AOS1, CDGAP
UniGene
Hs.668218
Gene Chromosome Location
Chr.3: 119294289-119419476 on Build GRCh38
Transcripts
NM_020754.3 XM_006713714.3 XM_017006955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3585727 c.418C>T p.P140S Chr.3: 119380973-119380973 on Build GRCh38
445351 c.386C>A p.A129D Chr.3: 119380941-119380941 on Build GRCh38
3992658 c.425A>G p.H142R Chr.3: 119380980-119380980 on Build GRCh38
5732089 c.409G>A p.E137K Chr.3: 119380964-119380964 on Build GRCh38
4800507 c.387C>T p.A129A Chr.3: 119380942-119380942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762846265 Chr.3: 119381289-119381289 on Build GRCh38
rs903962870 Chr.3: 119381101-119381101 on Build GRCh38
rs1032833293 Chr.3: 119381067-119381067 on Build GRCh38
rs371169065 Chr.3: 119380941-119380941 on Build GRCh38
rs1031186787 Chr.3: 119380893-119380893 on Build GRCh38
rs72966428 Chr.3: 119381242-119381242 on Build GRCh38
rs774589511 Chr.3: 119380925-119380927 on Build GRCh38
rs1026706541 Chr.3: 119381268-119381268 on Build GRCh38
rs753974872 Chr.3: 119381004-119381004 on Build GRCh38
rs374160242 Chr.3: 119380918-119380918 on Build GRCh38
rs201216830 Chr.3: 119380944-119380944 on Build GRCh38
rs768628066 Chr.3: 119381297-119381297 on Build GRCh38
rs779233884 Chr.3: 119380924-119380924 on Build GRCh38
rs533480514 Chr.3: 119381173-119381173 on Build GRCh38
rs899654492 Chr.3: 119381050-119381050 on Build GRCh38
rs1000800766 Chr.3: 119381121-119381121 on Build GRCh38
rs202229113 Chr.3: 119380920-119380920 on Build GRCh38
rs769861846 Chr.3: 119381037-119381037 on Build GRCh38
rs756366180 Chr.3: 119380984-119380984 on Build GRCh38
rs778255527 Chr.3: 119380862-119380862 on Build GRCh38
rs531872510 Chr.3: 119381010-119381010 on Build GRCh38
rs749558414 Chr.3: 119380876-119380884 on Build GRCh38
rs746489598 Chr.3: 119380942-119380942 on Build GRCh38
rs757501179 Chr.3: 119380906-119380906 on Build GRCh38
rs759589229 Chr.3: 119380888-119380888 on Build GRCh38
rs932526299 Chr.3: 119381054-119381054 on Build GRCh38
rs1050458654 Chr.3: 119381071-119381071 on Build GRCh38
rs770535119 Chr.3: 119380943-119380943 on Build GRCh38
rs532464648 Chr.3: 119381148-119381148 on Build GRCh38
rs200397968 Chr.3: 119380897-119380897 on Build GRCh38
rs548188313 Chr.3: 119381228-119381228 on Build GRCh38
rs150339878 Chr.3: 119380939-119380939 on Build GRCh38
rs892685721 Chr.3: 119381166-119381166 on Build GRCh38
rs771235886 Chr.3: 119380891-119380893 on Build GRCh38
rs766421058 Chr.3: 119380995-119380995 on Build GRCh38
rs965473995 Chr.3: 119380871-119380871 on Build GRCh38
rs370263730 Chr.3: 119381032-119381032 on Build GRCh38
rs1010107101 Chr.3: 119381165-119381165 on Build GRCh38
rs34026383 Chr.3: 119381103-119381104 on Build GRCh38
rs778240892 Chr.3: 119381020-119381020 on Build GRCh38
rs1000463228 Chr.3: 119381046-119381046 on Build GRCh38
rs775292252 Chr.3: 119380953-119380953 on Build GRCh38
rs374718588 Chr.3: 119380965-119380965 on Build GRCh38
rs763491256 Chr.3: 119380914-119380914 on Build GRCh38
rs755525923 Chr.3: 119380921-119380921 on Build GRCh38
rs781545145 Chr.3: 119381011-119381011 on Build GRCh38
rs533858905 Chr.3: 119381265-119381265 on Build GRCh38
rs1037251368 Chr.3: 119381291-119381291 on Build GRCh38
rs79505114 Chr.3: 119381176-119381176 on Build GRCh38
rs969618457 Chr.3: 119380931-119380931 on Build GRCh38
rs886057798 Chr.3: 119380895-119380897 on Build GRCh38
rs551087333 Chr.3: 119381170-119381170 on Build GRCh38
rs377580895 Chr.3: 119380858-119380858 on Build GRCh38
rs943953162 Chr.3: 119381024-119381024 on Build GRCh38
rs759535946 Chr.3: 119380951-119380951 on Build GRCh38
rs536293363 Chr.3: 119381200-119381200 on Build GRCh38
rs748526834 Chr.3: 119380932-119380932 on Build GRCh38
rs767724642 Chr.3: 119380975-119380975 on Build GRCh38
rs750455296 Chr.3: 119380915-119380915 on Build GRCh38
rs765104598 Chr.3: 119380892-119380892 on Build GRCh38
rs910046055 Chr.3: 119381279-119381279 on Build GRCh38
rs1036736187 Chr.3: 119380945-119380945 on Build GRCh38
rs770767862 Chr.3: 119381036-119381036 on Build GRCh38
rs189874575 Chr.3: 119381293-119381293 on Build GRCh38
rs1052389421 Chr.3: 119381155-119381155 on Build GRCh38
rs1014022084 Chr.3: 119380964-119380964 on Build GRCh38
rs758721508 Chr.3: 119381014-119381014 on Build GRCh38
rs200220489 Chr.3: 119380978-119380978 on Build GRCh38
rs984727532 Chr.3: 119381275-119381275 on Build GRCh38
rs940256583 Chr.3: 119381282-119381282 on Build GRCh38

View Full Product Details