Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119399114-119399611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP31
Entrez Gene ID
57514
Gene Name
Rho GTPase activating protein 31
Gene Aliases
AOS1, CDGAP
UniGene
Hs.668218
Gene Chromosome Location
Chr.3: 119294289-119419476 on Build GRCh38
Transcripts
NM_020754.3 XM_006713714.3 XM_017006955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6603430 c.1042G>A p.D348N Chr.3: 119399234-119399234 on Build GRCh38
362224 c.1007T>A p.V336E Chr.3: 119399199-119399199 on Build GRCh38
1670464 c.1060C>T p.P354S Chr.3: 119399252-119399252 on Build GRCh38
7110912 c.1025G>T p.R342L Chr.3: 119399217-119399217 on Build GRCh38
4112513 c.1034A>T p.K345I Chr.3: 119399226-119399226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145476777 Chr.3: 119399367-119399367 on Build GRCh38
rs558599576 Chr.3: 119399542-119399542 on Build GRCh38
rs746378167 Chr.3: 119399441-119399441 on Build GRCh38
rs747734889 Chr.3: 119399244-119399244 on Build GRCh38
rs568987461 Chr.3: 119399458-119399458 on Build GRCh38
rs377202519 Chr.3: 119399185-119399186 on Build GRCh38
rs931712016 Chr.3: 119399232-119399232 on Build GRCh38
rs905046066 Chr.3: 119399425-119399425 on Build GRCh38
rs1030895272 Chr.3: 119399336-119399336 on Build GRCh38
rs770941672 Chr.3: 119399297-119399297 on Build GRCh38
rs780554721 Chr.3: 119399182-119399182 on Build GRCh38
rs550735466 Chr.3: 119399431-119399431 on Build GRCh38
rs191045060 Chr.3: 119399495-119399495 on Build GRCh38
rs183600436 Chr.3: 119399332-119399332 on Build GRCh38
rs9819603 Chr.3: 119399498-119399498 on Build GRCh38
rs757992468 Chr.3: 119399303-119399303 on Build GRCh38
rs962272391 Chr.3: 119399576-119399576 on Build GRCh38
rs974970363 Chr.3: 119399415-119399415 on Build GRCh38
rs9819443 Chr.3: 119399379-119399379 on Build GRCh38
rs755330707 Chr.3: 119399235-119399235 on Build GRCh38
rs563761648 Chr.3: 119399216-119399216 on Build GRCh38
rs536345599 Chr.3: 119399485-119399485 on Build GRCh38
rs558573589 Chr.3: 119399207-119399207 on Build GRCh38
rs561324826 Chr.3: 119399274-119399274 on Build GRCh38
rs749938601 Chr.3: 119399180-119399180 on Build GRCh38
rs770778446 Chr.3: 119399302-119399302 on Build GRCh38
rs1013478006 Chr.3: 119399416-119399416 on Build GRCh38
rs1019069481 Chr.3: 119399409-119399409 on Build GRCh38
rs141111639 Chr.3: 119399390-119399390 on Build GRCh38
rs201855122 Chr.3: 119399250-119399250 on Build GRCh38
rs767567816 Chr.3: 119399158-119399158 on Build GRCh38
rs917708890 Chr.3: 119399537-119399537 on Build GRCh38
rs116811521 Chr.3: 119399527-119399527 on Build GRCh38
rs762141694 Chr.3: 119399150-119399150 on Build GRCh38
rs4688001 Chr.3: 119399257-119399257 on Build GRCh38
rs376404901 Chr.3: 119399300-119399300 on Build GRCh38
rs770150065 Chr.3: 119399567-119399567 on Build GRCh38
rs765959115 Chr.3: 119399344-119399344 on Build GRCh38
rs199940037 Chr.3: 119399176-119399176 on Build GRCh38
rs368101298 Chr.3: 119399144-119399144 on Build GRCh38
rs966521358 Chr.3: 119399410-119399410 on Build GRCh38
rs751269132 Chr.3: 119399354-119399354 on Build GRCh38
rs186809864 Chr.3: 119399422-119399422 on Build GRCh38
rs936356708 Chr.3: 119399345-119399345 on Build GRCh38
rs1056530035 Chr.3: 119399532-119399532 on Build GRCh38
rs183285772 Chr.3: 119399502-119399502 on Build GRCh38
rs746530408 Chr.3: 119399278-119399278 on Build GRCh38
rs369521143 Chr.3: 119399236-119399236 on Build GRCh38

View Full Product Details