Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121621751-121622256 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO40
Entrez Gene ID
51725
Gene Name
F-box protein 40
Gene Aliases
FBX40
UniGene
Hs.272564
Gene Chromosome Location
Chr.3: 121593323-121630292 on Build GRCh38
Transcripts
NM_016298.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4760730 c.485G>A p.G162D Chr.3: 121621914-121621914 on Build GRCh38
4827466 c.508G>A p.G170R Chr.3: 121621937-121621937 on Build GRCh38
1736192 c.791C>A p.P264H Chr.3: 121622220-121622220 on Build GRCh38
4683996 c.415A>G p.R139G Chr.3: 121621844-121621844 on Build GRCh38
1418158 c.682G>A p.A228T Chr.3: 121622111-121622111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372552970 Chr.3: 121621931-121621931 on Build GRCh38
rs909232803 Chr.3: 121621878-121621878 on Build GRCh38
rs773539981 Chr.3: 121622215-121622215 on Build GRCh38
rs754382635 Chr.3: 121622141-121622141 on Build GRCh38
rs748062583 Chr.3: 121621816-121621816 on Build GRCh38
rs780506306 Chr.3: 121622185-121622185 on Build GRCh38
rs748921319 Chr.3: 121621880-121621880 on Build GRCh38
rs770345962 Chr.3: 121622100-121622100 on Build GRCh38
rs775065669 Chr.3: 121621784-121621784 on Build GRCh38
rs780300146 Chr.3: 121621957-121621957 on Build GRCh38
rs142982539 Chr.3: 121621979-121621979 on Build GRCh38
rs369747294 Chr.3: 121622054-121622054 on Build GRCh38
rs760748351 Chr.3: 121622216-121622216 on Build GRCh38
rs755905296 Chr.3: 121622096-121622097 on Build GRCh38
rs774121135 Chr.3: 121621883-121621883 on Build GRCh38
rs938025587 Chr.3: 121621888-121621888 on Build GRCh38
rs756600680 Chr.3: 121621955-121621955 on Build GRCh38
rs770193085 Chr.3: 121621783-121621783 on Build GRCh38
rs752867730 Chr.3: 121621817-121621817 on Build GRCh38
rs201761738 Chr.3: 121621965-121621965 on Build GRCh38
rs769667052 Chr.3: 121622202-121622202 on Build GRCh38
rs750685750 Chr.3: 121622222-121622222 on Build GRCh38
rs755451725 Chr.3: 121622059-121622059 on Build GRCh38
rs578177867 Chr.3: 121621963-121621963 on Build GRCh38
rs746569619 Chr.3: 121622095-121622095 on Build GRCh38
rs770070135 Chr.3: 121621868-121621868 on Build GRCh38
rs767611473 Chr.3: 121621799-121621799 on Build GRCh38
rs758626441 Chr.3: 121622230-121622246 on Build GRCh38
rs761861151 Chr.3: 121621798-121621798 on Build GRCh38
rs775854273 Chr.3: 121621873-121621873 on Build GRCh38
rs759230547 Chr.3: 121621921-121621921 on Build GRCh38
rs760874090 Chr.3: 121622136-121622136 on Build GRCh38
rs757387582 Chr.3: 121621940-121621940 on Build GRCh38
rs1017101413 Chr.3: 121621814-121621814 on Build GRCh38
rs756038658 Chr.3: 121621809-121621809 on Build GRCh38
rs758806499 Chr.3: 121622167-121622167 on Build GRCh38
rs776532505 Chr.3: 121622230-121622230 on Build GRCh38
rs772810351 Chr.3: 121621975-121621975 on Build GRCh38
rs751796828 Chr.3: 121622008-121622008 on Build GRCh38
rs763515142 Chr.3: 121621787-121621787 on Build GRCh38
rs762041808 Chr.3: 121622123-121622123 on Build GRCh38
rs148776238 Chr.3: 121622108-121622108 on Build GRCh38
rs1056128667 Chr.3: 121622171-121622171 on Build GRCh38
rs760554899 Chr.3: 121621978-121621978 on Build GRCh38
rs267599566 Chr.3: 121621848-121621848 on Build GRCh38
rs769751719 Chr.3: 121622107-121622107 on Build GRCh38
rs1056199005 Chr.3: 121621788-121621788 on Build GRCh38
rs556118975 Chr.3: 121622073-121622073 on Build GRCh38
rs566081092 Chr.3: 121621885-121621885 on Build GRCh38
rs949854189 Chr.3: 121621977-121621977 on Build GRCh38
rs781193078 Chr.3: 121621941-121621941 on Build GRCh38
rs766483889 Chr.3: 121622223-121622223 on Build GRCh38
rs775221948 Chr.3: 121622119-121622119 on Build GRCh38
rs370367749 Chr.3: 121622131-121622131 on Build GRCh38
rs771720396 Chr.3: 121621970-121621970 on Build GRCh38
rs191688932 Chr.3: 121622137-121622137 on Build GRCh38
rs147957871 Chr.3: 121622197-121622197 on Build GRCh38
rs147448149 Chr.3: 121622003-121622003 on Build GRCh38
rs747786850 Chr.3: 121621969-121621969 on Build GRCh38
rs199575816 Chr.3: 121621928-121621928 on Build GRCh38
rs377409069 Chr.3: 121621786-121621786 on Build GRCh38
rs755468260 Chr.3: 121622155-121622155 on Build GRCh38
rs747835644 Chr.3: 121622068-121622068 on Build GRCh38
rs765719429 Chr.3: 121622160-121622160 on Build GRCh38
rs776402291 Chr.3: 121621986-121621986 on Build GRCh38
rs200154550 Chr.3: 121621901-121621901 on Build GRCh38
rs374090102 Chr.3: 121622057-121622057 on Build GRCh38
rs757299930 Chr.3: 121621858-121621858 on Build GRCh38
rs901626100 Chr.3: 121622115-121622115 on Build GRCh38
rs1007346895 Chr.3: 121621791-121621791 on Build GRCh38
rs760595092 Chr.3: 121621897-121621897 on Build GRCh38
rs768432931 Chr.3: 121622212-121622212 on Build GRCh38
rs777545387 Chr.3: 121622076-121622076 on Build GRCh38
rs756122429 Chr.3: 121622028-121622028 on Build GRCh38
rs777550766 Chr.3: 121622168-121622168 on Build GRCh38
rs997067161 Chr.3: 121621919-121621919 on Build GRCh38
rs767772217 Chr.3: 121622124-121622124 on Build GRCh38
rs368822929 Chr.3: 121622207-121622207 on Build GRCh38
rs775379711 Chr.3: 121622209-121622209 on Build GRCh38
rs763898208 Chr.3: 121622188-121622189 on Build GRCh38
rs377295452 Chr.3: 121622062-121622062 on Build GRCh38
rs549329473 Chr.3: 121621835-121621835 on Build GRCh38
rs756807753 Chr.3: 121622174-121622174 on Build GRCh38
rs142436040 Chr.3: 121622172-121622172 on Build GRCh38
rs746363538 Chr.3: 121621865-121621865 on Build GRCh38
rs976661978 Chr.3: 121621832-121621832 on Build GRCh38
rs766181328 Chr.3: 121621815-121621815 on Build GRCh38
rs753621120 Chr.3: 121621920-121621920 on Build GRCh38
rs376105815 Chr.3: 121622027-121622027 on Build GRCh38
rs769583651 Chr.3: 121622113-121622113 on Build GRCh38
rs375428007 Chr.3: 121621881-121621881 on Build GRCh38
rs768870158 Chr.3: 121621964-121621964 on Build GRCh38
rs767131081 Chr.3: 121621889-121621889 on Build GRCh38
rs145400219 Chr.3: 121622163-121622163 on Build GRCh38
rs939747557 Chr.3: 121622188-121622188 on Build GRCh38
rs759446632 Chr.3: 121621992-121621992 on Build GRCh38
rs370258531 Chr.3: 121621806-121621806 on Build GRCh38
rs752659519 Chr.3: 121621812-121621812 on Build GRCh38
rs34751480 Chr.3: 121621796-121621796 on Build GRCh38
rs749119690 Chr.3: 121622210-121622210 on Build GRCh38
rs761959890 Chr.3: 121622011-121622011 on Build GRCh38
rs371738482 Chr.3: 121621943-121621943 on Build GRCh38
rs138735736 Chr.3: 121621849-121621849 on Build GRCh38
rs1054116888 Chr.3: 121622089-121622089 on Build GRCh38
rs773338926 Chr.3: 121621894-121621894 on Build GRCh38

View Full Product Details