Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 121987770-121988281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR1
Entrez Gene ID
286676
Gene Name
immunoglobulin like domain containing receptor 1
Gene Aliases
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta
UniGene
Hs.98484
Gene Chromosome Location
Chr.3: 121987323-122060554 on Build GRCh38
Transcripts
NM_001199799.1 NM_001199800.1 NM_175924.3 XM_005247389.4 XM_011512738.2 XM_011512739.2

SNP Detail

SNP ID Chromosome Location
rs764562889 Chr.3: 121988089-121988089 on Build GRCh38
rs181628071 Chr.3: 121988101-121988101 on Build GRCh38
rs958864459 Chr.3: 121987866-121987866 on Build GRCh38
rs754309930 Chr.3: 121988235-121988235 on Build GRCh38
rs757100881 Chr.3: 121988236-121988236 on Build GRCh38
rs892575339 Chr.3: 121987822-121987822 on Build GRCh38
rs775889751 Chr.3: 121987915-121987915 on Build GRCh38
rs1043721475 Chr.3: 121987833-121987833 on Build GRCh38
rs930052001 Chr.3: 121988174-121988174 on Build GRCh38
rs1009583474 Chr.3: 121987827-121987827 on Build GRCh38
rs964199018 Chr.3: 121988123-121988123 on Build GRCh38
rs571262745 Chr.3: 121988177-121988177 on Build GRCh38
rs759586255 Chr.3: 121987892-121987892 on Build GRCh38
rs951787435 Chr.3: 121988038-121988038 on Build GRCh38
rs1403729 Chr.3: 121987905-121987905 on Build GRCh38
rs563334520 Chr.3: 121987963-121987963 on Build GRCh38
rs1047474505 Chr.3: 121988176-121988176 on Build GRCh38
rs775080019 Chr.3: 121988213-121988213 on Build GRCh38
rs138335629 Chr.3: 121988006-121988006 on Build GRCh38
rs772444510 Chr.3: 121987922-121987922 on Build GRCh38
rs929957411 Chr.3: 121987835-121987835 on Build GRCh38
rs910836218 Chr.3: 121988124-121988124 on Build GRCh38
rs565610957 Chr.3: 121988099-121988099 on Build GRCh38
rs753549007 Chr.3: 121988155-121988155 on Build GRCh38
rs1000523178 Chr.3: 121987850-121987850 on Build GRCh38
rs1006149312 Chr.3: 121987896-121987896 on Build GRCh38
rs574405506 Chr.3: 121987902-121987902 on Build GRCh38
rs1034308459 Chr.3: 121987856-121987856 on Build GRCh38
rs549594681 Chr.3: 121988101-121988102 on Build GRCh38
rs763038125 Chr.3: 121988231-121988231 on Build GRCh38
rs746000667 Chr.3: 121987831-121987831 on Build GRCh38
rs530892554 Chr.3: 121987881-121987881 on Build GRCh38
rs1024679873 Chr.3: 121987971-121987971 on Build GRCh38
rs922641725 Chr.3: 121988161-121988161 on Build GRCh38
rs145679236 Chr.3: 121987815-121987815 on Build GRCh38
rs761032107 Chr.3: 121988058-121988058 on Build GRCh38
rs976240063 Chr.3: 121988158-121988158 on Build GRCh38
rs373788728 Chr.3: 121988034-121988034 on Build GRCh38
rs754067165 Chr.3: 121988248-121988248 on Build GRCh38
rs1024124235 Chr.3: 121987882-121987882 on Build GRCh38
rs1037912911 Chr.3: 121987897-121987897 on Build GRCh38
rs1037619352 Chr.3: 121988219-121988219 on Build GRCh38
rs776379104 Chr.3: 121988170-121988170 on Build GRCh38
rs999114876 Chr.3: 121987990-121987990 on Build GRCh38
rs970455560 Chr.3: 121987973-121987973 on Build GRCh38
rs550947393 Chr.3: 121988151-121988151 on Build GRCh38
rs141335434 Chr.3: 121988234-121988234 on Build GRCh38
rs1047095709 Chr.3: 121987837-121987837 on Build GRCh38
rs941380433 Chr.3: 121988194-121988194 on Build GRCh38
rs1403730 Chr.3: 121987907-121987907 on Build GRCh38
rs767179327 Chr.3: 121988239-121988239 on Build GRCh38
rs189041551 Chr.3: 121988004-121988004 on Build GRCh38
rs1012071018 Chr.3: 121987941-121987941 on Build GRCh38
rs371813355 Chr.3: 121988105-121988106 on Build GRCh38
rs759223170 Chr.3: 121988197-121988197 on Build GRCh38
rs768023200 Chr.3: 121987913-121987913 on Build GRCh38
rs565305925 Chr.3: 121987846-121987846 on Build GRCh38
rs911492468 Chr.3: 121988113-121988113 on Build GRCh38

View Full Product Details