Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122001219-122001714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILDR1
Entrez Gene ID
286676
Gene Name
immunoglobulin like domain containing receptor 1
Gene Aliases
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta
UniGene
Hs.98484
Gene Chromosome Location
Chr.3: 121987323-122060554 on Build GRCh38
Transcripts
NM_001199799.1 NM_001199800.1 NM_175924.3 XM_005247389.4 XM_011512738.2 XM_011512739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685504 c.543C>G p.L181L Chr.3: 122001411-122001411 on Build GRCh38
1037054 c.525C>A p.I175I Chr.3: 122001429-122001429 on Build GRCh38
1037053 c.610C>T p.P204S Chr.3: 122001344-122001344 on Build GRCh38
4840581 c.605G>A p.R202H Chr.3: 122001349-122001349 on Build GRCh38
4840583 c.641G>A p.R214H Chr.3: 122001349-122001349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770277704 Chr.3: 122001497-122001497 on Build GRCh38
rs947703778 Chr.3: 122001459-122001460 on Build GRCh38
rs559607245 Chr.3: 122001672-122001672 on Build GRCh38
rs748439356 Chr.3: 122001292-122001292 on Build GRCh38
rs746431279 Chr.3: 122001485-122001485 on Build GRCh38
rs981185222 Chr.3: 122001606-122001606 on Build GRCh38
rs201297724 Chr.3: 122001422-122001422 on Build GRCh38
rs954646766 Chr.3: 122001674-122001674 on Build GRCh38
rs770157252 Chr.3: 122001382-122001382 on Build GRCh38
rs762656524 Chr.3: 122001394-122001394 on Build GRCh38
rs1022715828 Chr.3: 122001578-122001578 on Build GRCh38
rs778081230 Chr.3: 122001463-122001463 on Build GRCh38
rs35657414 Chr.3: 122001668-122001668 on Build GRCh38
rs1042522028 Chr.3: 122001669-122001669 on Build GRCh38
rs553331037 Chr.3: 122001403-122001403 on Build GRCh38
rs766344111 Chr.3: 122001438-122001438 on Build GRCh38
rs750052759 Chr.3: 122001684-122001684 on Build GRCh38
rs199599600 Chr.3: 122001271-122001271 on Build GRCh38
rs1010985902 Chr.3: 122001634-122001634 on Build GRCh38
rs375603020 Chr.3: 122001441-122001441 on Build GRCh38
rs943576212 Chr.3: 122001636-122001636 on Build GRCh38
rs62269207 Chr.3: 122001647-122001647 on Build GRCh38
rs375328614 Chr.3: 122001286-122001286 on Build GRCh38
rs935460503 Chr.3: 122001527-122001527 on Build GRCh38
rs148433157 Chr.3: 122001350-122001350 on Build GRCh38
rs747091345 Chr.3: 122001302-122001302 on Build GRCh38
rs372743080 Chr.3: 122001257-122001257 on Build GRCh38
rs1032950062 Chr.3: 122001491-122001491 on Build GRCh38
rs556774059 Chr.3: 122001626-122001626 on Build GRCh38
rs765141910 Chr.3: 122001435-122001437 on Build GRCh38
rs1049341194 Chr.3: 122001251-122001251 on Build GRCh38
rs766874607 Chr.3: 122001423-122001423 on Build GRCh38
rs998149933 Chr.3: 122001650-122001652 on Build GRCh38
rs991345897 Chr.3: 122001566-122001566 on Build GRCh38
rs779222843 Chr.3: 122001587-122001587 on Build GRCh38
rs775870033 Chr.3: 122001499-122001499 on Build GRCh38
rs146174741 Chr.3: 122001349-122001349 on Build GRCh38
rs1039284384 Chr.3: 122001644-122001644 on Build GRCh38
rs387907017 Chr.3: 122001371-122001371 on Build GRCh38
rs188126104 Chr.3: 122001630-122001630 on Build GRCh38
rs199584285 Chr.3: 122001682-122001682 on Build GRCh38
rs371665575 Chr.3: 122001272-122001272 on Build GRCh38
rs777756846 Chr.3: 122001300-122001300 on Build GRCh38
rs79120893 Chr.3: 122001258-122001258 on Build GRCh38
rs869222550 Chr.3: 122001680-122001682 on Build GRCh38
rs184163953 Chr.3: 122001617-122001617 on Build GRCh38
rs747157285 Chr.3: 122001469-122001469 on Build GRCh38
rs199829749 Chr.3: 122001370-122001370 on Build GRCh38
rs572716256 Chr.3: 122001690-122001690 on Build GRCh38
rs545010285 Chr.3: 122001691-122001691 on Build GRCh38
rs756111455 Chr.3: 122001436-122001436 on Build GRCh38
rs745900430 Chr.3: 122001614-122001614 on Build GRCh38
rs775815087 Chr.3: 122001393-122001393 on Build GRCh38
rs749628134 Chr.3: 122001502-122001502 on Build GRCh38
rs770865084 Chr.3: 122001344-122001344 on Build GRCh38
rs757394644 Chr.3: 122001576-122001576 on Build GRCh38
rs781074712 Chr.3: 122001480-122001480 on Build GRCh38
rs957487543 Chr.3: 122001496-122001496 on Build GRCh38
rs763768266 Chr.3: 122001400-122001400 on Build GRCh38
rs566703763 Chr.3: 122001513-122001513 on Build GRCh38
rs371521897 Chr.3: 122001377-122001377 on Build GRCh38
rs983318998 Chr.3: 122001331-122001331 on Build GRCh38
rs187493717 Chr.3: 122001254-122001254 on Build GRCh38
rs558595843 Chr.3: 122001601-122001601 on Build GRCh38
rs1039576896 Chr.3: 122001276-122001276 on Build GRCh38
rs772326986 Chr.3: 122001386-122001386 on Build GRCh38
rs191602232 Chr.3: 122001607-122001607 on Build GRCh38
rs984835456 Chr.3: 122001620-122001620 on Build GRCh38
rs758744952 Chr.3: 122001458-122001458 on Build GRCh38
rs879019325 Chr.3: 122001667-122001668 on Build GRCh38
rs755981549 Chr.3: 122001281-122001281 on Build GRCh38
rs777537563 Chr.3: 122001297-122001297 on Build GRCh38
rs200106561 Chr.3: 122001667-122001667 on Build GRCh38
rs775517605 Chr.3: 122001521-122001521 on Build GRCh38
rs772420417 Chr.3: 122001294-122001294 on Build GRCh38
rs747644884 Chr.3: 122001474-122001474 on Build GRCh38
rs56126849 Chr.3: 122001682-122001682 on Build GRCh38
rs539036232 Chr.3: 122001591-122001591 on Build GRCh38
rs750391703 Chr.3: 122001431-122001431 on Build GRCh38

View Full Product Details