Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122055174-122055702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
UniGene
Hs.171182
Gene Chromosome Location
Chr.3: 122055362-122121143 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_175862.4 XM_011512739.2

Gene Symbol
ILDR1
Entrez Gene ID
286676
Gene Name
immunoglobulin like domain containing receptor 1
Gene Aliases
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta
UniGene
Hs.98484
Gene Chromosome Location
Chr.3: 121987323-122060554 on Build GRCh38
Transcripts
NM_001206924.1 NM_001206925.1 NM_175862.4 XM_011512739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266243 c.14+5G>T p.? Chr.3: 122055508-122055508 on Build GRCh38
3586188 c.8C>T p.P3L Chr.3: 122055497-122055497 on Build GRCh38
3586187 c.3G>A p.M1I Chr.3: 122055492-122055492 on Build GRCh38
5290700 c.4G>A p.D2N Chr.3: 122055493-122055493 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781190589 Chr.3: 122055443-122055443 on Build GRCh38
rs369658926 Chr.3: 122055516-122055516 on Build GRCh38
rs749638145 Chr.3: 122055482-122055482 on Build GRCh38
rs879068118 Chr.3: 122055623-122055623 on Build GRCh38
rs951240338 Chr.3: 122055450-122055450 on Build GRCh38
rs887839539 Chr.3: 122055612-122055612 on Build GRCh38
rs536015475 Chr.3: 122055454-122055454 on Build GRCh38
rs369591256 Chr.3: 122055470-122055470 on Build GRCh38
rs72965464 Chr.3: 122055213-122055213 on Build GRCh38
rs1037699889 Chr.3: 122055586-122055586 on Build GRCh38
rs1017627268 Chr.3: 122055678-122055678 on Build GRCh38
rs993583288 Chr.3: 122055203-122055203 on Build GRCh38
rs368171668 Chr.3: 122055405-122055405 on Build GRCh38
rs552322262 Chr.3: 122055331-122055331 on Build GRCh38
rs1022910257 Chr.3: 122055674-122055674 on Build GRCh38
rs868571613 Chr.3: 122055458-122055458 on Build GRCh38
rs532828793 Chr.3: 122055268-122055268 on Build GRCh38
rs975044185 Chr.3: 122055476-122055476 on Build GRCh38
rs967193193 Chr.3: 122055372-122055372 on Build GRCh38
rs1031354591 Chr.3: 122055417-122055417 on Build GRCh38
rs774402174 Chr.3: 122055411-122055411 on Build GRCh38
rs1018621781 Chr.3: 122055264-122055264 on Build GRCh38
rs373812101 Chr.3: 122055502-122055502 on Build GRCh38
rs774565765 Chr.3: 122055536-122055536 on Build GRCh38
rs942102286 Chr.3: 122055560-122055560 on Build GRCh38
rs538365270 Chr.3: 122055403-122055403 on Build GRCh38
rs569250515 Chr.3: 122055361-122055361 on Build GRCh38
rs914272281 Chr.3: 122055451-122055451 on Build GRCh38
rs199530326 Chr.3: 122055462-122055462 on Build GRCh38
rs976645151 Chr.3: 122055327-122055327 on Build GRCh38
rs767687225 Chr.3: 122055440-122055440 on Build GRCh38
rs1007535442 Chr.3: 122055252-122055252 on Build GRCh38
rs1029540089 Chr.3: 122055371-122055371 on Build GRCh38
rs747745893 Chr.3: 122055501-122055501 on Build GRCh38
rs780505271 Chr.3: 122055473-122055473 on Build GRCh38
rs1049629514 Chr.3: 122055598-122055598 on Build GRCh38
rs538910339 Chr.3: 122055529-122055529 on Build GRCh38
rs765012010 Chr.3: 122055531-122055531 on Build GRCh38
rs1012871902 Chr.3: 122055383-122055383 on Build GRCh38
rs377482392 Chr.3: 122055506-122055506 on Build GRCh38
rs574858206 Chr.3: 122055670-122055670 on Build GRCh38
rs558158767 Chr.3: 122055662-122055662 on Build GRCh38
rs141814734 Chr.3: 122055497-122055497 on Build GRCh38
rs559325317 Chr.3: 122055248-122055248 on Build GRCh38
rs897980841 Chr.3: 122055201-122055201 on Build GRCh38
rs1023218887 Chr.3: 122055385-122055385 on Build GRCh38
rs963984645 Chr.3: 122055311-122055311 on Build GRCh38
rs886534555 Chr.3: 122055247-122055247 on Build GRCh38
rs376177740 Chr.3: 122055511-122055511 on Build GRCh38
rs762008820 Chr.3: 122055547-122055547 on Build GRCh38
rs11575853 Chr.3: 122055434-122055434 on Build GRCh38

View Full Product Details