Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123282218-123282720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY5
Entrez Gene ID
111
Gene Name
adenylate cyclase 5
Gene Aliases
AC5, FDFM
UniGene
Hs.593292
Gene Chromosome Location
Chr.3: 123282296-123448632 on Build GRCh38
Transcripts
NM_001199642.1 NM_183357.2 XM_005247077.3 XM_005247078.1 XM_006713483.1 XM_006713484.1 XM_011512359.2 XM_011512360.2 XM_011512361.1 XM_017005638.1 XM_017005639.1 XM_017005640.1

SNP Detail

SNP ID Chromosome Location
rs137920080 Chr.3: 123282255-123282255 on Build GRCh38
rs6784930 Chr.3: 123282647-123282647 on Build GRCh38
rs894089985 Chr.3: 123282559-123282559 on Build GRCh38
rs528077852 Chr.3: 123282366-123282366 on Build GRCh38
rs566780333 Chr.3: 123282586-123282586 on Build GRCh38
rs747487242 Chr.3: 123282276-123282276 on Build GRCh38
rs890612000 Chr.3: 123282322-123282322 on Build GRCh38
rs920970378 Chr.3: 123282675-123282675 on Build GRCh38
rs145986442 Chr.3: 123282646-123282646 on Build GRCh38
rs941032206 Chr.3: 123282491-123282491 on Build GRCh38
rs547887822 Chr.3: 123282650-123282650 on Build GRCh38
rs80312230 Chr.3: 123282499-123282499 on Build GRCh38
rs755324719 Chr.3: 123282325-123282325 on Build GRCh38
rs1052692333 Chr.3: 123282546-123282546 on Build GRCh38
rs933752699 Chr.3: 123282506-123282506 on Build GRCh38
rs547054063 Chr.3: 123282582-123282582 on Build GRCh38
rs1006018708 Chr.3: 123282517-123282517 on Build GRCh38
rs142457222 Chr.3: 123282379-123282379 on Build GRCh38
rs1010871463 Chr.3: 123282377-123282377 on Build GRCh38
rs113985515 Chr.3: 123282404-123282404 on Build GRCh38
rs949149802 Chr.3: 123282247-123282247 on Build GRCh38
rs943617784 Chr.3: 123282653-123282653 on Build GRCh38
rs776404904 Chr.3: 123282555-123282557 on Build GRCh38
rs1031231427 Chr.3: 123282333-123282333 on Build GRCh38
rs13322080 Chr.3: 123282532-123282532 on Build GRCh38
rs903332932 Chr.3: 123282259-123282259 on Build GRCh38
rs759003230 Chr.3: 123282642-123282642 on Build GRCh38
rs770537664 Chr.3: 123282563-123282563 on Build GRCh38
rs969537021 Chr.3: 123282397-123282397 on Build GRCh38
rs559103938 Chr.3: 123282344-123282344 on Build GRCh38
rs565882366 Chr.3: 123282328-123282328 on Build GRCh38
rs1054565666 Chr.3: 123282321-123282321 on Build GRCh38
rs999673146 Chr.3: 123282542-123282542 on Build GRCh38
rs962698321 Chr.3: 123282648-123282648 on Build GRCh38
rs990768584 Chr.3: 123282562-123282562 on Build GRCh38
rs1039406087 Chr.3: 123282494-123282494 on Build GRCh38
rs923685260 Chr.3: 123282611-123282611 on Build GRCh38
rs760379374 Chr.3: 123282367-123282368 on Build GRCh38
rs549652838 Chr.3: 123282649-123282649 on Build GRCh38
rs978180355 Chr.3: 123282592-123282592 on Build GRCh38
rs951218910 Chr.3: 123282635-123282635 on Build GRCh38
rs866433407 Chr.3: 123282667-123282667 on Build GRCh38
rs113236387 Chr.3: 123282530-123282530 on Build GRCh38
rs561450800 Chr.3: 123282389-123282391 on Build GRCh38
rs934822654 Chr.3: 123282265-123282265 on Build GRCh38
rs1030544006 Chr.3: 123282317-123282317 on Build GRCh38
rs192261002 Chr.3: 123282400-123282400 on Build GRCh38
rs1039283692 Chr.3: 123282696-123282696 on Build GRCh38
rs982141479 Chr.3: 123282420-123282420 on Build GRCh38
rs1003595364 Chr.3: 123282606-123282606 on Build GRCh38
rs909760432 Chr.3: 123282473-123282473 on Build GRCh38
rs927829358 Chr.3: 123282438-123282438 on Build GRCh38
rs1026372994 Chr.3: 123282484-123282485 on Build GRCh38
rs770470582 Chr.3: 123282318-123282318 on Build GRCh38
rs531186924 Chr.3: 123282469-123282469 on Build GRCh38
rs998335959 Chr.3: 123282308-123282308 on Build GRCh38
rs969778494 Chr.3: 123282587-123282587 on Build GRCh38
rs1042169217 Chr.3: 123282258-123282259 on Build GRCh38
rs951189566 Chr.3: 123282466-123282466 on Build GRCh38
rs76135430 Chr.3: 123282616-123282616 on Build GRCh38
rs557671604 Chr.3: 123282634-123282634 on Build GRCh38

View Full Product Details