Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 123295960-123296453 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY5
Entrez Gene ID
111
Gene Name
adenylate cyclase 5
Gene Aliases
AC5, FDFM
UniGene
Hs.593292
Gene Chromosome Location
Chr.3: 123282296-123448632 on Build GRCh38
Transcripts
NM_001199642.1 NM_183357.2 XM_005247077.3 XM_005247078.1 XM_006713483.1 XM_006713484.1 XM_011512359.2 XM_011512360.2 XM_011512361.1 XM_017005638.1 XM_017005639.1 XM_017005640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6162945 c.2997G>A p.L999L Chr.3: 123296150-123296150 on Build GRCh38
5626843 c.2984C>A p.S995* Chr.3: 123296163-123296163 on Build GRCh38
7160744 c.3043G>C p.D1015H Chr.3: 123296104-123296104 on Build GRCh38
4138378 c.2931-1G>C p.? Chr.3: 123296217-123296217 on Build GRCh38
1037311 c.3043G>A p.D1015N Chr.3: 123296104-123296104 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749178622 Chr.3: 123296179-123296179 on Build GRCh38
rs773680641 Chr.3: 123296191-123296191 on Build GRCh38
rs1042617652 Chr.3: 123296316-123296316 on Build GRCh38
rs764637048 Chr.3: 123296249-123296249 on Build GRCh38
rs745615891 Chr.3: 123296170-123296170 on Build GRCh38
rs770248837 Chr.3: 123296224-123296224 on Build GRCh38
rs937113516 Chr.3: 123296004-123296004 on Build GRCh38
rs115200463 Chr.3: 123296248-123296248 on Build GRCh38
rs771812569 Chr.3: 123296199-123296199 on Build GRCh38
rs1034007271 Chr.3: 123296185-123296185 on Build GRCh38
rs116911025 Chr.3: 123296368-123296368 on Build GRCh38
rs9882534 Chr.3: 123296247-123296247 on Build GRCh38
rs1025161900 Chr.3: 123296232-123296232 on Build GRCh38
rs564846157 Chr.3: 123296203-123296203 on Build GRCh38
rs918822267 Chr.3: 123296016-123296016 on Build GRCh38
rs1024085084 Chr.3: 123296424-123296424 on Build GRCh38
rs56322900 Chr.3: 123296053-123296053 on Build GRCh38
rs774471785 Chr.3: 123296281-123296281 on Build GRCh38
rs13079140 Chr.3: 123296220-123296220 on Build GRCh38
rs754808533 Chr.3: 123296264-123296264 on Build GRCh38
rs200579862 Chr.3: 123296251-123296251 on Build GRCh38
rs780951116 Chr.3: 123296048-123296048 on Build GRCh38
rs753118981 Chr.3: 123296137-123296137 on Build GRCh38
rs761362128 Chr.3: 123296105-123296105 on Build GRCh38
rs900987825 Chr.3: 123296090-123296090 on Build GRCh38
rs191382776 Chr.3: 123296362-123296362 on Build GRCh38
rs972981759 Chr.3: 123296015-123296015 on Build GRCh38
rs757362005 Chr.3: 123296165-123296165 on Build GRCh38
rs1007363034 Chr.3: 123296102-123296102 on Build GRCh38
rs56407630 Chr.3: 123296057-123296057 on Build GRCh38
rs145908478 Chr.3: 123296178-123296178 on Build GRCh38
rs9881951 Chr.3: 123296030-123296030 on Build GRCh38
rs141428146 Chr.3: 123296343-123296343 on Build GRCh38
rs549426827 Chr.3: 123296324-123296324 on Build GRCh38
rs751096975 Chr.3: 123296038-123296038 on Build GRCh38
rs768701782 Chr.3: 123296183-123296183 on Build GRCh38
rs374609115 Chr.3: 123296265-123296265 on Build GRCh38
rs768021637 Chr.3: 123296076-123296076 on Build GRCh38
rs918050645 Chr.3: 123295994-123295994 on Build GRCh38
rs370344584 Chr.3: 123296266-123296266 on Build GRCh38
rs760450179 Chr.3: 123296208-123296208 on Build GRCh38
rs1048875178 Chr.3: 123296039-123296039 on Build GRCh38
rs571005355 Chr.3: 123296206-123296206 on Build GRCh38
rs894118531 Chr.3: 123296414-123296414 on Build GRCh38
rs879938595 Chr.3: 123295992-123295992 on Build GRCh38
rs749298665 Chr.3: 123296068-123296068 on Build GRCh38
rs781396013 Chr.3: 123296166-123296166 on Build GRCh38
rs755882167 Chr.3: 123296177-123296177 on Build GRCh38
rs767294732 Chr.3: 123296109-123296109 on Build GRCh38
rs531784183 Chr.3: 123296242-123296242 on Build GRCh38
rs142086014 Chr.3: 123296135-123296135 on Build GRCh38
rs554029464 Chr.3: 123296367-123296367 on Build GRCh38
rs533884895 Chr.3: 123296361-123296361 on Build GRCh38
rs753685469 Chr.3: 123296254-123296254 on Build GRCh38
rs545626654 Chr.3: 123295993-123295993 on Build GRCh38
rs757955112 Chr.3: 123295998-123295998 on Build GRCh38
rs764076808 Chr.3: 123296156-123296156 on Build GRCh38
rs992947151 Chr.3: 123296276-123296276 on Build GRCh38
rs991020410 Chr.3: 123295985-123295985 on Build GRCh38
rs9844230 Chr.3: 123296066-123296066 on Build GRCh38
rs1013916001 Chr.3: 123296417-123296417 on Build GRCh38
rs917012403 Chr.3: 123296278-123296278 on Build GRCh38
rs763355875 Chr.3: 123296231-123296231 on Build GRCh38
rs772074478 Chr.3: 123296366-123296366 on Build GRCh38
rs949523618 Chr.3: 123296287-123296287 on Build GRCh38
rs535925010 Chr.3: 123296321-123296321 on Build GRCh38
rs984096883 Chr.3: 123296255-123296255 on Build GRCh38
rs926921515 Chr.3: 123296252-123296252 on Build GRCh38
rs758980207 Chr.3: 123296140-123296140 on Build GRCh38
rs775985458 Chr.3: 123296225-123296225 on Build GRCh38
rs751717928 Chr.3: 123296159-123296159 on Build GRCh38
rs764674688 Chr.3: 123296239-123296239 on Build GRCh38
rs901306925 Chr.3: 123296428-123296428 on Build GRCh38
rs747701438 Chr.3: 123296198-123296198 on Build GRCh38
rs866033544 Chr.3: 123296275-123296275 on Build GRCh38
rs761061698 Chr.3: 123296386-123296386 on Build GRCh38
rs113525130 Chr.3: 123296132-123296132 on Build GRCh38
rs779701154 Chr.3: 123296253-123296253 on Build GRCh38
rs201365334 Chr.3: 123296250-123296250 on Build GRCh38
rs376634525 Chr.3: 123296096-123296096 on Build GRCh38
rs777071342 Chr.3: 123296123-123296123 on Build GRCh38

View Full Product Details