Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129555091-129555596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H1FOO
Entrez Gene ID
132243
Gene Name
H1 histone family member O, oocyte specific
Gene Aliases
H1.8, H1oo, osH1
UniGene
Hs.97358
Gene Chromosome Location
Chr.3: 129543214-129555185 on Build GRCh38
Transcripts
XM_017005733.1 NM_015103.2 XM_011512589.1 XM_011512592.1

Gene Symbol
PLXND1
Entrez Gene ID
23129
Gene Name
plexin D1
Gene Aliases
PLEXD1
UniGene
Hs.301685
Gene Chromosome Location
Chr.3: 129555213-129606739 on Build GRCh38
Transcripts
XM_017005733.1 NM_015103.2 XM_011512589.1 XM_011512592.1

SNP Detail

SNP ID Chromosome Location
rs929155573 Chr.3: 129555317-129555326 on Build GRCh38
rs763974908 Chr.3: 129555477-129555477 on Build GRCh38
rs563784696 Chr.3: 129555303-129555303 on Build GRCh38
rs529561303 Chr.3: 129555421-129555421 on Build GRCh38
rs932352358 Chr.3: 129555439-129555439 on Build GRCh38
rs372921867 Chr.3: 129555349-129555350 on Build GRCh38
rs1052731140 Chr.3: 129555466-129555466 on Build GRCh38
rs1040781112 Chr.3: 129555214-129555214 on Build GRCh38
rs1006902211 Chr.3: 129555496-129555496 on Build GRCh38
rs947743633 Chr.3: 129555414-129555414 on Build GRCh38
rs574603526 Chr.3: 129555197-129555197 on Build GRCh38
rs545444461 Chr.3: 129555297-129555297 on Build GRCh38
rs991826213 Chr.3: 129555395-129555395 on Build GRCh38
rs182162812 Chr.3: 129555471-129555471 on Build GRCh38
rs764604078 Chr.3: 129555480-129555480 on Build GRCh38
rs1048337244 Chr.3: 129555161-129555161 on Build GRCh38
rs113891900 Chr.3: 129555126-129555126 on Build GRCh38
rs541379324 Chr.3: 129555121-129555121 on Build GRCh38
rs190445996 Chr.3: 129555117-129555117 on Build GRCh38
rs906581039 Chr.3: 129555565-129555566 on Build GRCh38
rs1026261367 Chr.3: 129555560-129555560 on Build GRCh38
rs766948849 Chr.3: 129555315-129555315 on Build GRCh38
rs112023993 Chr.3: 129555357-129555357 on Build GRCh38
rs934091628 Chr.3: 129555475-129555475 on Build GRCh38
rs757891693 Chr.3: 129555331-129555331 on Build GRCh38
rs569429461 Chr.3: 129555478-129555478 on Build GRCh38
rs1007124442 Chr.3: 129555332-129555332 on Build GRCh38
rs1009828684 Chr.3: 129555559-129555559 on Build GRCh38
rs948002276 Chr.3: 129555247-129555247 on Build GRCh38
rs745618643 Chr.3: 129555548-129555548 on Build GRCh38
rs892679856 Chr.3: 129555549-129555550 on Build GRCh38
rs1043524331 Chr.3: 129555262-129555262 on Build GRCh38
rs1001117159 Chr.3: 129555286-129555286 on Build GRCh38
rs79523423 Chr.3: 129555225-129555225 on Build GRCh38
rs529728173 Chr.3: 129555547-129555548 on Build GRCh38
rs972447695 Chr.3: 129555147-129555147 on Build GRCh38
rs962836355 Chr.3: 129555355-129555355 on Build GRCh38
rs908478731 Chr.3: 129555404-129555404 on Build GRCh38
rs112382780 Chr.3: 129555372-129555372 on Build GRCh38
rs1052910 Chr.3: 129555520-129555520 on Build GRCh38
rs568635585 Chr.3: 129555394-129555394 on Build GRCh38
rs541680693 Chr.3: 129555224-129555224 on Build GRCh38
rs925212375 Chr.3: 129555419-129555419 on Build GRCh38
rs891913156 Chr.3: 129555274-129555274 on Build GRCh38
rs145262581 Chr.3: 129555120-129555120 on Build GRCh38
rs746693895 Chr.3: 129555417-129555417 on Build GRCh38
rs558188852 Chr.3: 129555118-129555118 on Build GRCh38
rs916256581 Chr.3: 129555403-129555403 on Build GRCh38
rs749931793 Chr.3: 129555269-129555269 on Build GRCh38
rs775450317 Chr.3: 129555469-129555469 on Build GRCh38
rs575668143 Chr.3: 129555113-129555113 on Build GRCh38
rs529477028 Chr.3: 129555277-129555277 on Build GRCh38
rs762475207 Chr.3: 129555343-129555343 on Build GRCh38
rs1030102642 Chr.3: 129555287-129555287 on Build GRCh38
rs530530194 Chr.3: 129555268-129555268 on Build GRCh38
rs750024224 Chr.3: 129555525-129555525 on Build GRCh38
rs549207860 Chr.3: 129555322-129555322 on Build GRCh38
rs866189419 Chr.3: 129555178-129555178 on Build GRCh38
rs370213668 Chr.3: 129555495-129555495 on Build GRCh38
rs569209264 Chr.3: 129555465-129555465 on Build GRCh38
rs938992091 Chr.3: 129555204-129555204 on Build GRCh38
rs926454275 Chr.3: 129555377-129555377 on Build GRCh38
rs922301923 Chr.3: 129555422-129555422 on Build GRCh38
rs960774051 Chr.3: 129555373-129555373 on Build GRCh38
rs1053177860 Chr.3: 129555271-129555271 on Build GRCh38
rs575924570 Chr.3: 129555524-129555524 on Build GRCh38
rs866113650 Chr.3: 129555281-129555281 on Build GRCh38

View Full Product Details