Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130934215-130934720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2C1
Entrez Gene ID
27032
Gene Name
ATPase secretory pathway Ca2+ transporting 1
Gene Aliases
ATP2C1A, BCPM, HHD, PMR1, SPCA1, hSPCA1
UniGene
Hs.584884
Gene Chromosome Location
Chr.3: 130850500-131016712 on Build GRCh38
Transcripts
NM_001001485.2 NM_001001486.1 NM_001001487.1 NM_001199179.1 NM_001199180.1 NM_001199181.1 NM_001199182.1 NM_001199183.1 NM_001199184.1 NM_001199185.1 NM_014382.3 XM_005247354.2 XM_005247355.2 XM_005247356.2 XM_005247358.2 XM_017006164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3846216 c.257T>C p.L86P Chr.3: 130934644-130934644 on Build GRCh38
4404465 c.309C>T p.A103A Chr.3: 130934696-130934696 on Build GRCh38
6607868 c.424G>A p.V142M Chr.3: 130934709-130934709 on Build GRCh38
4404464 c.411C>T p.A137A Chr.3: 130934696-130934696 on Build GRCh38
6607840 c.310G>A p.V104I Chr.3: 130934697-130934697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926921690 Chr.3: 130934477-130934477 on Build GRCh38
rs76156891 Chr.3: 130934557-130934557 on Build GRCh38
rs1045786877 Chr.3: 130934243-130934243 on Build GRCh38
rs528067816 Chr.3: 130934461-130934461 on Build GRCh38
rs539674450 Chr.3: 130934580-130934580 on Build GRCh38
rs750648361 Chr.3: 130934571-130934571 on Build GRCh38
rs569007329 Chr.3: 130934534-130934534 on Build GRCh38
rs559081037 Chr.3: 130934696-130934696 on Build GRCh38
rs907776593 Chr.3: 130934508-130934508 on Build GRCh38
rs778659106 Chr.3: 130934585-130934585 on Build GRCh38
rs911570257 Chr.3: 130934405-130934405 on Build GRCh38
rs930485455 Chr.3: 130934465-130934465 on Build GRCh38
rs773030549 Chr.3: 130934631-130934631 on Build GRCh38
rs886299310 Chr.3: 130934282-130934284 on Build GRCh38
rs142250638 Chr.3: 130934528-130934528 on Build GRCh38
rs1004699848 Chr.3: 130934310-130934310 on Build GRCh38
rs992285744 Chr.3: 130934426-130934426 on Build GRCh38
rs16835390 Chr.3: 130934245-130934245 on Build GRCh38
rs893248094 Chr.3: 130934599-130934599 on Build GRCh38
rs139944706 Chr.3: 130934506-130934506 on Build GRCh38
rs56864440 Chr.3: 130934457-130934457 on Build GRCh38
rs768588962 Chr.3: 130934581-130934581 on Build GRCh38
rs34266677 Chr.3: 130934548-130934549 on Build GRCh38
rs374413602 Chr.3: 130934256-130934256 on Build GRCh38
rs957565644 Chr.3: 130934317-130934317 on Build GRCh38
rs940618557 Chr.3: 130934513-130934513 on Build GRCh38
rs535701338 Chr.3: 130934507-130934507 on Build GRCh38
rs996187189 Chr.3: 130934360-130934360 on Build GRCh38
rs751063866 Chr.3: 130934459-130934459 on Build GRCh38
rs762778075 Chr.3: 130934257-130934257 on Build GRCh38
rs1012190037 Chr.3: 130934341-130934341 on Build GRCh38
rs747998104 Chr.3: 130934591-130934591 on Build GRCh38
rs780149863 Chr.3: 130934286-130934288 on Build GRCh38
rs767093129 Chr.3: 130934540-130934540 on Build GRCh38
rs977712688 Chr.3: 130934396-130934396 on Build GRCh38
rs1004884952 Chr.3: 130934242-130934242 on Build GRCh38
rs907223608 Chr.3: 130934567-130934567 on Build GRCh38
rs397877386 Chr.3: 130934554-130934555 on Build GRCh38
rs967626027 Chr.3: 130934355-130934355 on Build GRCh38
rs534628809 Chr.3: 130934467-130934467 on Build GRCh38
rs746716018 Chr.3: 130934693-130934693 on Build GRCh38
rs551827258 Chr.3: 130934421-130934421 on Build GRCh38
rs1029369387 Chr.3: 130934380-130934380 on Build GRCh38
rs568171743 Chr.3: 130934619-130934619 on Build GRCh38
rs76226805 Chr.3: 130934558-130934558 on Build GRCh38
rs1050464154 Chr.3: 130934604-130934604 on Build GRCh38
rs985134692 Chr.3: 130934525-130934525 on Build GRCh38
rs756309330 Chr.3: 130934577-130934577 on Build GRCh38
rs79613660 Chr.3: 130934554-130934554 on Build GRCh38
rs940538686 Chr.3: 130934543-130934543 on Build GRCh38
rs112314930 Chr.3: 130934411-130934411 on Build GRCh38
rs35022847 Chr.3: 130934268-130934268 on Build GRCh38
rs1020529919 Chr.3: 130934347-130934347 on Build GRCh38
rs71333682 Chr.3: 130934505-130934505 on Build GRCh38
rs148794392 Chr.3: 130934407-130934408 on Build GRCh38
rs771725725 Chr.3: 130934618-130934618 on Build GRCh38
rs1011726157 Chr.3: 130934615-130934615 on Build GRCh38

View Full Product Details