Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134360015-134360507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMOTL2
Entrez Gene ID
51421
Gene Name
angiomotin like 2
Gene Aliases
LCCP
UniGene
Hs.426312
Gene Chromosome Location
Chr.3: 134355345-134375417 on Build GRCh38
Transcripts
NM_001278683.1 NM_001278685.1 NM_016201.3 XM_006713654.2 XM_011512881.1 XM_017006580.1 XM_017006581.1 XM_017006582.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661420 c.1805C>T p.S602F Chr.3: 134360184-134360184 on Build GRCh38
3974176 c.1593A>C p.P531P Chr.3: 134360396-134360396 on Build GRCh38
3992752 c.1836T>G p.N612K Chr.3: 134360153-134360153 on Build GRCh38
3122646 c.1730G>A p.R577H Chr.3: 134360259-134360259 on Build GRCh38
6599355 c.1683G>A p.A561A Chr.3: 134360306-134360306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199600324 Chr.3: 134360402-134360402 on Build GRCh38
rs770056566 Chr.3: 134360229-134360229 on Build GRCh38
rs773202954 Chr.3: 134360067-134360067 on Build GRCh38
rs771059774 Chr.3: 134360071-134360071 on Build GRCh38
rs765712043 Chr.3: 134360195-134360195 on Build GRCh38
rs758990000 Chr.3: 134360297-134360297 on Build GRCh38
rs140347461 Chr.3: 134360255-134360255 on Build GRCh38
rs908045586 Chr.3: 134360183-134360183 on Build GRCh38
rs372354405 Chr.3: 134360226-134360227 on Build GRCh38
rs777933631 Chr.3: 134360425-134360426 on Build GRCh38
rs768070890 Chr.3: 134360447-134360447 on Build GRCh38
rs142861055 Chr.3: 134360382-134360382 on Build GRCh38
rs371556971 Chr.3: 134360338-134360338 on Build GRCh38
rs151045996 Chr.3: 134360177-134360177 on Build GRCh38
rs748357501 Chr.3: 134360225-134360233 on Build GRCh38
rs137933371 Chr.3: 134360356-134360356 on Build GRCh38
rs775857485 Chr.3: 134360178-134360178 on Build GRCh38
rs369922508 Chr.3: 134360057-134360057 on Build GRCh38
rs780877639 Chr.3: 134360117-134360117 on Build GRCh38
rs780246478 Chr.3: 134360343-134360343 on Build GRCh38
rs756549466 Chr.3: 134360306-134360306 on Build GRCh38
rs762907607 Chr.3: 134360464-134360464 on Build GRCh38
rs776959994 Chr.3: 134360191-134360191 on Build GRCh38
rs757200684 Chr.3: 134360228-134360228 on Build GRCh38
rs760293974 Chr.3: 134360312-134360312 on Build GRCh38
rs751684213 Chr.3: 134360401-134360401 on Build GRCh38
rs367916829 Chr.3: 134360072-134360072 on Build GRCh38
rs779335973 Chr.3: 134360484-134360484 on Build GRCh38
rs35938212 Chr.3: 134360367-134360367 on Build GRCh38
rs777633596 Chr.3: 134360150-134360150 on Build GRCh38
rs764985432 Chr.3: 134360458-134360458 on Build GRCh38
rs771467653 Chr.3: 134360357-134360357 on Build GRCh38
rs570965109 Chr.3: 134360391-134360391 on Build GRCh38
rs766621996 Chr.3: 134360349-134360349 on Build GRCh38
rs781603704 Chr.3: 134360068-134360068 on Build GRCh38
rs192757705 Chr.3: 134360148-134360148 on Build GRCh38
rs745957994 Chr.3: 134360232-134360232 on Build GRCh38
rs781416079 Chr.3: 134360407-134360407 on Build GRCh38
rs896175583 Chr.3: 134360294-134360294 on Build GRCh38
rs747135063 Chr.3: 134360345-134360345 on Build GRCh38
rs923450390 Chr.3: 134360139-134360139 on Build GRCh38
rs562821811 Chr.3: 134360152-134360152 on Build GRCh38
rs570339300 Chr.3: 134360421-134360421 on Build GRCh38
rs750280687 Chr.3: 134360396-134360396 on Build GRCh38
rs780371170 Chr.3: 134360237-134360237 on Build GRCh38
rs753620282 Chr.3: 134360224-134360224 on Build GRCh38
rs145301642 Chr.3: 134360260-134360260 on Build GRCh38
rs149751650 Chr.3: 134360200-134360200 on Build GRCh38
rs974746598 Chr.3: 134360085-134360085 on Build GRCh38
rs772288782 Chr.3: 134360236-134360236 on Build GRCh38
rs547949613 Chr.3: 134360352-134360352 on Build GRCh38
rs766482164 Chr.3: 134360400-134360400 on Build GRCh38
rs148948420 Chr.3: 134360039-134360039 on Build GRCh38
rs753439957 Chr.3: 134360386-134360386 on Build GRCh38
rs761182369 Chr.3: 134360092-134360092 on Build GRCh38
rs755855347 Chr.3: 134360216-134360216 on Build GRCh38
rs770877034 Chr.3: 134360153-134360153 on Build GRCh38
rs757068719 Chr.3: 134360325-134360325 on Build GRCh38
rs774962408 Chr.3: 134360360-134360360 on Build GRCh38
rs147176018 Chr.3: 134360074-134360075 on Build GRCh38
rs754633628 Chr.3: 134360107-134360107 on Build GRCh38
rs752306623 Chr.3: 134360307-134360307 on Build GRCh38
rs762140866 Chr.3: 134360259-134360259 on Build GRCh38
rs550324555 Chr.3: 134360209-134360209 on Build GRCh38
rs764970176 Chr.3: 134360393-134360393 on Build GRCh38
rs754408398 Chr.3: 134360101-134360101 on Build GRCh38
rs755463014 Chr.3: 134360451-134360451 on Build GRCh38
rs752361624 Chr.3: 134360211-134360211 on Build GRCh38
rs753657091 Chr.3: 134360314-134360314 on Build GRCh38
rs140871979 Chr.3: 134360186-134360186 on Build GRCh38
rs764628027 Chr.3: 134360097-134360097 on Build GRCh38
rs184398270 Chr.3: 134360437-134360437 on Build GRCh38
rs530571349 Chr.3: 134360406-134360406 on Build GRCh38
rs371941299 Chr.3: 134360154-134360154 on Build GRCh38
rs983480047 Chr.3: 134360479-134360479 on Build GRCh38
rs747935308 Chr.3: 134360127-134360127 on Build GRCh38
rs376869456 Chr.3: 134360246-134360246 on Build GRCh38
rs755183683 Chr.3: 134360067-134360068 on Build GRCh38
rs758384203 Chr.3: 134360399-134360399 on Build GRCh38
rs201755719 Chr.3: 134360169-134360169 on Build GRCh38
rs547887711 Chr.3: 134360208-134360208 on Build GRCh38
rs565023057 Chr.3: 134360192-134360192 on Build GRCh38
rs371137555 Chr.3: 134360069-134360069 on Build GRCh38
rs769036048 Chr.3: 134360249-134360249 on Build GRCh38
rs376883450 Chr.3: 134360231-134360231 on Build GRCh38
rs2241558 Chr.3: 134360222-134360222 on Build GRCh38
rs781335285 Chr.3: 134360350-134360350 on Build GRCh38
rs200489847 Chr.3: 134360073-134360073 on Build GRCh38
rs773127658 Chr.3: 134360459-134360459 on Build GRCh38
rs750405650 Chr.3: 134360332-134360332 on Build GRCh38
rs751067676 Chr.3: 134360103-134360103 on Build GRCh38
rs937594727 Chr.3: 134360250-134360250 on Build GRCh38
rs1030072155 Chr.3: 134360472-134360472 on Build GRCh38
rs777969756 Chr.3: 134360435-134360435 on Build GRCh38
rs761337309 Chr.3: 134360454-134360454 on Build GRCh38
rs561578417 Chr.3: 134360331-134360331 on Build GRCh38
rs923266542 Chr.3: 134360095-134360095 on Build GRCh38
rs573334002 Chr.3: 134360102-134360102 on Build GRCh38
rs147624210 Chr.3: 134360436-134360436 on Build GRCh38
rs756461602 Chr.3: 134360422-134360422 on Build GRCh38

View Full Product Details