Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138124203-138124736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A4GNT
Entrez Gene ID
51146
Gene Name
alpha-1,4-N-acetylglucosaminyltransferase
Gene Aliases
alpha4GnT
UniGene
Hs.278960
Gene Chromosome Location
Chr.3: 138123718-138133310 on Build GRCh38
Transcripts
NM_016161.2 XM_017006543.1 XM_017006544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6163534 c.671G>T p.G224V Chr.3: 138124616-138124616 on Build GRCh38
4657824 c.707G>A p.C236Y Chr.3: 138124580-138124580 on Build GRCh38
6163536 c.954G>T p.R318S Chr.3: 138124333-138124333 on Build GRCh38
3587935 c.963G>A p.R321R Chr.3: 138124324-138124324 on Build GRCh38
7447436 c.713T>C p.L238P Chr.3: 138124574-138124574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142287900 Chr.3: 138124521-138124521 on Build GRCh38
rs781246221 Chr.3: 138124620-138124620 on Build GRCh38
rs757747703 Chr.3: 138124247-138124247 on Build GRCh38
rs752287411 Chr.3: 138124536-138124536 on Build GRCh38
rs892441560 Chr.3: 138124287-138124287 on Build GRCh38
rs113881039 Chr.3: 138124491-138124491 on Build GRCh38
rs371977803 Chr.3: 138124399-138124399 on Build GRCh38
rs774666633 Chr.3: 138124652-138124652 on Build GRCh38
rs777006950 Chr.3: 138124423-138124423 on Build GRCh38
rs763565730 Chr.3: 138124531-138124531 on Build GRCh38
rs777531048 Chr.3: 138124472-138124472 on Build GRCh38
rs76043052 Chr.3: 138124404-138124404 on Build GRCh38
rs369495396 Chr.3: 138124525-138124525 on Build GRCh38
rs749300342 Chr.3: 138124318-138124318 on Build GRCh38
rs762168578 Chr.3: 138124517-138124517 on Build GRCh38
rs558347002 Chr.3: 138124496-138124496 on Build GRCh38
rs149488891 Chr.3: 138124695-138124695 on Build GRCh38
rs533935341 Chr.3: 138124365-138124365 on Build GRCh38
rs774383385 Chr.3: 138124395-138124395 on Build GRCh38
rs953566772 Chr.3: 138124340-138124340 on Build GRCh38
rs759650530 Chr.3: 138124325-138124325 on Build GRCh38
rs764265840 Chr.3: 138124422-138124422 on Build GRCh38
rs372771000 Chr.3: 138124236-138124236 on Build GRCh38
rs868624165 Chr.3: 138124451-138124451 on Build GRCh38
rs146581075 Chr.3: 138124352-138124352 on Build GRCh38
rs563180536 Chr.3: 138124699-138124699 on Build GRCh38
rs766765241 Chr.3: 138124533-138124533 on Build GRCh38
rs780846256 Chr.3: 138124283-138124283 on Build GRCh38
rs778897391 Chr.3: 138124579-138124579 on Build GRCh38
rs775713265 Chr.3: 138124349-138124349 on Build GRCh38
rs375179797 Chr.3: 138124444-138124444 on Build GRCh38
rs974466026 Chr.3: 138124465-138124465 on Build GRCh38
rs1004739394 Chr.3: 138124659-138124659 on Build GRCh38
rs764577493 Chr.3: 138124351-138124351 on Build GRCh38
rs1031037973 Chr.3: 138124265-138124265 on Build GRCh38
rs745937519 Chr.3: 138124397-138124397 on Build GRCh38
rs1016010634 Chr.3: 138124680-138124680 on Build GRCh38
rs143849375 Chr.3: 138124330-138124330 on Build GRCh38
rs768654443 Chr.3: 138124612-138124612 on Build GRCh38
rs776256364 Chr.3: 138124672-138124672 on Build GRCh38
rs764560674 Chr.3: 138124313-138124313 on Build GRCh38
rs751089246 Chr.3: 138124368-138124368 on Build GRCh38
rs984052061 Chr.3: 138124428-138124428 on Build GRCh38
rs553089322 Chr.3: 138124551-138124551 on Build GRCh38
rs983751562 Chr.3: 138124692-138124692 on Build GRCh38
rs137971144 Chr.3: 138124682-138124682 on Build GRCh38
rs1035488344 Chr.3: 138124440-138124440 on Build GRCh38
rs61740810 Chr.3: 138124441-138124441 on Build GRCh38
rs201500783 Chr.3: 138124541-138124541 on Build GRCh38
rs570351376 Chr.3: 138124478-138124478 on Build GRCh38
rs773808328 Chr.3: 138124676-138124676 on Build GRCh38
rs151042505 Chr.3: 138124476-138124476 on Build GRCh38
rs780715163 Chr.3: 138124372-138124372 on Build GRCh38
rs755712762 Chr.3: 138124382-138124382 on Build GRCh38
rs750254800 Chr.3: 138124683-138124683 on Build GRCh38
rs6794722 Chr.3: 138124252-138124252 on Build GRCh38
rs760058289 Chr.3: 138124539-138124539 on Build GRCh38
rs777813994 Chr.3: 138124709-138124709 on Build GRCh38
rs770447419 Chr.3: 138124511-138124512 on Build GRCh38
rs748875746 Chr.3: 138124426-138124426 on Build GRCh38
rs748428271 Chr.3: 138124630-138124630 on Build GRCh38
rs906582929 Chr.3: 138124261-138124261 on Build GRCh38
rs758069750 Chr.3: 138124684-138124684 on Build GRCh38
rs761593650 Chr.3: 138124681-138124681 on Build GRCh38
rs763191886 Chr.3: 138124640-138124640 on Build GRCh38
rs747910981 Chr.3: 138124309-138124309 on Build GRCh38
rs762115616 Chr.3: 138124432-138124432 on Build GRCh38
rs755834975 Chr.3: 138124471-138124471 on Build GRCh38
rs776775041 Chr.3: 138124503-138124503 on Build GRCh38
rs201383957 Chr.3: 138124654-138124654 on Build GRCh38
rs766342848 Chr.3: 138124687-138124687 on Build GRCh38
rs767113376 Chr.3: 138124448-138124448 on Build GRCh38
rs753037101 Chr.3: 138124227-138124227 on Build GRCh38
rs369687841 Chr.3: 138124392-138124392 on Build GRCh38
rs2246945 Chr.3: 138124634-138124634 on Build GRCh38
rs1045110452 Chr.3: 138124534-138124534 on Build GRCh38
rs200109290 Chr.3: 138124398-138124398 on Build GRCh38
rs1052933001 Chr.3: 138124490-138124490 on Build GRCh38
rs771025283 Chr.3: 138124394-138124394 on Build GRCh38
rs1007758052 Chr.3: 138124601-138124601 on Build GRCh38
rs540681277 Chr.3: 138124608-138124608 on Build GRCh38
rs763752728 Chr.3: 138124664-138124664 on Build GRCh38
rs889333352 Chr.3: 138124591-138124591 on Build GRCh38
rs774614227 Chr.3: 138124324-138124324 on Build GRCh38
rs747124106 Chr.3: 138124611-138124611 on Build GRCh38
rs922172316 Chr.3: 138124475-138124475 on Build GRCh38
rs755608685 Chr.3: 138124498-138124498 on Build GRCh38
rs770970333 Chr.3: 138124323-138124323 on Build GRCh38
rs764244115 Chr.3: 138124370-138124370 on Build GRCh38
rs554487175 Chr.3: 138124371-138124371 on Build GRCh38
rs146992672 Chr.3: 138124645-138124645 on Build GRCh38
rs747903977 Chr.3: 138124377-138124377 on Build GRCh38
rs757796942 Chr.3: 138124362-138124362 on Build GRCh38
rs777760014 Chr.3: 138124386-138124386 on Build GRCh38
rs756953597 Chr.3: 138124552-138124552 on Build GRCh38
rs1022472732 Chr.3: 138124333-138124333 on Build GRCh38
rs1002758031 Chr.3: 138124622-138124622 on Build GRCh38
rs538771809 Chr.3: 138124493-138124493 on Build GRCh38
rs750864199 Chr.3: 138124442-138124442 on Build GRCh38
rs769568570 Chr.3: 138124310-138124310 on Build GRCh38
rs758391584 Chr.3: 138124598-138124598 on Build GRCh38
rs750149302 Chr.3: 138124589-138124589 on Build GRCh38
rs760801510 Chr.3: 138124350-138124350 on Build GRCh38
rs754408747 Chr.3: 138124239-138124239 on Build GRCh38
rs753610351 Chr.3: 138124543-138124543 on Build GRCh38
rs1051973525 Chr.3: 138124262-138124262 on Build GRCh38
rs2170309 Chr.3: 138124264-138124264 on Build GRCh38
rs745872612 Chr.3: 138124479-138124479 on Build GRCh38
rs771878705 Chr.3: 138124484-138124484 on Build GRCh38
rs143852911 Chr.3: 138124704-138124704 on Build GRCh38

View Full Product Details