Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138619629-138620097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAIM
Entrez Gene ID
55179
Gene Name
Fas apoptotic inhibitory molecule
Gene Aliases
FAIM1
UniGene
Hs.173438
Gene Chromosome Location
Chr.3: 138608163-138633376 on Build GRCh38
Transcripts
NM_001033030.1 NM_001033031.1 NM_001033032.1 NM_018147.3 XM_011512950.2

SNP Detail

SNP ID Chromosome Location
rs772731493 Chr.3: 138619777-138619777 on Build GRCh38
rs990754079 Chr.3: 138619838-138619838 on Build GRCh38
rs767433804 Chr.3: 138619820-138619820 on Build GRCh38
rs996332195 Chr.3: 138619988-138619988 on Build GRCh38
rs932799391 Chr.3: 138619909-138619909 on Build GRCh38
rs145207138 Chr.3: 138619918-138619918 on Build GRCh38
rs927903064 Chr.3: 138620069-138620069 on Build GRCh38
rs1049388998 Chr.3: 138620057-138620057 on Build GRCh38
rs755794690 Chr.3: 138619715-138619715 on Build GRCh38
rs752132304 Chr.3: 138620017-138620017 on Build GRCh38
rs921414653 Chr.3: 138619852-138619852 on Build GRCh38
rs571505482 Chr.3: 138619654-138619654 on Build GRCh38
rs368433430 Chr.3: 138619671-138619671 on Build GRCh38
rs535723683 Chr.3: 138619764-138619764 on Build GRCh38
rs971064478 Chr.3: 138619657-138619657 on Build GRCh38
rs772543081 Chr.3: 138619729-138619729 on Build GRCh38
rs749343681 Chr.3: 138619692-138619692 on Build GRCh38
rs774117690 Chr.3: 138619796-138619796 on Build GRCh38
rs746049347 Chr.3: 138619725-138619725 on Build GRCh38
rs779257835 Chr.3: 138619758-138619758 on Build GRCh38
rs746072831 Chr.3: 138619767-138619772 on Build GRCh38
rs148995487 Chr.3: 138620034-138620034 on Build GRCh38
rs199822647 Chr.3: 138619746-138619746 on Build GRCh38
rs762554134 Chr.3: 138619699-138619699 on Build GRCh38
rs866048180 Chr.3: 138619724-138619724 on Build GRCh38
rs764636307 Chr.3: 138620005-138620005 on Build GRCh38
rs146881980 Chr.3: 138619937-138619937 on Build GRCh38
rs66470363 Chr.3: 138620074-138620074 on Build GRCh38
rs779247481 Chr.3: 138619707-138619707 on Build GRCh38
rs969219882 Chr.3: 138620050-138620050 on Build GRCh38
rs578012912 Chr.3: 138619996-138619996 on Build GRCh38
rs956475270 Chr.3: 138620026-138620026 on Build GRCh38
rs1012014718 Chr.3: 138619693-138619693 on Build GRCh38
rs569224502 Chr.3: 138619845-138619845 on Build GRCh38
rs762572898 Chr.3: 138619787-138619787 on Build GRCh38
rs912782417 Chr.3: 138619919-138619919 on Build GRCh38
rs778005257 Chr.3: 138619690-138619690 on Build GRCh38
rs759099314 Chr.3: 138619814-138619814 on Build GRCh38
rs1672935 Chr.3: 138620073-138620073 on Build GRCh38
rs145680493 Chr.3: 138619739-138619739 on Build GRCh38
rs765773567 Chr.3: 138619795-138619795 on Build GRCh38
rs552371549 Chr.3: 138620016-138620016 on Build GRCh38
rs977080151 Chr.3: 138619689-138619689 on Build GRCh38
rs980625075 Chr.3: 138620059-138620059 on Build GRCh38
rs185858212 Chr.3: 138619839-138619839 on Build GRCh38
rs756059216 Chr.3: 138619682-138619682 on Build GRCh38
rs899303365 Chr.3: 138619973-138619973 on Build GRCh38
rs954976530 Chr.3: 138620007-138620007 on Build GRCh38
rs757447487 Chr.3: 138619700-138619700 on Build GRCh38
rs987153727 Chr.3: 138619655-138619655 on Build GRCh38
rs113179945 Chr.3: 138619706-138619706 on Build GRCh38
rs927882853 Chr.3: 138619895-138619895 on Build GRCh38
rs1037035032 Chr.3: 138619963-138619963 on Build GRCh38

View Full Product Details