Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110061348-110061853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALX3
Entrez Gene ID
257
Gene Name
ALX homeobox 3
Gene Aliases
FND, FND1
UniGene
Hs.669953
Gene Chromosome Location
Chr.1: 110060375-110070700 on Build GRCh38
Transcripts
NM_006492.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3471022 c.659G>A p.G220E Chr.1: 110061499-110061499 on Build GRCh38
4391950 c.632G>A p.R211H Chr.1: 110061526-110061526 on Build GRCh38
2118364 c.704G>A p.R235H Chr.1: 110061454-110061454 on Build GRCh38
3984068 c.669C>A p.P223P Chr.1: 110061489-110061489 on Build GRCh38
893616 c.674C>T p.T225M Chr.1: 110061484-110061484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs909367938 Chr.1: 110061780-110061780 on Build GRCh38
rs753136806 Chr.1: 110061593-110061593 on Build GRCh38
rs771081739 Chr.1: 110061472-110061472 on Build GRCh38
rs942298501 Chr.1: 110061785-110061785 on Build GRCh38
rs765747640 Chr.1: 110061497-110061497 on Build GRCh38
rs1013439707 Chr.1: 110061761-110061761 on Build GRCh38
rs763479585 Chr.1: 110061411-110061411 on Build GRCh38
rs897853201 Chr.1: 110061626-110061626 on Build GRCh38
rs778041172 Chr.1: 110061464-110061464 on Build GRCh38
rs773879847 Chr.1: 110061489-110061489 on Build GRCh38
rs375503292 Chr.1: 110061496-110061496 on Build GRCh38
rs538430669 Chr.1: 110061493-110061493 on Build GRCh38
rs757293052 Chr.1: 110061526-110061526 on Build GRCh38
rs139935098 Chr.1: 110061405-110061405 on Build GRCh38
rs761076201 Chr.1: 110061413-110061414 on Build GRCh38
rs199801560 Chr.1: 110061469-110061469 on Build GRCh38
rs121908166 Chr.1: 110061550-110061550 on Build GRCh38
rs374651713 Chr.1: 110061547-110061547 on Build GRCh38
rs12749726 Chr.1: 110061458-110061458 on Build GRCh38
rs776854762 Chr.1: 110061474-110061474 on Build GRCh38
rs116442064 Chr.1: 110061818-110061818 on Build GRCh38
rs759007659 Chr.1: 110061521-110061521 on Build GRCh38
rs776099801 Chr.1: 110061410-110061410 on Build GRCh38
rs574975276 Chr.1: 110061799-110061799 on Build GRCh38
rs145535704 Chr.1: 110061455-110061455 on Build GRCh38
rs779144086 Chr.1: 110061459-110061459 on Build GRCh38
rs748725555 Chr.1: 110061462-110061462 on Build GRCh38
rs113851340 Chr.1: 110061531-110061531 on Build GRCh38
rs765099441 Chr.1: 110061396-110061396 on Build GRCh38
rs201353439 Chr.1: 110061584-110061584 on Build GRCh38
rs772578809 Chr.1: 110061579-110061579 on Build GRCh38
rs766980097 Chr.1: 110061427-110061427 on Build GRCh38
rs929301500 Chr.1: 110061627-110061627 on Build GRCh38
rs752816606 Chr.1: 110061619-110061619 on Build GRCh38
rs199753639 Chr.1: 110061454-110061454 on Build GRCh38
rs761334835 Chr.1: 110061491-110061491 on Build GRCh38
rs78415858 Chr.1: 110061809-110061809 on Build GRCh38
rs897926893 Chr.1: 110061381-110061381 on Build GRCh38
rs905015317 Chr.1: 110061781-110061781 on Build GRCh38
rs781367314 Chr.1: 110061525-110061525 on Build GRCh38
rs866514586 Chr.1: 110061399-110061399 on Build GRCh38
rs370985645 Chr.1: 110061494-110061494 on Build GRCh38
rs745653169 Chr.1: 110061475-110061475 on Build GRCh38
rs766089717 Chr.1: 110061588-110061588 on Build GRCh38
rs140822231 Chr.1: 110061481-110061481 on Build GRCh38
rs781127904 Chr.1: 110061527-110061527 on Build GRCh38
rs895851483 Chr.1: 110061755-110061755 on Build GRCh38
rs942256863 Chr.1: 110061600-110061600 on Build GRCh38
rs186135023 Chr.1: 110061483-110061483 on Build GRCh38
rs1012716668 Chr.1: 110061573-110061573 on Build GRCh38
rs1032260186 Chr.1: 110061831-110061831 on Build GRCh38
rs965013210 Chr.1: 110061727-110061727 on Build GRCh38
rs998772057 Chr.1: 110061719-110061719 on Build GRCh38
rs953563606 Chr.1: 110061760-110061760 on Build GRCh38
rs1027987881 Chr.1: 110061759-110061759 on Build GRCh38
rs150129126 Chr.1: 110061520-110061520 on Build GRCh38
rs769697275 Chr.1: 110061476-110061476 on Build GRCh38
rs749039409 Chr.1: 110061533-110061533 on Build GRCh38
rs910822954 Chr.1: 110061583-110061583 on Build GRCh38
rs201764848 Chr.1: 110061514-110061514 on Build GRCh38
rs775250647 Chr.1: 110061478-110061478 on Build GRCh38
rs774210543 Chr.1: 110061538-110061538 on Build GRCh38
rs750158911 Chr.1: 110061495-110061495 on Build GRCh38
rs896354929 Chr.1: 110061410-110061410 on Build GRCh38
rs751044375 Chr.1: 110061449-110061449 on Build GRCh38
rs778377987 Chr.1: 110061524-110061524 on Build GRCh38
rs755307144 Chr.1: 110061456-110061456 on Build GRCh38
rs761654304 Chr.1: 110061541-110061541 on Build GRCh38
rs562601005 Chr.1: 110061710-110061710 on Build GRCh38
rs705279 Chr.1: 110061544-110061544 on Build GRCh38
rs1031435746 Chr.1: 110061386-110061386 on Build GRCh38
rs546279305 Chr.1: 110061690-110061690 on Build GRCh38
rs953835530 Chr.1: 110061720-110061731 on Build GRCh38
rs775299987 Chr.1: 110061532-110061532 on Build GRCh38
rs191002897 Chr.1: 110061666-110061666 on Build GRCh38
rs986673436 Chr.1: 110061774-110061774 on Build GRCh38
rs1023863452 Chr.1: 110061771-110061771 on Build GRCh38

View Full Product Details