Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110211301-110211798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNC4
Entrez Gene ID
3749
Gene Name
potassium voltage-gated channel subfamily C member 4
Gene Aliases
C1orf30, HKSHIIIC, KSHIIIC, KV3.4
UniGene
Hs.153521
Gene Chromosome Location
Chr.1: 110210714-110282649 on Build GRCh38
Transcripts
NM_001039574.2 NM_004978.4 NR_036437.1 XM_006710625.3 XM_006710627.3 XM_011541401.2 XM_011541402.1 XM_011541403.1 XM_011541404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6400366 c.192G>A p.W64* Chr.1: 110211691-110211691 on Build GRCh38
7165928 c.119T>G p.I40S Chr.1: 110211618-110211618 on Build GRCh38
6428996 c.275G>A p.C92Y Chr.1: 110211774-110211774 on Build GRCh38
1583253 c.144G>T p.E48D Chr.1: 110211643-110211643 on Build GRCh38
4694231 c.264C>T p.G88G Chr.1: 110211763-110211763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749209326 Chr.1: 110211736-110211736 on Build GRCh38
rs774956058 Chr.1: 110211746-110211754 on Build GRCh38
rs950751782 Chr.1: 110211683-110211683 on Build GRCh38
rs890978139 Chr.1: 110211353-110211353 on Build GRCh38
rs913923591 Chr.1: 110211550-110211550 on Build GRCh38
rs758889583 Chr.1: 110211667-110211667 on Build GRCh38
rs926699641 Chr.1: 110211520-110211520 on Build GRCh38
rs781619537 Chr.1: 110211715-110211715 on Build GRCh38
rs374585082 Chr.1: 110211587-110211587 on Build GRCh38
rs945927012 Chr.1: 110211555-110211555 on Build GRCh38
rs763639630 Chr.1: 110211525-110211525 on Build GRCh38
rs781774815 Chr.1: 110211512-110211513 on Build GRCh38
rs557834733 Chr.1: 110211420-110211420 on Build GRCh38
rs200808980 Chr.1: 110211728-110211728 on Build GRCh38
rs761468792 Chr.1: 110211648-110211648 on Build GRCh38
rs747202398 Chr.1: 110211489-110211489 on Build GRCh38
rs994937743 Chr.1: 110211654-110211654 on Build GRCh38
rs773855578 Chr.1: 110211752-110211752 on Build GRCh38
rs777960075 Chr.1: 110211562-110211562 on Build GRCh38
rs753174150 Chr.1: 110211664-110211664 on Build GRCh38
rs746078372 Chr.1: 110211716-110211716 on Build GRCh38
rs544030029 Chr.1: 110211334-110211334 on Build GRCh38
rs770207460 Chr.1: 110211576-110211578 on Build GRCh38
rs755391792 Chr.1: 110211472-110211472 on Build GRCh38
rs772190080 Chr.1: 110211485-110211485 on Build GRCh38
rs564226668 Chr.1: 110211439-110211439 on Build GRCh38
rs964820226 Chr.1: 110211360-110211360 on Build GRCh38
rs745527329 Chr.1: 110211625-110211625 on Build GRCh38
rs748484156 Chr.1: 110211520-110211522 on Build GRCh38
rs763718457 Chr.1: 110211459-110211459 on Build GRCh38
rs755514721 Chr.1: 110211554-110211554 on Build GRCh38
rs756778946 Chr.1: 110211407-110211407 on Build GRCh38
rs533097583 Chr.1: 110211578-110211578 on Build GRCh38
rs542783839 Chr.1: 110211382-110211382 on Build GRCh38
rs762669016 Chr.1: 110211455-110211455 on Build GRCh38
rs760216181 Chr.1: 110211764-110211769 on Build GRCh38
rs1048505118 Chr.1: 110211700-110211700 on Build GRCh38
rs768527573 Chr.1: 110211744-110211744 on Build GRCh38
rs529393270 Chr.1: 110211380-110211380 on Build GRCh38
rs758446326 Chr.1: 110211558-110211558 on Build GRCh38
rs772777425 Chr.1: 110211771-110211771 on Build GRCh38
rs749221236 Chr.1: 110211600-110211600 on Build GRCh38
rs762425011 Chr.1: 110211523-110211523 on Build GRCh38
rs946011162 Chr.1: 110211325-110211325 on Build GRCh38
rs1017810707 Chr.1: 110211605-110211605 on Build GRCh38
rs113402589 Chr.1: 110211462-110211462 on Build GRCh38
rs559671892 Chr.1: 110211417-110211417 on Build GRCh38
rs980943199 Chr.1: 110211511-110211511 on Build GRCh38
rs35783710 Chr.1: 110211541-110211541 on Build GRCh38
rs748111654 Chr.1: 110211482-110211482 on Build GRCh38
rs912348081 Chr.1: 110211585-110211585 on Build GRCh38
rs149102895 Chr.1: 110211533-110211533 on Build GRCh38
rs775341759 Chr.1: 110211598-110211598 on Build GRCh38
rs373728555 Chr.1: 110211368-110211369 on Build GRCh38
rs1025221723 Chr.1: 110211748-110211748 on Build GRCh38
rs771559402 Chr.1: 110211769-110211769 on Build GRCh38
rs899572113 Chr.1: 110211638-110211638 on Build GRCh38
rs775490508 Chr.1: 110211522-110211522 on Build GRCh38
rs751618642 Chr.1: 110211694-110211694 on Build GRCh38
rs367820335 Chr.1: 110211658-110211658 on Build GRCh38
rs747724381 Chr.1: 110211761-110211761 on Build GRCh38
rs114915950 Chr.1: 110211430-110211430 on Build GRCh38
rs764282684 Chr.1: 110211675-110211675 on Build GRCh38
rs891467221 Chr.1: 110211514-110211514 on Build GRCh38
rs578185313 Chr.1: 110211328-110211328 on Build GRCh38
rs776517423 Chr.1: 110211492-110211492 on Build GRCh38
rs891057468 Chr.1: 110211677-110211677 on Build GRCh38
rs549548759 Chr.1: 110211631-110211631 on Build GRCh38
rs535148746 Chr.1: 110211704-110211704 on Build GRCh38
rs756521904 Chr.1: 110211463-110211463 on Build GRCh38
rs765602937 Chr.1: 110211662-110211662 on Build GRCh38
rs779357026 Chr.1: 110211557-110211557 on Build GRCh38
rs191936427 Chr.1: 110211730-110211730 on Build GRCh38
rs368490387 Chr.1: 110211646-110211646 on Build GRCh38
rs771678546 Chr.1: 110211742-110211743 on Build GRCh38
rs759526594 Chr.1: 110211496-110211496 on Build GRCh38
rs917167958 Chr.1: 110211720-110211720 on Build GRCh38
rs983067357 Chr.1: 110211424-110211424 on Build GRCh38
rs770880157 Chr.1: 110211491-110211491 on Build GRCh38
rs187164679 Chr.1: 110211362-110211362 on Build GRCh38
rs943849942 Chr.1: 110211614-110211614 on Build GRCh38
rs757509542 Chr.1: 110211526-110211526 on Build GRCh38
rs773807471 Chr.1: 110211606-110211608 on Build GRCh38
rs780370480 Chr.1: 110211466-110211466 on Build GRCh38
rs372085585 Chr.1: 110211419-110211419 on Build GRCh38
rs202062820 Chr.1: 110211532-110211532 on Build GRCh38
rs769547803 Chr.1: 110211500-110211500 on Build GRCh38
rs375782593 Chr.1: 110211564-110211564 on Build GRCh38
rs370049320 Chr.1: 110211577-110211577 on Build GRCh38
rs778188117 Chr.1: 110211487-110211487 on Build GRCh38
rs779660366 Chr.1: 110211571-110211571 on Build GRCh38
rs768189871 Chr.1: 110211628-110211628 on Build GRCh38

View Full Product Details