Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111722376-111722885 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212B
Entrez Gene ID
55924
Gene Name
family with sequence similarity 212 member B
Gene Aliases
C1orf183
UniGene
Hs.193406
Gene Chromosome Location
Chr.1: 111722064-111755797 on Build GRCh38
Transcripts
NM_019099.4 NM_198926.2 XM_011541783.2

SNP Detail

SNP ID Chromosome Location
rs915233910 Chr.1: 111722402-111722402 on Build GRCh38
rs944767634 Chr.1: 111722775-111722775 on Build GRCh38
rs1044585918 Chr.1: 111722437-111722437 on Build GRCh38
rs76860170 Chr.1: 111722710-111722710 on Build GRCh38
rs756334029 Chr.1: 111722573-111722573 on Build GRCh38
rs974046054 Chr.1: 111722843-111722843 on Build GRCh38
rs369868680 Chr.1: 111722643-111722643 on Build GRCh38
rs563076634 Chr.1: 111722616-111722616 on Build GRCh38
rs1005606581 Chr.1: 111722545-111722545 on Build GRCh38
rs148046710 Chr.1: 111722674-111722676 on Build GRCh38
rs952393685 Chr.1: 111722540-111722540 on Build GRCh38
rs986300386 Chr.1: 111722782-111722782 on Build GRCh38
rs948565434 Chr.1: 111722435-111722435 on Build GRCh38
rs117364116 Chr.1: 111722688-111722688 on Build GRCh38
rs1000511744 Chr.1: 111722507-111722507 on Build GRCh38
rs969076350 Chr.1: 111722613-111722613 on Build GRCh38
rs562308812 Chr.1: 111722628-111722628 on Build GRCh38
rs953741392 Chr.1: 111722760-111722760 on Build GRCh38
rs752859719 Chr.1: 111722600-111722602 on Build GRCh38
rs946753889 Chr.1: 111722720-111722720 on Build GRCh38
rs904476705 Chr.1: 111722442-111722442 on Build GRCh38
rs147385845 Chr.1: 111722801-111722801 on Build GRCh38
rs994076020 Chr.1: 111722475-111722475 on Build GRCh38
rs1040863298 Chr.1: 111722449-111722449 on Build GRCh38
rs901127289 Chr.1: 111722454-111722454 on Build GRCh38
rs4838921 Chr.1: 111722463-111722463 on Build GRCh38
rs530323252 Chr.1: 111722617-111722617 on Build GRCh38
rs1023396247 Chr.1: 111722557-111722557 on Build GRCh38
rs1025758746 Chr.1: 111722509-111722509 on Build GRCh38
rs142750240 Chr.1: 111722574-111722574 on Build GRCh38
rs994107671 Chr.1: 111722829-111722829 on Build GRCh38
rs910644394 Chr.1: 111722751-111722751 on Build GRCh38
rs556494681 Chr.1: 111722418-111722418 on Build GRCh38
rs543303342 Chr.1: 111722406-111722407 on Build GRCh38
rs1050825661 Chr.1: 111722746-111722746 on Build GRCh38
rs572673585 Chr.1: 111722663-111722663 on Build GRCh38
rs181996028 Chr.1: 111722828-111722828 on Build GRCh38

View Full Product Details