Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 111726478-111726987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM212B
Entrez Gene ID
55924
Gene Name
family with sequence similarity 212 member B
Gene Aliases
C1orf183
UniGene
Hs.193406
Gene Chromosome Location
Chr.1: 111722064-111755797 on Build GRCh38
Transcripts
NM_019099.4 NM_198926.2 XM_011541783.2

SNP Detail

SNP ID Chromosome Location
rs761142215 Chr.1: 111726945-111726945 on Build GRCh38
rs1049885318 Chr.1: 111726528-111726528 on Build GRCh38
rs189860232 Chr.1: 111726730-111726730 on Build GRCh38
rs575895630 Chr.1: 111726841-111726841 on Build GRCh38
rs138918319 Chr.1: 111726730-111726731 on Build GRCh38
rs1030826341 Chr.1: 111726643-111726643 on Build GRCh38
rs776801113 Chr.1: 111726950-111726950 on Build GRCh38
rs138611018 Chr.1: 111726835-111726835 on Build GRCh38
rs984379959 Chr.1: 111726718-111726718 on Build GRCh38
rs918739475 Chr.1: 111726747-111726747 on Build GRCh38
rs984453538 Chr.1: 111726920-111726920 on Build GRCh38
rs117488967 Chr.1: 111726648-111726648 on Build GRCh38
rs41314013 Chr.1: 111726731-111726731 on Build GRCh38
rs755025047 Chr.1: 111726540-111726540 on Build GRCh38
rs772413340 Chr.1: 111726929-111726929 on Build GRCh38
rs985653817 Chr.1: 111726730-111726731 on Build GRCh38
rs780246122 Chr.1: 111726515-111726515 on Build GRCh38
rs533615115 Chr.1: 111726682-111726682 on Build GRCh38
rs1022790943 Chr.1: 111726700-111726700 on Build GRCh38
rs766744826 Chr.1: 111726947-111726947 on Build GRCh38
rs953774246 Chr.1: 111726728-111726728 on Build GRCh38
rs759463669 Chr.1: 111726954-111726954 on Build GRCh38
rs140925986 Chr.1: 111726711-111726712 on Build GRCh38
rs971750855 Chr.1: 111726710-111726710 on Build GRCh38
rs1043744553 Chr.1: 111726840-111726840 on Build GRCh38
rs939493797 Chr.1: 111726759-111726759 on Build GRCh38
rs1037192857 Chr.1: 111726741-111726741 on Build GRCh38
rs895354224 Chr.1: 111726798-111726798 on Build GRCh38
rs748526100 Chr.1: 111726927-111726927 on Build GRCh38
rs997722562 Chr.1: 111726898-111726898 on Build GRCh38
rs773775871 Chr.1: 111726931-111726931 on Build GRCh38
rs560898650 Chr.1: 111726877-111726877 on Build GRCh38
rs1003787784 Chr.1: 111726522-111726522 on Build GRCh38
rs764294512 Chr.1: 111726901-111726901 on Build GRCh38
rs944289358 Chr.1: 111726560-111726560 on Build GRCh38
rs772051535 Chr.1: 111726797-111726797 on Build GRCh38
rs11809456 Chr.1: 111726517-111726517 on Build GRCh38
rs775866010 Chr.1: 111726694-111726697 on Build GRCh38
rs1012633781 Chr.1: 111726592-111726592 on Build GRCh38
rs796358800 Chr.1: 111726712-111726713 on Build GRCh38
rs955704646 Chr.1: 111726712-111726717 on Build GRCh38
rs976042329 Chr.1: 111726675-111726675 on Build GRCh38
rs79913188 Chr.1: 111726863-111726863 on Build GRCh38
rs200882564 Chr.1: 111726783-111726783 on Build GRCh38
rs953413300 Chr.1: 111726909-111726909 on Build GRCh38
rs752849252 Chr.1: 111726957-111726957 on Build GRCh38
rs968085489 Chr.1: 111726608-111726608 on Build GRCh38
rs780449662 Chr.1: 111726713-111726714 on Build GRCh38
rs1022397256 Chr.1: 111726607-111726607 on Build GRCh38
rs141683410 Chr.1: 111726582-111726582 on Build GRCh38
rs76435252 Chr.1: 111726550-111726550 on Build GRCh38
rs150129230 Chr.1: 111726689-111726689 on Build GRCh38
rs377517685 Chr.1: 111726889-111726889 on Build GRCh38
rs1044646116 Chr.1: 111726864-111726864 on Build GRCh38
rs368595246 Chr.1: 111726783-111726784 on Build GRCh38
rs765506487 Chr.1: 111726955-111726955 on Build GRCh38
rs532878533 Chr.1: 111726507-111726507 on Build GRCh38
rs74400511 Chr.1: 111726888-111726888 on Build GRCh38
rs749204498 Chr.1: 111726634-111726634 on Build GRCh38
rs914327517 Chr.1: 111726728-111726731 on Build GRCh38
rs554579547 Chr.1: 111726611-111726611 on Build GRCh38

View Full Product Details