Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 139360960-139361462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB2
Entrez Gene ID
9276
Gene Name
coatomer protein complex subunit beta 2
Gene Aliases
beta'-COP
UniGene
Hs.681616
Gene Chromosome Location
Chr.3: 139355595-139389680 on Build GRCh38
Transcripts
NM_004766.2 NR_023350.1 XM_017007512.1 XM_017007513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5785725 c.2176A>G p.N726D Chr.3: 139361115-139361115 on Build GRCh38
1637064 c.2125G>A p.A709T Chr.3: 139361166-139361166 on Build GRCh38
1039101 c.2157G>A p.A719A Chr.3: 139361134-139361134 on Build GRCh38
5769176 c.1996-7C>G p.? Chr.3: 139361302-139361302 on Build GRCh38
6634040 c.2093G>A p.G698D Chr.3: 139361198-139361198 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053754913 Chr.3: 139361338-139361338 on Build GRCh38
rs765963919 Chr.3: 139361298-139361298 on Build GRCh38
rs764631027 Chr.3: 139361049-139361049 on Build GRCh38
rs751220094 Chr.3: 139361104-139361104 on Build GRCh38
rs778790033 Chr.3: 139361164-139361164 on Build GRCh38
rs948105943 Chr.3: 139361374-139361377 on Build GRCh38
rs954106550 Chr.3: 139361347-139361347 on Build GRCh38
rs772403942 Chr.3: 139361176-139361176 on Build GRCh38
rs752302628 Chr.3: 139361340-139361340 on Build GRCh38
rs369565965 Chr.3: 139361289-139361289 on Build GRCh38
rs7374710 Chr.3: 139361111-139361111 on Build GRCh38
rs563198810 Chr.3: 139361354-139361354 on Build GRCh38
rs776037503 Chr.3: 139361185-139361185 on Build GRCh38
rs540969150 Chr.3: 139361114-139361114 on Build GRCh38
rs935330619 Chr.3: 139361220-139361220 on Build GRCh38
rs757951607 Chr.3: 139361059-139361059 on Build GRCh38
rs986695408 Chr.3: 139361358-139361358 on Build GRCh38
rs201087921 Chr.3: 139361127-139361127 on Build GRCh38
rs971127344 Chr.3: 139361004-139361004 on Build GRCh38
rs762608793 Chr.3: 139361051-139361054 on Build GRCh38
rs754623890 Chr.3: 139361110-139361110 on Build GRCh38
rs755317775 Chr.3: 139361007-139361007 on Build GRCh38
rs768857801 Chr.3: 139361215-139361215 on Build GRCh38
rs754260482 Chr.3: 139361052-139361052 on Build GRCh38
rs775370744 Chr.3: 139361415-139361415 on Build GRCh38
rs777212739 Chr.3: 139361219-139361219 on Build GRCh38
rs922053443 Chr.3: 139361003-139361003 on Build GRCh38
rs765983959 Chr.3: 139361265-139361265 on Build GRCh38
rs377734699 Chr.3: 139361256-139361256 on Build GRCh38
rs772123622 Chr.3: 139361173-139361173 on Build GRCh38
rs779145946 Chr.3: 139361008-139361008 on Build GRCh38
rs772103492 Chr.3: 139361315-139361315 on Build GRCh38
rs761153916 Chr.3: 139361200-139361200 on Build GRCh38
rs561119379 Chr.3: 139361120-139361120 on Build GRCh38
rs759233209 Chr.3: 139361290-139361290 on Build GRCh38
rs748398802 Chr.3: 139361169-139361169 on Build GRCh38
rs749130594 Chr.3: 139361135-139361135 on Build GRCh38
rs912465584 Chr.3: 139361373-139361373 on Build GRCh38
rs771055699 Chr.3: 139361157-139361157 on Build GRCh38
rs113529756 Chr.3: 139361128-139361128 on Build GRCh38
rs756081148 Chr.3: 139361125-139361125 on Build GRCh38
rs543622675 Chr.3: 139361134-139361134 on Build GRCh38
rs765753032 Chr.3: 139361073-139361073 on Build GRCh38
rs944757946 Chr.3: 139361410-139361410 on Build GRCh38

View Full Product Details