Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149140636-149141138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS3
Entrez Gene ID
84343
Gene Name
HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
Gene Aliases
BLOC2S1, SUTAL
UniGene
Hs.558314
Gene Chromosome Location
Chr.3: 149129584-149173196 on Build GRCh38
Transcripts
NM_001308258.1 NM_032383.4 XM_005247834.3 XM_017007323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630832 c.769C>T p.P257S Chr.3: 149141073-149141073 on Build GRCh38
3729023 c.713-10delT p.? Chr.3: 149141007-149141007 on Build GRCh38
4450729 c.809_817delTATCTGTTA p.L270_T273>S Chr.3: 149141113-149141121 on Build GRCh38
445777 c.812C>G p.S271C Chr.3: 149141116-149141116 on Build GRCh38
7142103 c.756C>A p.V252V Chr.3: 149141060-149141060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1057018509 Chr.3: 149141107-149141107 on Build GRCh38
rs557158646 Chr.3: 149140903-149140903 on Build GRCh38
rs1023178089 Chr.3: 149140796-149140796 on Build GRCh38
rs745431040 Chr.3: 149141110-149141110 on Build GRCh38
rs554010598 Chr.3: 149140774-149140774 on Build GRCh38
rs1019076581 Chr.3: 149140762-149140762 on Build GRCh38
rs999079198 Chr.3: 149140768-149140768 on Build GRCh38
rs1010316506 Chr.3: 149140798-149140798 on Build GRCh38
rs1033245955 Chr.3: 149140772-149140772 on Build GRCh38
rs755372867 Chr.3: 149141038-149141038 on Build GRCh38
rs181701127 Chr.3: 149140912-149140912 on Build GRCh38
rs868067117 Chr.3: 149140718-149140718 on Build GRCh38
rs752004185 Chr.3: 149141118-149141118 on Build GRCh38
rs534328122 Chr.3: 149140755-149140755 on Build GRCh38
rs767334706 Chr.3: 149141029-149141030 on Build GRCh38
rs545439652 Chr.3: 149141064-149141064 on Build GRCh38
rs752034836 Chr.3: 149141037-149141037 on Build GRCh38
rs942091926 Chr.3: 149140746-149140746 on Build GRCh38
rs773842435 Chr.3: 149140940-149140940 on Build GRCh38
rs568260386 Chr.3: 149140708-149140708 on Build GRCh38
rs1037729992 Chr.3: 149141075-149141075 on Build GRCh38
rs80142888 Chr.3: 149140914-149140914 on Build GRCh38
rs750257088 Chr.3: 149140698-149140698 on Build GRCh38
rs150098273 Chr.3: 149140675-149140675 on Build GRCh38
rs886525128 Chr.3: 149140743-149140743 on Build GRCh38
rs143594510 Chr.3: 149141000-149141000 on Build GRCh38
rs754423156 Chr.3: 149140924-149140924 on Build GRCh38
rs565928278 Chr.3: 149140698-149140698 on Build GRCh38
rs750402363 Chr.3: 149141023-149141023 on Build GRCh38
rs760577035 Chr.3: 149141032-149141033 on Build GRCh38
rs886058075 Chr.3: 149141020-149141020 on Build GRCh38
rs1037509423 Chr.3: 149140752-149140752 on Build GRCh38
rs1019911520 Chr.3: 149140834-149140834 on Build GRCh38
rs1047847056 Chr.3: 149140741-149140741 on Build GRCh38
rs1050450815 Chr.3: 149141020-149141020 on Build GRCh38
rs765371581 Chr.3: 149141017-149141017 on Build GRCh38
rs186810630 Chr.3: 149140985-149140985 on Build GRCh38
rs999919813 Chr.3: 149140866-149140866 on Build GRCh38
rs374691328 Chr.3: 149141013-149141013 on Build GRCh38
rs373488474 Chr.3: 149141029-149141029 on Build GRCh38
rs756796449 Chr.3: 149141079-149141079 on Build GRCh38
rs759560948 Chr.3: 149141006-149141007 on Build GRCh38
rs78383046 Chr.3: 149140775-149140775 on Build GRCh38
rs112117971 Chr.3: 149140785-149140785 on Build GRCh38
rs1032531224 Chr.3: 149140869-149140869 on Build GRCh38
rs927155737 Chr.3: 149141093-149141093 on Build GRCh38
rs775449783 Chr.3: 149141030-149141031 on Build GRCh38
rs376398872 Chr.3: 149141101-149141101 on Build GRCh38
rs1040411263 Chr.3: 149141076-149141076 on Build GRCh38
rs903384018 Chr.3: 149141098-149141098 on Build GRCh38
rs778626931 Chr.3: 149141085-149141085 on Build GRCh38
rs766853921 Chr.3: 149141030-149141030 on Build GRCh38
rs781494097 Chr.3: 149141050-149141050 on Build GRCh38
rs543095487 Chr.3: 149140804-149140804 on Build GRCh38
rs981154550 Chr.3: 149140882-149140882 on Build GRCh38
rs776727453 Chr.3: 149140942-149140942 on Build GRCh38
rs73164975 Chr.3: 149140766-149140766 on Build GRCh38
rs923291302 Chr.3: 149140674-149140674 on Build GRCh38
rs762079803 Chr.3: 149141005-149141005 on Build GRCh38
rs971140878 Chr.3: 149140835-149140835 on Build GRCh38

View Full Product Details