Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 149141117-149141516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPS3
Entrez Gene ID
84343
Gene Name
HPS3, biogenesis of lysosomal organelles complex 2 subunit 1
Gene Aliases
BLOC2S1, SUTAL
UniGene
Hs.558314
Gene Chromosome Location
Chr.3: 149129584-149173196 on Build GRCh38
Transcripts
NM_001308258.1 NM_032383.4 XM_005247834.3 XM_017007323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3002628 c.905C>T p.S302L Chr.3: 149141315-149141315 on Build GRCh38
6681948 c.906G>A p.S302S Chr.3: 149141316-149141316 on Build GRCh38
3660254 c.860C>A p.S287Y Chr.3: 149141164-149141164 on Build GRCh38
3784358 c.970+4A>G p.? Chr.3: 149141384-149141384 on Build GRCh38
4450729 c.809_817delTATCTGTTA p.L270_T273>S Chr.3: 149141113-149141121 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758012717 Chr.3: 149141302-149141302 on Build GRCh38
rs775294702 Chr.3: 149141388-149141389 on Build GRCh38
rs763192167 Chr.3: 149141253-149141253 on Build GRCh38
rs773468722 Chr.3: 149141161-149141161 on Build GRCh38
rs544577394 Chr.3: 149141487-149141487 on Build GRCh38
rs760630186 Chr.3: 149141429-149141432 on Build GRCh38
rs200702850 Chr.3: 149141406-149141406 on Build GRCh38
rs1022637234 Chr.3: 149141163-149141163 on Build GRCh38
rs146928432 Chr.3: 149141168-149141168 on Build GRCh38
rs759609460 Chr.3: 149141288-149141288 on Build GRCh38
rs779663433 Chr.3: 149141214-149141214 on Build GRCh38
rs367758594 Chr.3: 149141200-149141200 on Build GRCh38
rs530919536 Chr.3: 149141445-149141445 on Build GRCh38
rs201568134 Chr.3: 149141403-149141403 on Build GRCh38
rs944688815 Chr.3: 149141366-149141366 on Build GRCh38
rs764381762 Chr.3: 149141356-149141356 on Build GRCh38
rs756488920 Chr.3: 149141202-149141203 on Build GRCh38
rs758679569 Chr.3: 149141201-149141201 on Build GRCh38
rs767875239 Chr.3: 149141194-149141194 on Build GRCh38
rs781269202 Chr.3: 149141220-149141220 on Build GRCh38
rs377428730 Chr.3: 149141330-149141330 on Build GRCh38
rs927381356 Chr.3: 149141481-149141481 on Build GRCh38
rs536794577 Chr.3: 149141353-149141353 on Build GRCh38
rs71304453 Chr.3: 149141404-149141404 on Build GRCh38
rs766446900 Chr.3: 149141172-149141172 on Build GRCh38
rs886058076 Chr.3: 149141199-149141200 on Build GRCh38
rs529089901 Chr.3: 149141238-149141238 on Build GRCh38
rs910569690 Chr.3: 149141348-149141348 on Build GRCh38
rs754520795 Chr.3: 149141309-149141309 on Build GRCh38
rs774355427 Chr.3: 149141273-149141273 on Build GRCh38
rs751180573 Chr.3: 149141305-149141305 on Build GRCh38
rs141832148 Chr.3: 149141297-149141297 on Build GRCh38
rs770934699 Chr.3: 149141259-149141259 on Build GRCh38
rs751233801 Chr.3: 149141367-149141367 on Build GRCh38
rs878887233 Chr.3: 149141201-149141202 on Build GRCh38
rs781327575 Chr.3: 149141315-149141315 on Build GRCh38
rs778483476 Chr.3: 149141199-149141199 on Build GRCh38
rs371430031 Chr.3: 149141341-149141341 on Build GRCh38
rs769609430 Chr.3: 149141225-149141225 on Build GRCh38
rs752595588 Chr.3: 149141397-149141397 on Build GRCh38
rs756491989 Chr.3: 149141197-149141197 on Build GRCh38
rs565326513 Chr.3: 149141408-149141408 on Build GRCh38
rs114029765 Chr.3: 149141387-149141387 on Build GRCh38
rs10693502 Chr.3: 149141213-149141214 on Build GRCh38
rs769503746 Chr.3: 149141328-149141328 on Build GRCh38
rs1040463306 Chr.3: 149141368-149141368 on Build GRCh38
rs202215373 Chr.3: 149141195-149141195 on Build GRCh38
rs200882543 Chr.3: 149141142-149141142 on Build GRCh38
rs1013548674 Chr.3: 149141159-149141159 on Build GRCh38
rs1054634505 Chr.3: 149141398-149141398 on Build GRCh38
rs368415299 Chr.3: 149141364-149141364 on Build GRCh38
rs770541463 Chr.3: 149141214-149141219 on Build GRCh38
rs906891890 Chr.3: 149141438-149141438 on Build GRCh38
rs749010472 Chr.3: 149141214-149141220 on Build GRCh38
rs137952312 Chr.3: 149141296-149141296 on Build GRCh38
rs759983514 Chr.3: 149141187-149141187 on Build GRCh38
rs772162331 Chr.3: 149141417-149141417 on Build GRCh38
rs757879257 Chr.3: 149141207-149141207 on Build GRCh38
rs200395770 Chr.3: 149141342-149141342 on Build GRCh38
rs1045020679 Chr.3: 149141416-149141416 on Build GRCh38
rs747476250 Chr.3: 149141424-149141424 on Build GRCh38
rs749167874 Chr.3: 149141405-149141405 on Build GRCh38
rs750685598 Chr.3: 149141154-149141155 on Build GRCh38
rs111598115 Chr.3: 149141200-149141201 on Build GRCh38
rs370943042 Chr.3: 149141380-149141380 on Build GRCh38
rs200156408 Chr.3: 149141185-149141185 on Build GRCh38
rs570886288 Chr.3: 149141349-149141349 on Build GRCh38
rs762077425 Chr.3: 149141362-149141362 on Build GRCh38
rs775582716 Chr.3: 149141419-149141419 on Build GRCh38
rs201040693 Chr.3: 149141215-149141215 on Build GRCh38
rs370670658 Chr.3: 149141219-149141219 on Build GRCh38
rs771072752 Chr.3: 149141334-149141334 on Build GRCh38
rs768073408 Chr.3: 149141289-149141289 on Build GRCh38
rs879006087 Chr.3: 149141474-149141474 on Build GRCh38
rs373049759 Chr.3: 149141303-149141303 on Build GRCh38
rs753183516 Chr.3: 149141291-149141291 on Build GRCh38
rs886058077 Chr.3: 149141213-149141213 on Build GRCh38
rs752087954 Chr.3: 149141186-149141186 on Build GRCh38
rs778100609 Chr.3: 149141205-149141206 on Build GRCh38
rs762991548 Chr.3: 149141162-149141162 on Build GRCh38
rs878939393 Chr.3: 149141399-149141399 on Build GRCh38
rs199556788 Chr.3: 149141379-149141379 on Build GRCh38
rs748088057 Chr.3: 149141321-149141321 on Build GRCh38

View Full Product Details