Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172443826-172444364 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHSR
Entrez Gene ID
2693
Gene Name
growth hormone secretagogue receptor
Gene Aliases
GHDP
UniGene
Hs.130212
Gene Chromosome Location
Chr.3: 172443291-172448456 on Build GRCh38
Transcripts
NM_198407.2

SNP Detail

SNP ID Chromosome Location
rs747895075 Chr.3: 172443871-172443871 on Build GRCh38
rs564915227 Chr.3: 172444288-172444288 on Build GRCh38
rs754292678 Chr.3: 172444093-172444101 on Build GRCh38
rs539145341 Chr.3: 172444058-172444058 on Build GRCh38
rs896106040 Chr.3: 172444136-172444136 on Build GRCh38
rs988711038 Chr.3: 172444103-172444103 on Build GRCh38
rs906051136 Chr.3: 172444075-172444075 on Build GRCh38
rs978622113 Chr.3: 172444070-172444070 on Build GRCh38
rs185937419 Chr.3: 172444200-172444200 on Build GRCh38
rs966902598 Chr.3: 172443950-172443950 on Build GRCh38
rs1000263913 Chr.3: 172444336-172444336 on Build GRCh38
rs539823726 Chr.3: 172444251-172444251 on Build GRCh38
rs772213395 Chr.3: 172444099-172444099 on Build GRCh38
rs775843480 Chr.3: 172444100-172444100 on Build GRCh38
rs973182049 Chr.3: 172444023-172444023 on Build GRCh38
rs139844524 Chr.3: 172444000-172444000 on Build GRCh38
rs888646747 Chr.3: 172444264-172444264 on Build GRCh38
rs554362985 Chr.3: 172443899-172443899 on Build GRCh38
rs755925335 Chr.3: 172443893-172443893 on Build GRCh38
rs1048637895 Chr.3: 172443968-172443968 on Build GRCh38
rs542461021 Chr.3: 172444164-172444164 on Build GRCh38
rs182732562 Chr.3: 172444003-172444003 on Build GRCh38
rs1057309895 Chr.3: 172444135-172444135 on Build GRCh38
rs886058182 Chr.3: 172444319-172444319 on Build GRCh38
rs777058233 Chr.3: 172444227-172444227 on Build GRCh38
rs145364247 Chr.3: 172444322-172444322 on Build GRCh38
rs79483527 Chr.3: 172444064-172444064 on Build GRCh38
rs998207476 Chr.3: 172444324-172444324 on Build GRCh38
rs1013700640 Chr.3: 172443882-172443882 on Build GRCh38
rs531192936 Chr.3: 172443865-172443865 on Build GRCh38
rs535110477 Chr.3: 172444332-172444332 on Build GRCh38
rs756728640 Chr.3: 172444115-172444115 on Build GRCh38
rs1037305474 Chr.3: 172444319-172444319 on Build GRCh38
rs555579461 Chr.3: 172444321-172444321 on Build GRCh38
rs565105 Chr.3: 172444327-172444327 on Build GRCh38
rs572834585 Chr.3: 172444218-172444218 on Build GRCh38
rs749217235 Chr.3: 172443973-172443973 on Build GRCh38
rs972641218 Chr.3: 172444338-172444338 on Build GRCh38
rs73039028 Chr.3: 172443853-172443853 on Build GRCh38
rs112111487 Chr.3: 172444004-172444004 on Build GRCh38
rs752822264 Chr.3: 172444107-172444107 on Build GRCh38
rs764510468 Chr.3: 172444139-172444139 on Build GRCh38
rs562416 Chr.3: 172444052-172444052 on Build GRCh38
rs905329424 Chr.3: 172443945-172443945 on Build GRCh38
rs979165376 Chr.3: 172443959-172443959 on Build GRCh38
rs896053509 Chr.3: 172443868-172443868 on Build GRCh38
rs925830346 Chr.3: 172444078-172444078 on Build GRCh38
rs1048584905 Chr.3: 172444211-172444211 on Build GRCh38
rs570105204 Chr.3: 172443854-172443854 on Build GRCh38
rs953981809 Chr.3: 172444065-172444065 on Build GRCh38
rs544302265 Chr.3: 172444318-172444318 on Build GRCh38
rs777638365 Chr.3: 172443954-172443954 on Build GRCh38
rs9880206 Chr.3: 172444137-172444137 on Build GRCh38
rs1044504290 Chr.3: 172444123-172444123 on Build GRCh38
rs553367905 Chr.3: 172444032-172444032 on Build GRCh38
rs955941508 Chr.3: 172444095-172444095 on Build GRCh38
rs532071665 Chr.3: 172444316-172444316 on Build GRCh38

View Full Product Details