Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 172448251-172448751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHSR
Entrez Gene ID
2693
Gene Name
growth hormone secretagogue receptor
Gene Aliases
GHDP
UniGene
Hs.130212
Gene Chromosome Location
Chr.3: 172443291-172448456 on Build GRCh38
Transcripts
NM_004122.2 NM_198407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4115285 c.10G>A p.A4T Chr.3: 172448404-172448404 on Build GRCh38
5571565 c.151G>C p.V51L Chr.3: 172448263-172448263 on Build GRCh38
3392215 c.94G>A p.D32N Chr.3: 172448320-172448320 on Build GRCh38
1420646 c.132G>A p.A44A Chr.3: 172448282-172448282 on Build GRCh38
6670865 c.15G>A p.T5T Chr.3: 172448399-172448399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533287787 Chr.3: 172448616-172448616 on Build GRCh38
rs200044818 Chr.3: 172448472-172448472 on Build GRCh38
rs746082893 Chr.3: 172448444-172448444 on Build GRCh38
rs199859784 Chr.3: 172448447-172448447 on Build GRCh38
rs1044966868 Chr.3: 172448451-172448451 on Build GRCh38
rs919779255 Chr.3: 172448674-172448674 on Build GRCh38
rs980594821 Chr.3: 172448643-172448643 on Build GRCh38
rs755709287 Chr.3: 172448419-172448419 on Build GRCh38
rs761345817 Chr.3: 172448514-172448514 on Build GRCh38
rs140224509 Chr.3: 172448290-172448290 on Build GRCh38
rs762767250 Chr.3: 172448389-172448389 on Build GRCh38
rs899122413 Chr.3: 172448679-172448679 on Build GRCh38
rs543942634 Chr.3: 172448537-172448537 on Build GRCh38
rs199870358 Chr.3: 172448336-172448336 on Build GRCh38
rs746106545 Chr.3: 172448362-172448362 on Build GRCh38
rs1029157553 Chr.3: 172448532-172448532 on Build GRCh38
rs1047159253 Chr.3: 172448459-172448459 on Build GRCh38
rs200419561 Chr.3: 172448343-172448343 on Build GRCh38
rs267606843 Chr.3: 172448408-172448408 on Build GRCh38
rs756727375 Chr.3: 172448333-172448333 on Build GRCh38
rs750269236 Chr.3: 172448312-172448312 on Build GRCh38
rs773044485 Chr.3: 172448384-172448384 on Build GRCh38
rs201076298 Chr.3: 172448450-172448450 on Build GRCh38
rs767104789 Chr.3: 172448410-172448410 on Build GRCh38
rs776383117 Chr.3: 172448428-172448428 on Build GRCh38
rs1031390876 Chr.3: 172448568-172448568 on Build GRCh38
rs899069867 Chr.3: 172448394-172448394 on Build GRCh38
rs545162620 Chr.3: 172448728-172448728 on Build GRCh38
rs149430564 Chr.3: 172448346-172448346 on Build GRCh38
rs752306875 Chr.3: 172448418-172448418 on Build GRCh38
rs923988666 Chr.3: 172448432-172448432 on Build GRCh38
rs754896970 Chr.3: 172448305-172448307 on Build GRCh38
rs773134895 Chr.3: 172448457-172448457 on Build GRCh38
rs766092209 Chr.3: 172448392-172448392 on Build GRCh38
rs74791529 Chr.3: 172448706-172448706 on Build GRCh38
rs200380996 Chr.3: 172448317-172448317 on Build GRCh38
rs199691534 Chr.3: 172448425-172448425 on Build GRCh38
rs992611926 Chr.3: 172448602-172448602 on Build GRCh38
rs1004821824 Chr.3: 172448374-172448374 on Build GRCh38
rs921560250 Chr.3: 172448572-172448572 on Build GRCh38
rs373569448 Chr.3: 172448382-172448382 on Build GRCh38
rs751202593 Chr.3: 172448318-172448318 on Build GRCh38
rs201558252 Chr.3: 172448278-172448278 on Build GRCh38
rs761792362 Chr.3: 172448298-172448298 on Build GRCh38
rs771233160 Chr.3: 172448356-172448356 on Build GRCh38
rs748661863 Chr.3: 172448330-172448330 on Build GRCh38
rs765009184 Chr.3: 172448310-172448310 on Build GRCh38
rs185959116 Chr.3: 172448564-172448564 on Build GRCh38
rs940548794 Chr.3: 172448670-172448670 on Build GRCh38
rs2232165 Chr.3: 172448354-172448354 on Build GRCh38
rs754471403 Chr.3: 172448323-172448323 on Build GRCh38
rs202007365 Chr.3: 172448283-172448283 on Build GRCh38
rs947921947 Chr.3: 172448306-172448306 on Build GRCh38
rs1056682592 Chr.3: 172448279-172448279 on Build GRCh38
rs767592667 Chr.3: 172448291-172448291 on Build GRCh38
rs201998267 Chr.3: 172448481-172448481 on Build GRCh38
rs200320085 Chr.3: 172448487-172448487 on Build GRCh38
rs200645708 Chr.3: 172448436-172448436 on Build GRCh38
rs377344701 Chr.3: 172448541-172448541 on Build GRCh38
rs772317940 Chr.3: 172448445-172448445 on Build GRCh38
rs369278883 Chr.3: 172448364-172448364 on Build GRCh38
rs931196804 Chr.3: 172448675-172448675 on Build GRCh38
rs372133942 Chr.3: 172448453-172448453 on Build GRCh38
rs201206843 Chr.3: 172448480-172448480 on Build GRCh38
rs201599372 Chr.3: 172448439-172448439 on Build GRCh38
rs931684248 Chr.3: 172448328-172448328 on Build GRCh38
rs144006748 Chr.3: 172448400-172448400 on Build GRCh38
rs553299240 Chr.3: 172448369-172448369 on Build GRCh38
rs969493362 Chr.3: 172448624-172448624 on Build GRCh38
rs996294178 Chr.3: 172448519-172448519 on Build GRCh38
rs374126118 Chr.3: 172448603-172448603 on Build GRCh38
rs979737367 Chr.3: 172448567-172448567 on Build GRCh38
rs561816644 Chr.3: 172448539-172448539 on Build GRCh38
rs988252217 Chr.3: 172448468-172448468 on Build GRCh38
rs754745493 Chr.3: 172448402-172448402 on Build GRCh38
rs573873988 Chr.3: 172448551-172448551 on Build GRCh38
rs201109603 Chr.3: 172448491-172448491 on Build GRCh38
rs150293762 Chr.3: 172448313-172448313 on Build GRCh38
rs138443343 Chr.3: 172448358-172448358 on Build GRCh38
rs776496820 Chr.3: 172448297-172448297 on Build GRCh38
rs147528033 Chr.3: 172448584-172448584 on Build GRCh38
rs773780095 Chr.3: 172448277-172448277 on Build GRCh38
rs2232166 Chr.3: 172448270-172448270 on Build GRCh38
rs201682882 Chr.3: 172448456-172448456 on Build GRCh38
rs761706124 Chr.3: 172448376-172448376 on Build GRCh38
rs947869836 Chr.3: 172448448-172448448 on Build GRCh38
rs200861497 Chr.3: 172448381-172448381 on Build GRCh38
rs137953416 Chr.3: 172448327-172448327 on Build GRCh38
rs778306245 Chr.3: 172448420-172448420 on Build GRCh38

View Full Product Details