Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184136471-184136997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B5
Entrez Gene ID
8893
Gene Name
eukaryotic translation initiation factor 2B subunit epsilon
Gene Aliases
CACH, CLE, EIF-2B, EIF2Bepsilon, LVWM
UniGene
Hs.283551
Gene Chromosome Location
Chr.3: 184134435-184145311 on Build GRCh38
Transcripts
NM_003907.2 XM_011513266.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4115646 c.311A>C p.E104A Chr.3: 184136727-184136727 on Build GRCh38
5014400 c.232T>C p.Y78H Chr.3: 184136648-184136648 on Build GRCh38
4115645 c.233A>G p.Y78C Chr.3: 184136649-184136649 on Build GRCh38
3427386 c.278T>C p.V93A Chr.3: 184136694-184136694 on Build GRCh38
6316341 c.217G>T p.V73L Chr.3: 184136633-184136633 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs567030655 Chr.3: 184136881-184136881 on Build GRCh38
rs761050107 Chr.3: 184136583-184136583 on Build GRCh38
rs762701294 Chr.3: 184136770-184136770 on Build GRCh38
rs150793860 Chr.3: 184136531-184136531 on Build GRCh38
rs768761281 Chr.3: 184136581-184136582 on Build GRCh38
rs142679815 Chr.3: 184136943-184136943 on Build GRCh38
rs374876865 Chr.3: 184136769-184136769 on Build GRCh38
rs766064882 Chr.3: 184136782-184136782 on Build GRCh38
rs754345535 Chr.3: 184136601-184136601 on Build GRCh38
rs543426662 Chr.3: 184136500-184136500 on Build GRCh38
rs755979083 Chr.3: 184136784-184136784 on Build GRCh38
rs113994045 Chr.3: 184136634-184136634 on Build GRCh38
rs370609212 Chr.3: 184136571-184136571 on Build GRCh38
rs886058214 Chr.3: 184136632-184136632 on Build GRCh38
rs968181452 Chr.3: 184136650-184136650 on Build GRCh38
rs778822941 Chr.3: 184136693-184136693 on Build GRCh38
rs776734512 Chr.3: 184136587-184136587 on Build GRCh38
rs775719092 Chr.3: 184136726-184136726 on Build GRCh38
rs923447162 Chr.3: 184136914-184136914 on Build GRCh38
rs113994047 Chr.3: 184136657-184136657 on Build GRCh38
rs1034411626 Chr.3: 184136809-184136809 on Build GRCh38
rs942079682 Chr.3: 184136863-184136863 on Build GRCh38
rs976309388 Chr.3: 184136895-184136895 on Build GRCh38
rs571561821 Chr.3: 184136844-184136844 on Build GRCh38
rs767056172 Chr.3: 184136972-184136972 on Build GRCh38
rs368375006 Chr.3: 184136568-184136568 on Build GRCh38
rs146048484 Chr.3: 184136970-184136970 on Build GRCh38
rs747305325 Chr.3: 184136729-184136729 on Build GRCh38
rs530149499 Chr.3: 184136569-184136569 on Build GRCh38
rs141748678 Chr.3: 184136644-184136644 on Build GRCh38
rs371931237 Chr.3: 184136763-184136763 on Build GRCh38
rs28939717 Chr.3: 184136687-184136687 on Build GRCh38
rs746008885 Chr.3: 184136694-184136694 on Build GRCh38
rs762222639 Chr.3: 184136747-184136747 on Build GRCh38
rs759670014 Chr.3: 184136564-184136564 on Build GRCh38
rs972180837 Chr.3: 184136878-184136878 on Build GRCh38
rs548249072 Chr.3: 184136682-184136682 on Build GRCh38
rs113994044 Chr.3: 184136619-184136619 on Build GRCh38
rs138072251 Chr.3: 184136939-184136939 on Build GRCh38
rs867266088 Chr.3: 184136823-184136823 on Build GRCh38
rs764543925 Chr.3: 184136585-184136585 on Build GRCh38
rs938660226 Chr.3: 184136923-184136923 on Build GRCh38
rs919218257 Chr.3: 184136908-184136908 on Build GRCh38
rs1023494334 Chr.3: 184136575-184136575 on Build GRCh38
rs150531914 Chr.3: 184136715-184136715 on Build GRCh38
rs923517578 Chr.3: 184136813-184136813 on Build GRCh38
rs1000985816 Chr.3: 184136976-184136976 on Build GRCh38
rs2271595 Chr.3: 184136538-184136538 on Build GRCh38
rs1012483241 Chr.3: 184136547-184136547 on Build GRCh38
rs761986445 Chr.3: 184136783-184136784 on Build GRCh38
rs751354540 Chr.3: 184136666-184136666 on Build GRCh38
rs750017665 Chr.3: 184136647-184136647 on Build GRCh38
rs903693906 Chr.3: 184136975-184136975 on Build GRCh38
rs113994046 Chr.3: 184136636-184136636 on Build GRCh38
rs754854073 Chr.3: 184136678-184136678 on Build GRCh38
rs908141283 Chr.3: 184136839-184136839 on Build GRCh38
rs191998519 Chr.3: 184136938-184136938 on Build GRCh38
rs116835002 Chr.3: 184136745-184136745 on Build GRCh38
rs752573705 Chr.3: 184136778-184136778 on Build GRCh38
rs113994048 Chr.3: 184136734-184136734 on Build GRCh38
rs909400446 Chr.3: 184136850-184136850 on Build GRCh38
rs762550544 Chr.3: 184136768-184136768 on Build GRCh38
rs374801184 Chr.3: 184136603-184136603 on Build GRCh38
rs1000636351 Chr.3: 184136677-184136677 on Build GRCh38
rs538507828 Chr.3: 184136845-184136845 on Build GRCh38
rs111248061 Chr.3: 184136530-184136530 on Build GRCh38
rs781211740 Chr.3: 184136681-184136681 on Build GRCh38
rs565612701 Chr.3: 184136504-184136504 on Build GRCh38
rs765813754 Chr.3: 184136757-184136757 on Build GRCh38
rs932110011 Chr.3: 184136935-184136941 on Build GRCh38
rs774344255 Chr.3: 184136663-184136663 on Build GRCh38

View Full Product Details