Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184163578-184164081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DVL3
Entrez Gene ID
1857
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
DRS3
UniGene
Hs.388116
Gene Chromosome Location
Chr.3: 184155311-184173614 on Build GRCh38
Transcripts
NM_004423.3 XM_005247172.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5696604 c.217C>G p.R73G Chr.3: 184163712-184163712 on Build GRCh38
4115655 c.183G>A p.S61S Chr.3: 184163678-184163678 on Build GRCh38
4115654 c.182C>T p.S61L Chr.3: 184163677-184163677 on Build GRCh38
1041922 c.202C>T p.P68S Chr.3: 184163697-184163697 on Build GRCh38
382316 c.183G>T p.S61S Chr.3: 184163678-184163678 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775385632 Chr.3: 184163637-184163637 on Build GRCh38
rs116156353 Chr.3: 184163770-184163770 on Build GRCh38
rs746909865 Chr.3: 184163751-184163751 on Build GRCh38
rs928777173 Chr.3: 184163984-184163984 on Build GRCh38
rs956173111 Chr.3: 184163797-184163797 on Build GRCh38
rs1024592706 Chr.3: 184163908-184163914 on Build GRCh38
rs984871687 Chr.3: 184163904-184163904 on Build GRCh38
rs773097646 Chr.3: 184163766-184163766 on Build GRCh38
rs766628577 Chr.3: 184163678-184163678 on Build GRCh38
rs551827820 Chr.3: 184163956-184163956 on Build GRCh38
rs889499262 Chr.3: 184163739-184163739 on Build GRCh38
rs758158693 Chr.3: 184163746-184163746 on Build GRCh38
rs988730208 Chr.3: 184163849-184163849 on Build GRCh38
rs1709643 Chr.3: 184163943-184163943 on Build GRCh38
rs965878495 Chr.3: 184163826-184163826 on Build GRCh38
rs752861795 Chr.3: 184164037-184164037 on Build GRCh38
rs749485607 Chr.3: 184163641-184163641 on Build GRCh38
rs759896212 Chr.3: 184163902-184163902 on Build GRCh38
rs763713678 Chr.3: 184164049-184164049 on Build GRCh38
rs981724844 Chr.3: 184163981-184163981 on Build GRCh38
rs1055866796 Chr.3: 184164026-184164026 on Build GRCh38
rs34601858 Chr.3: 184163852-184163853 on Build GRCh38
rs774816506 Chr.3: 184163679-184163679 on Build GRCh38
rs760647520 Chr.3: 184163644-184163644 on Build GRCh38
rs971214413 Chr.3: 184163927-184163927 on Build GRCh38
rs756823260 Chr.3: 184163727-184163727 on Build GRCh38
rs1029178121 Chr.3: 184163867-184163867 on Build GRCh38
rs267599710 Chr.3: 184163698-184163698 on Build GRCh38
rs371684582 Chr.3: 184163685-184163685 on Build GRCh38
rs768039340 Chr.3: 184163696-184163696 on Build GRCh38
rs753395619 Chr.3: 184163713-184163713 on Build GRCh38
rs774688011 Chr.3: 184163778-184163778 on Build GRCh38
rs370079001 Chr.3: 184163753-184163753 on Build GRCh38
rs187470505 Chr.3: 184163901-184163901 on Build GRCh38
rs908079613 Chr.3: 184163909-184163909 on Build GRCh38
rs756709038 Chr.3: 184163711-184163711 on Build GRCh38
rs759326723 Chr.3: 184163872-184163872 on Build GRCh38
rs995863020 Chr.3: 184163851-184163851 on Build GRCh38
rs2943477 Chr.3: 184163979-184163979 on Build GRCh38
rs773397133 Chr.3: 184163663-184163663 on Build GRCh38
rs778562744 Chr.3: 184163732-184163732 on Build GRCh38
rs1012012688 Chr.3: 184163608-184163608 on Build GRCh38
rs926709784 Chr.3: 184163941-184163941 on Build GRCh38
rs1018724447 Chr.3: 184163824-184163824 on Build GRCh38
rs947368265 Chr.3: 184164044-184164044 on Build GRCh38
rs143411607 Chr.3: 184163677-184163677 on Build GRCh38
rs796286446 Chr.3: 184163955-184163955 on Build GRCh38
rs771010090 Chr.3: 184163777-184163777 on Build GRCh38
rs753220846 Chr.3: 184163699-184163699 on Build GRCh38
rs369009696 Chr.3: 184163651-184163651 on Build GRCh38
rs375976533 Chr.3: 184163748-184163748 on Build GRCh38
rs769760411 Chr.3: 184163659-184163659 on Build GRCh38
rs979890856 Chr.3: 184163930-184163930 on Build GRCh38
rs745697751 Chr.3: 184163626-184163626 on Build GRCh38
rs767360360 Chr.3: 184163907-184163907 on Build GRCh38
rs1709642 Chr.3: 184163987-184163987 on Build GRCh38
rs771948026 Chr.3: 184163634-184163634 on Build GRCh38
rs73185719 Chr.3: 184164056-184164056 on Build GRCh38
rs745569187 Chr.3: 184163733-184163733 on Build GRCh38

View Full Product Details