Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186854711-186855224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOQ
Entrez Gene ID
9370
Gene Name
adiponectin, C1Q and collagen domain containing
Gene Aliases
ACDC, ACRP30, ADIPQTL1, ADPN, APM-1, APM1, GBP28
UniGene
Hs.80485
Gene Chromosome Location
Chr.3: 186842674-186858463 on Build GRCh38
Transcripts
NM_001177800.1 NM_004797.3 NR_046662.1

Gene Symbol
ADIPOQ-AS1
Entrez Gene ID
100874095
Gene Name
ADIPOQ antisense RNA 1
Gene Aliases
-
UniGene
Hs.638968
Gene Chromosome Location
Chr.3: 186851886-186856123 on Build GRCh38
Transcripts
NM_001177800.1 NM_004797.3 NR_046662.1

SNP Detail

SNP ID Chromosome Location
rs200142714 Chr.3: 186855033-186855033 on Build GRCh38
rs369636003 Chr.3: 186854789-186854789 on Build GRCh38
rs1030524980 Chr.3: 186854821-186854821 on Build GRCh38
rs200726766 Chr.3: 186855196-186855196 on Build GRCh38
rs961071399 Chr.3: 186854872-186854872 on Build GRCh38
rs201402829 Chr.3: 186854740-186854740 on Build GRCh38
rs548289893 Chr.3: 186854740-186854740 on Build GRCh38
rs1018085651 Chr.3: 186855165-186855165 on Build GRCh38
rs760763719 Chr.3: 186854747-186854747 on Build GRCh38
rs199701452 Chr.3: 186855176-186855176 on Build GRCh38
rs397806285 Chr.3: 186855076-186855077 on Build GRCh38
rs936874485 Chr.3: 186855019-186855019 on Build GRCh38
rs758362372 Chr.3: 186854998-186854998 on Build GRCh38
rs972551158 Chr.3: 186854770-186854770 on Build GRCh38
rs199658515 Chr.3: 186855185-186855185 on Build GRCh38
rs367776984 Chr.3: 186854743-186854743 on Build GRCh38
rs201995003 Chr.3: 186854991-186854991 on Build GRCh38
rs56354395 Chr.3: 186855075-186855076 on Build GRCh38
rs955065908 Chr.3: 186854852-186854852 on Build GRCh38
rs923301090 Chr.3: 186854949-186854949 on Build GRCh38
rs192491840 Chr.3: 186854955-186854955 on Build GRCh38
rs200307801 Chr.3: 186854851-186854853 on Build GRCh38
rs1501297 Chr.3: 186855137-186855137 on Build GRCh38
rs1015307005 Chr.3: 186854855-186854855 on Build GRCh38
rs753554665 Chr.3: 186854792-186854792 on Build GRCh38
rs201008329 Chr.3: 186855038-186855038 on Build GRCh38
rs771723603 Chr.3: 186855193-186855193 on Build GRCh38
rs201139232 Chr.3: 186854980-186854980 on Build GRCh38
rs569794385 Chr.3: 186854760-186854760 on Build GRCh38
rs543894441 Chr.3: 186854997-186854997 on Build GRCh38
rs199695100 Chr.3: 186854767-186854767 on Build GRCh38
rs200695138 Chr.3: 186854877-186854877 on Build GRCh38
rs1042210263 Chr.3: 186855002-186855002 on Build GRCh38
rs199678941 Chr.3: 186854907-186854907 on Build GRCh38
rs201407353 Chr.3: 186855169-186855169 on Build GRCh38
rs895412624 Chr.3: 186855035-186855035 on Build GRCh38
rs911158444 Chr.3: 186854861-186854861 on Build GRCh38
rs200346097 Chr.3: 186854759-186854759 on Build GRCh38
rs538819678 Chr.3: 186854846-186854846 on Build GRCh38
rs1054325309 Chr.3: 186855025-186855025 on Build GRCh38
rs540831577 Chr.3: 186855173-186855173 on Build GRCh38
rs201023062 Chr.3: 186855100-186855100 on Build GRCh38
rs565186343 Chr.3: 186855009-186855009 on Build GRCh38
rs949254946 Chr.3: 186855001-186855001 on Build GRCh38
rs184535507 Chr.3: 186854974-186854974 on Build GRCh38
rs752111207 Chr.3: 186854951-186854951 on Build GRCh38
rs4068 Chr.3: 186854769-186854769 on Build GRCh38
rs764259370 Chr.3: 186854750-186854750 on Build GRCh38
rs200775572 Chr.3: 186854936-186854936 on Build GRCh38
rs895379856 Chr.3: 186854746-186854746 on Build GRCh38
rs201687733 Chr.3: 186854883-186854883 on Build GRCh38
rs758853656 Chr.3: 186855045-186855046 on Build GRCh38
rs201686646 Chr.3: 186855146-186855146 on Build GRCh38
rs201246459 Chr.3: 186855179-186855179 on Build GRCh38
rs529994963 Chr.3: 186855131-186855131 on Build GRCh38
rs200502743 Chr.3: 186855177-186855177 on Build GRCh38
rs4686440 Chr.3: 186854891-186854891 on Build GRCh38
rs942552973 Chr.3: 186854862-186854862 on Build GRCh38
rs542641384 Chr.3: 186854926-186854926 on Build GRCh38
rs759155653 Chr.3: 186854801-186854801 on Build GRCh38
rs200429302 Chr.3: 186855153-186855153 on Build GRCh38
rs201671249 Chr.3: 186854961-186854961 on Build GRCh38
rs73888309 Chr.3: 186855075-186855075 on Build GRCh38
rs952544317 Chr.3: 186855088-186855088 on Build GRCh38
rs375881971 Chr.3: 186854948-186854948 on Build GRCh38
rs188947568 Chr.3: 186854793-186854793 on Build GRCh38
rs1042506645 Chr.3: 186855010-186855010 on Build GRCh38
rs752360035 Chr.3: 186854780-186854783 on Build GRCh38
rs1501298 Chr.3: 186854853-186854853 on Build GRCh38
rs576170533 Chr.3: 186854975-186854975 on Build GRCh38
rs199943987 Chr.3: 186854973-186854973 on Build GRCh38
rs1052462527 Chr.3: 186855181-186855181 on Build GRCh38
rs200326826 Chr.3: 186855077-186855077 on Build GRCh38
rs1046632166 Chr.3: 186855047-186855047 on Build GRCh38
rs201026846 Chr.3: 186854766-186854766 on Build GRCh38
rs973747355 Chr.3: 186854909-186854909 on Build GRCh38
rs374215428 Chr.3: 186854883-186854884 on Build GRCh38
rs906230149 Chr.3: 186855069-186855069 on Build GRCh38
rs770841121 Chr.3: 186854738-186854738 on Build GRCh38
rs201816379 Chr.3: 186854745-186854745 on Build GRCh38

View Full Product Details