Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194138195-194138705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HES1
Entrez Gene ID
3280
Gene Name
hes family bHLH transcription factor 1
Gene Aliases
HES-1, HHL, HRY, bHLHb39
UniGene
Hs.250666
Gene Chromosome Location
Chr.3: 194136142-194138612 on Build GRCh38
Transcripts
NM_005524.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6477699 c.827G>T p.R276M Chr.3: 194138217-194138217 on Build GRCh38
5011230 c.805C>A p.L269I Chr.3: 194138195-194138195 on Build GRCh38
71051 c.812C>T p.A271V Chr.3: 194138202-194138202 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755187324 Chr.3: 194138558-194138558 on Build GRCh38
rs777698207 Chr.3: 194138282-194138282 on Build GRCh38
rs970749985 Chr.3: 194138670-194138670 on Build GRCh38
rs180757055 Chr.3: 194138544-194138544 on Build GRCh38
rs140091125 Chr.3: 194138658-194138658 on Build GRCh38
rs1025356231 Chr.3: 194138659-194138659 on Build GRCh38
rs754662672 Chr.3: 194138267-194138267 on Build GRCh38
rs999817608 Chr.3: 194138653-194138653 on Build GRCh38
rs945165722 Chr.3: 194138557-194138557 on Build GRCh38
rs1007042618 Chr.3: 194138620-194138620 on Build GRCh38
rs565442140 Chr.3: 194138343-194138343 on Build GRCh38
rs1017876700 Chr.3: 194138414-194138424 on Build GRCh38
rs146572113 Chr.3: 194138649-194138649 on Build GRCh38
rs984599442 Chr.3: 194138436-194138436 on Build GRCh38
rs983162843 Chr.3: 194138434-194138434 on Build GRCh38
rs971017075 Chr.3: 194138457-194138457 on Build GRCh38
rs750899762 Chr.3: 194138243-194138243 on Build GRCh38
rs889887207 Chr.3: 194138585-194138585 on Build GRCh38
rs918628755 Chr.3: 194138541-194138541 on Build GRCh38
rs1051463168 Chr.3: 194138261-194138261 on Build GRCh38
rs749323414 Chr.3: 194138451-194138451 on Build GRCh38
rs940641317 Chr.3: 194138518-194138518 on Build GRCh38
rs1037425428 Chr.3: 194138519-194138519 on Build GRCh38
rs879147653 Chr.3: 194138295-194138295 on Build GRCh38
rs551670541 Chr.3: 194138393-194138393 on Build GRCh38
rs964210765 Chr.3: 194138386-194138387 on Build GRCh38
rs11541817 Chr.3: 194138414-194138414 on Build GRCh38
rs749749936 Chr.3: 194138233-194138234 on Build GRCh38
rs1018421837 Chr.3: 194138307-194138307 on Build GRCh38
rs751170908 Chr.3: 194138266-194138266 on Build GRCh38
rs765188215 Chr.3: 194138508-194138508 on Build GRCh38
rs747710157 Chr.3: 194138275-194138275 on Build GRCh38
rs978145882 Chr.3: 194138230-194138230 on Build GRCh38
rs926580257 Chr.3: 194138498-194138498 on Build GRCh38
rs371726631 Chr.3: 194138248-194138248 on Build GRCh38
rs916332893 Chr.3: 194138413-194138413 on Build GRCh38
rs546816281 Chr.3: 194138262-194138262 on Build GRCh38
rs1043943658 Chr.3: 194138635-194138635 on Build GRCh38
rs936501360 Chr.3: 194138534-194138534 on Build GRCh38
rs371030330 Chr.3: 194138283-194138283 on Build GRCh38
rs765840154 Chr.3: 194138238-194138238 on Build GRCh38
rs779151380 Chr.3: 194138671-194138671 on Build GRCh38
rs540227829 Chr.3: 194138501-194138502 on Build GRCh38
rs1008447423 Chr.3: 194138589-194138593 on Build GRCh38
rs762400765 Chr.3: 194138235-194138235 on Build GRCh38
rs890139291 Chr.3: 194138564-194138564 on Build GRCh38
rs758733317 Chr.3: 194138249-194138249 on Build GRCh38
rs778900204 Chr.3: 194138275-194138276 on Build GRCh38
rs142811204 Chr.3: 194138391-194138391 on Build GRCh38
rs757778396 Chr.3: 194138234-194138235 on Build GRCh38
rs980657290 Chr.3: 194138464-194138466 on Build GRCh38
rs748299771 Chr.3: 194138679-194138679 on Build GRCh38
rs780654821 Chr.3: 194138270-194138270 on Build GRCh38

View Full Product Details