Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194687989-194688480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM43A
Entrez Gene ID
131583
Gene Name
family with sequence similarity 43 member A
Gene Aliases
-
UniGene
Hs.435080
Gene Chromosome Location
Chr.3: 194685893-194689037 on Build GRCh38
Transcripts
NM_153690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6429550 c.1202C>A p.T401N Chr.3: 194688028-194688028 on Build GRCh38
6381339 c.1207G>A p.A403T Chr.3: 194688033-194688033 on Build GRCh38
7281558 c.1197C>T p.D399D Chr.3: 194688023-194688023 on Build GRCh38
6500451 c.1215C>T p.A405A Chr.3: 194688041-194688041 on Build GRCh38
3974323 c.1248T>C p.S416S Chr.3: 194688074-194688074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571591959 Chr.3: 194688270-194688270 on Build GRCh38
rs959942467 Chr.3: 194688080-194688080 on Build GRCh38
rs757995420 Chr.3: 194688059-194688059 on Build GRCh38
rs185128492 Chr.3: 194688054-194688054 on Build GRCh38
rs539013069 Chr.3: 194688273-194688273 on Build GRCh38
rs1035821750 Chr.3: 194688233-194688233 on Build GRCh38
rs910514948 Chr.3: 194688379-194688379 on Build GRCh38
rs1043247562 Chr.3: 194688188-194688188 on Build GRCh38
rs775992819 Chr.3: 194688041-194688041 on Build GRCh38
rs771229405 Chr.3: 194688023-194688023 on Build GRCh38
rs947625503 Chr.3: 194688403-194688403 on Build GRCh38
rs577609750 Chr.3: 194688319-194688320 on Build GRCh38
rs1043299280 Chr.3: 194688416-194688416 on Build GRCh38
rs998626221 Chr.3: 194688201-194688201 on Build GRCh38
rs894098068 Chr.3: 194688249-194688249 on Build GRCh38
rs867924850 Chr.3: 194688069-194688069 on Build GRCh38
rs747993988 Chr.3: 194688084-194688084 on Build GRCh38
rs1007949519 Chr.3: 194688297-194688297 on Build GRCh38
rs779116440 Chr.3: 194688116-194688116 on Build GRCh38
rs959756353 Chr.3: 194688244-194688244 on Build GRCh38
rs1050726587 Chr.3: 194688158-194688158 on Build GRCh38
rs952694249 Chr.3: 194688361-194688361 on Build GRCh38
rs79296405 Chr.3: 194688092-194688092 on Build GRCh38
rs1021414634 Chr.3: 194688049-194688049 on Build GRCh38
rs774504244 Chr.3: 194688028-194688028 on Build GRCh38
rs1011735195 Chr.3: 194688017-194688017 on Build GRCh38
rs903315910 Chr.3: 194688190-194688190 on Build GRCh38
rs933366397 Chr.3: 194688329-194688329 on Build GRCh38
rs557524670 Chr.3: 194688274-194688274 on Build GRCh38
rs753755026 Chr.3: 194688125-194688126 on Build GRCh38
rs909341297 Chr.3: 194688119-194688119 on Build GRCh38
rs776273624 Chr.3: 194688350-194688350 on Build GRCh38
rs986500156 Chr.3: 194688042-194688042 on Build GRCh38
rs759646259 Chr.3: 194688034-194688034 on Build GRCh38
rs923335095 Chr.3: 194688317-194688317 on Build GRCh38
rs556168852 Chr.3: 194688166-194688166 on Build GRCh38
rs750029279 Chr.3: 194688058-194688058 on Build GRCh38
rs567425883 Chr.3: 194688220-194688220 on Build GRCh38
rs983910453 Chr.3: 194688118-194688118 on Build GRCh38
rs532935945 Chr.3: 194688055-194688055 on Build GRCh38
rs781105753 Chr.3: 194688014-194688014 on Build GRCh38
rs190421742 Chr.3: 194688135-194688135 on Build GRCh38
rs964226485 Chr.3: 194688340-194688340 on Build GRCh38
rs1046496330 Chr.3: 194688184-194688184 on Build GRCh38
rs982679717 Chr.3: 194688365-194688365 on Build GRCh38
rs754980164 Chr.3: 194688081-194688081 on Build GRCh38
rs192109430 Chr.3: 194688308-194688308 on Build GRCh38
rs761305993 Chr.3: 194688045-194688045 on Build GRCh38
rs905376980 Chr.3: 194688194-194688194 on Build GRCh38
rs766042722 Chr.3: 194688063-194688063 on Build GRCh38
rs751027248 Chr.3: 194688065-194688065 on Build GRCh38
rs546832315 Chr.3: 194688262-194688262 on Build GRCh38
rs530650379 Chr.3: 194688153-194688153 on Build GRCh38
rs908563885 Chr.3: 194688415-194688416 on Build GRCh38
rs1027229578 Chr.3: 194688354-194688354 on Build GRCh38
rs747862923 Chr.3: 194688018-194688018 on Build GRCh38
rs919818207 Chr.3: 194688110-194688110 on Build GRCh38
rs926688897 Chr.3: 194688440-194688440 on Build GRCh38
rs73890206 Chr.3: 194688432-194688432 on Build GRCh38
rs777096008 Chr.3: 194688052-194688052 on Build GRCh38
rs915795590 Chr.3: 194688089-194688089 on Build GRCh38
rs1011787603 Chr.3: 194688170-194688170 on Build GRCh38
rs764504812 Chr.3: 194688050-194688050 on Build GRCh38
rs772076724 Chr.3: 194688035-194688035 on Build GRCh38
rs1012983351 Chr.3: 194688261-194688261 on Build GRCh38
rs1036821514 Chr.3: 194688137-194688137 on Build GRCh38
rs991440426 Chr.3: 194688085-194688085 on Build GRCh38
rs908391565 Chr.3: 194688306-194688306 on Build GRCh38
rs949473679 Chr.3: 194688181-194688181 on Build GRCh38
rs996651275 Chr.3: 194688341-194688341 on Build GRCh38
rs772762821 Chr.3: 194688155-194688155 on Build GRCh38
rs770356108 Chr.3: 194688314-194688314 on Build GRCh38
rs534615738 Chr.3: 194688250-194688250 on Build GRCh38
rs1022869985 Chr.3: 194688269-194688269 on Build GRCh38
rs1054449141 Chr.3: 194688206-194688206 on Build GRCh38
rs934805704 Chr.3: 194688435-194688435 on Build GRCh38

View Full Product Details