Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 2963690-2964185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
UniGene
Hs.32959
Gene Chromosome Location
Chr.4: 2963504-3040760 on Build GRCh38
Transcripts
NM_001004056.1 NM_001004057.1 NM_005307.2 NM_182982.2 XM_005247962.3 XM_006713880.3 XM_011513447.2 XM_011513448.2 XM_011513449.2 XM_011513450.2 XM_011513451.2 XM_011513452.2 XM_011513453.2 XM_011513454.2 XM_011513455.2 XM_011513456.2 XM_011513457.2 XM_017008052.1 XM_017008053.1 XM_017008054.1 XM_017008055.1 XM_017008056.1 XM_017008058.1 XM_017008059.1 XM_017008060.1 XM_017008062.1 XM_017008064.1 XM_017008065.1 XM_017008066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5746245 c.123T>C p.A41A Chr.4: 2964105-2964105 on Build GRCh38
4688849 c.107G>A p.R36H Chr.4: 2964089-2964089 on Build GRCh38
5516293 c.40T>G p.S14A Chr.4: 2964022-2964022 on Build GRCh38
7189906 c.140+2T>A p.? Chr.4: 2964124-2964124 on Build GRCh38
5746244 c.35T>C p.L12P Chr.4: 2964105-2964105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905222549 Chr.4: 2964039-2964039 on Build GRCh38
rs779116091 Chr.4: 2963953-2963953 on Build GRCh38
rs901364412 Chr.4: 2963713-2963713 on Build GRCh38
rs763490759 Chr.4: 2964103-2964103 on Build GRCh38
rs1052354097 Chr.4: 2963988-2963988 on Build GRCh38
rs774261636 Chr.4: 2964154-2964154 on Build GRCh38
rs773183375 Chr.4: 2964150-2964150 on Build GRCh38
rs779542767 Chr.4: 2964008-2964008 on Build GRCh38
rs1003394148 Chr.4: 2963728-2963728 on Build GRCh38
rs931601199 Chr.4: 2964158-2964158 on Build GRCh38
rs756011582 Chr.4: 2964014-2964014 on Build GRCh38
rs773860718 Chr.4: 2964156-2964157 on Build GRCh38
rs150529414 Chr.4: 2964074-2964074 on Build GRCh38
rs752220487 Chr.4: 2963992-2963992 on Build GRCh38
rs1024092359 Chr.4: 2963757-2963757 on Build GRCh38
rs768950361 Chr.4: 2963967-2963967 on Build GRCh38
rs758860503 Chr.4: 2964122-2964122 on Build GRCh38
rs753340331 Chr.4: 2964115-2964115 on Build GRCh38
rs142156088 Chr.4: 2964147-2964148 on Build GRCh38
rs770791192 Chr.4: 2964153-2964153 on Build GRCh38
rs1031210892 Chr.4: 2963775-2963775 on Build GRCh38
rs764521757 Chr.4: 2964158-2964158 on Build GRCh38
rs985806094 Chr.4: 2963865-2963865 on Build GRCh38
rs577569608 Chr.4: 2963804-2963804 on Build GRCh38
rs972190958 Chr.4: 2963823-2963823 on Build GRCh38
rs769962087 Chr.4: 2964148-2964148 on Build GRCh38
rs13120295 Chr.4: 2963884-2963884 on Build GRCh38
rs757292288 Chr.4: 2963947-2963947 on Build GRCh38
rs1034908485 Chr.4: 2964066-2964066 on Build GRCh38
rs780759212 Chr.4: 2963991-2963991 on Build GRCh38
rs546530585 Chr.4: 2963943-2963943 on Build GRCh38
rs746331314 Chr.4: 2964011-2964011 on Build GRCh38
rs759272038 Chr.4: 2964086-2964086 on Build GRCh38
rs930429095 Chr.4: 2963863-2963863 on Build GRCh38
rs770980793 Chr.4: 2964052-2964053 on Build GRCh38
rs761916337 Chr.4: 2963969-2963969 on Build GRCh38
rs574844470 Chr.4: 2964049-2964050 on Build GRCh38
rs893885258 Chr.4: 2963954-2963954 on Build GRCh38
rs753867455 Chr.4: 2963945-2963945 on Build GRCh38
rs755609795 Chr.4: 2963995-2963995 on Build GRCh38
rs1010907425 Chr.4: 2963746-2963746 on Build GRCh38
rs1044988632 Chr.4: 2963716-2963716 on Build GRCh38
rs748347102 Chr.4: 2964139-2964139 on Build GRCh38
rs769987888 Chr.4: 2964071-2964071 on Build GRCh38
rs999585137 Chr.4: 2963724-2963724 on Build GRCh38
rs765278018 Chr.4: 2963971-2963971 on Build GRCh38
rs775568801 Chr.4: 2963939-2963939 on Build GRCh38
rs747337441 Chr.4: 2964038-2964038 on Build GRCh38
rs769003409 Chr.4: 2964042-2964042 on Build GRCh38
rs765218914 Chr.4: 2963948-2963948 on Build GRCh38
rs958242757 Chr.4: 2963729-2963729 on Build GRCh38
rs762300034 Chr.4: 2964153-2964154 on Build GRCh38
rs983876172 Chr.4: 2963894-2963894 on Build GRCh38
rs563044095 Chr.4: 2963832-2963832 on Build GRCh38
rs10021191 Chr.4: 2963968-2963968 on Build GRCh38
rs780542048 Chr.4: 2964127-2964127 on Build GRCh38
rs111635147 Chr.4: 2963975-2963975 on Build GRCh38
rs568743246 Chr.4: 2963981-2963981 on Build GRCh38
rs199840994 Chr.4: 2964155-2964155 on Build GRCh38
rs189750996 Chr.4: 2963901-2963901 on Build GRCh38
rs766053438 Chr.4: 2963977-2963977 on Build GRCh38
rs546431906 Chr.4: 2963813-2963813 on Build GRCh38
rs760786893 Chr.4: 2963965-2963965 on Build GRCh38
rs765922759 Chr.4: 2964154-2964155 on Build GRCh38
rs754660701 Chr.4: 2963984-2963984 on Build GRCh38
rs138608466 Chr.4: 2964101-2964101 on Build GRCh38
rs754535957 Chr.4: 2963950-2963950 on Build GRCh38
rs756868180 Chr.4: 2964050-2964052 on Build GRCh38
rs753475880 Chr.4: 2964081-2964081 on Build GRCh38
rs771383653 Chr.4: 2963933-2963933 on Build GRCh38
rs982020306 Chr.4: 2963778-2963778 on Build GRCh38
rs753292642 Chr.4: 2964157-2964157 on Build GRCh38
rs776479037 Chr.4: 2964148-2964148 on Build GRCh38
rs1041337657 Chr.4: 2963913-2963913 on Build GRCh38
rs930822111 Chr.4: 2963885-2963886 on Build GRCh38
rs901311939 Chr.4: 2963917-2963917 on Build GRCh38
rs920176402 Chr.4: 2964146-2964146 on Build GRCh38
rs772536951 Chr.4: 2964022-2964022 on Build GRCh38
rs892559757 Chr.4: 2963799-2963799 on Build GRCh38
rs761762910 Chr.4: 2964146-2964147 on Build GRCh38
rs768430935 Chr.4: 2964094-2964096 on Build GRCh38
rs774716937 Chr.4: 2963937-2963937 on Build GRCh38
rs752345106 Chr.4: 2964096-2964096 on Build GRCh38
rs1034793970 Chr.4: 2963803-2963803 on Build GRCh38
rs755378365 Chr.4: 2964133-2964133 on Build GRCh38
rs759327790 Chr.4: 2964157-2964158 on Build GRCh38
rs376025838 Chr.4: 2964052-2964052 on Build GRCh38
rs749327248 Chr.4: 2964152-2964152 on Build GRCh38
rs79477485 Chr.4: 2964130-2964130 on Build GRCh38
rs759759307 Chr.4: 2963938-2963938 on Build GRCh38
rs374639395 Chr.4: 2964077-2964077 on Build GRCh38
rs761757762 Chr.4: 2963870-2963870 on Build GRCh38
rs538512099 Chr.4: 2963956-2963956 on Build GRCh38
rs780304364 Chr.4: 2964032-2964032 on Build GRCh38
rs767218370 Chr.4: 2964093-2964093 on Build GRCh38
rs13120899 Chr.4: 2964137-2964137 on Build GRCh38
rs1031097131 Chr.4: 2963726-2963726 on Build GRCh38
rs538031762 Chr.4: 2964061-2964061 on Build GRCh38
rs892331865 Chr.4: 2964002-2964002 on Build GRCh38
rs986118518 Chr.4: 2963838-2963838 on Build GRCh38
rs969266903 Chr.4: 2963773-2963773 on Build GRCh38
rs745888367 Chr.4: 2963959-2963960 on Build GRCh38
rs775829839 Chr.4: 2963962-2963962 on Build GRCh38
rs16843684 Chr.4: 2963949-2963949 on Build GRCh38
rs772309969 Chr.4: 2963958-2963958 on Build GRCh38
rs972264036 Chr.4: 2963881-2963881 on Build GRCh38
rs370523679 Chr.4: 2964079-2964079 on Build GRCh38
rs531919230 Chr.4: 2963842-2963842 on Build GRCh38
rs550436937 Chr.4: 2963871-2963871 on Build GRCh38
rs758538455 Chr.4: 2963962-2963968 on Build GRCh38
rs766303986 Chr.4: 2964078-2964078 on Build GRCh38
rs554098887 Chr.4: 2964156-2964156 on Build GRCh38
rs781627042 Chr.4: 2964138-2964138 on Build GRCh38
rs750312486 Chr.4: 2963974-2963974 on Build GRCh38
rs762039266 Chr.4: 2964063-2964063 on Build GRCh38
rs746308109 Chr.4: 2963957-2963957 on Build GRCh38
rs920360106 Chr.4: 2963835-2963835 on Build GRCh38
rs906450413 Chr.4: 2963976-2963983 on Build GRCh38

View Full Product Details