Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 3019484-3019982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK4
Entrez Gene ID
2868
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
GPRK2L, GPRK4, GRK4a, IT11
UniGene
Hs.32959
Gene Chromosome Location
Chr.4: 2963504-3040760 on Build GRCh38
Transcripts
NM_001004056.1 NM_001004057.1 NM_005307.2 NM_182982.2 XM_005247962.3 XM_006713880.3 XM_011513447.2 XM_011513448.2 XM_011513449.2 XM_011513450.2 XM_011513451.2 XM_011513452.2 XM_011513453.2 XM_011513454.2 XM_011513455.2 XM_011513456.2 XM_017008052.1 XM_017008053.1 XM_017008054.1 XM_017008055.1 XM_017008056.1 XM_017008057.1 XM_017008058.1 XM_017008059.1 XM_017008060.1 XM_017008061.1 XM_017008062.1 XM_017008063.1 XM_017008064.1 XM_017008065.1 XM_017008066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6862555 c.932+9G>A p.? Chr.4: 3019840-3019840 on Build GRCh38
342482 c.895G>A p.G299S Chr.4: 3019794-3019794 on Build GRCh38
4802383 c.927A>G p.V309V Chr.4: 3019826-3019826 on Build GRCh38
5576689 c.823C>T p.H275Y Chr.4: 3019722-3019722 on Build GRCh38
4688851 c.783C>T p.C261C Chr.4: 3019682-3019682 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759084345 Chr.4: 3019712-3019712 on Build GRCh38
rs763588250 Chr.4: 3019768-3019768 on Build GRCh38
rs751779365 Chr.4: 3019842-3019842 on Build GRCh38
rs368535647 Chr.4: 3019849-3019849 on Build GRCh38
rs541598491 Chr.4: 3019810-3019810 on Build GRCh38
rs371878306 Chr.4: 3019596-3019596 on Build GRCh38
rs377174142 Chr.4: 3019657-3019657 on Build GRCh38
rs769679366 Chr.4: 3019774-3019774 on Build GRCh38
rs148508490 Chr.4: 3019573-3019573 on Build GRCh38
rs752109205 Chr.4: 3019824-3019824 on Build GRCh38
rs541421364 Chr.4: 3019949-3019949 on Build GRCh38
rs878864791 Chr.4: 3019701-3019701 on Build GRCh38
rs1048767145 Chr.4: 3019943-3019943 on Build GRCh38
rs770692592 Chr.4: 3019874-3019874 on Build GRCh38
rs755103498 Chr.4: 3019845-3019845 on Build GRCh38
rs748135091 Chr.4: 3019773-3019773 on Build GRCh38
rs150897108 Chr.4: 3019789-3019789 on Build GRCh38
rs1009865198 Chr.4: 3019559-3019559 on Build GRCh38
rs780412255 Chr.4: 3019754-3019754 on Build GRCh38
rs765784630 Chr.4: 3019834-3019834 on Build GRCh38
rs781363172 Chr.4: 3019846-3019846 on Build GRCh38
rs561864770 Chr.4: 3019835-3019835 on Build GRCh38
rs761653747 Chr.4: 3019593-3019593 on Build GRCh38
rs944472004 Chr.4: 3019640-3019640 on Build GRCh38
rs201827860 Chr.4: 3019799-3019799 on Build GRCh38
rs934200137 Chr.4: 3019954-3019954 on Build GRCh38
rs144663016 Chr.4: 3019522-3019522 on Build GRCh38
rs930351529 Chr.4: 3019940-3019940 on Build GRCh38
rs928989339 Chr.4: 3019508-3019508 on Build GRCh38
rs971865093 Chr.4: 3019927-3019927 on Build GRCh38
rs758828477 Chr.4: 3019839-3019839 on Build GRCh38
rs919021919 Chr.4: 3019931-3019931 on Build GRCh38
rs371623105 Chr.4: 3019859-3019859 on Build GRCh38
rs761549972 Chr.4: 3019683-3019683 on Build GRCh38
rs868173300 Chr.4: 3019721-3019721 on Build GRCh38
rs773968907 Chr.4: 3019875-3019875 on Build GRCh38
rs746120601 Chr.4: 3019636-3019636 on Build GRCh38
rs3213502 Chr.4: 3019656-3019656 on Build GRCh38
rs772388217 Chr.4: 3019655-3019655 on Build GRCh38
rs961831965 Chr.4: 3019912-3019912 on Build GRCh38
rs376559594 Chr.4: 3019866-3019866 on Build GRCh38
rs201271230 Chr.4: 3019664-3019664 on Build GRCh38
rs779328447 Chr.4: 3019631-3019631 on Build GRCh38
rs763686350 Chr.4: 3019886-3019886 on Build GRCh38
rs773963698 Chr.4: 3019794-3019794 on Build GRCh38
rs190288267 Chr.4: 3019857-3019857 on Build GRCh38
rs772942998 Chr.4: 3019689-3019689 on Build GRCh38
rs765948134 Chr.4: 3019699-3019699 on Build GRCh38
rs778283499 Chr.4: 3019625-3019625 on Build GRCh38
rs371662212 Chr.4: 3019814-3019814 on Build GRCh38
rs749123882 Chr.4: 3019864-3019864 on Build GRCh38
rs748109584 Chr.4: 3019852-3019852 on Build GRCh38
rs765892881 Chr.4: 3019609-3019609 on Build GRCh38
rs757656701 Chr.4: 3019746-3019746 on Build GRCh38
rs986941523 Chr.4: 3019544-3019544 on Build GRCh38
rs868582090 Chr.4: 3019744-3019744 on Build GRCh38
rs577114989 Chr.4: 3019753-3019753 on Build GRCh38
rs185967676 Chr.4: 3019748-3019748 on Build GRCh38
rs751458381 Chr.4: 3019836-3019836 on Build GRCh38
rs1039852556 Chr.4: 3019590-3019590 on Build GRCh38
rs757040676 Chr.4: 3019840-3019840 on Build GRCh38
rs910160206 Chr.4: 3019561-3019561 on Build GRCh38
rs762623772 Chr.4: 3019695-3019695 on Build GRCh38
rs201996842 Chr.4: 3019854-3019854 on Build GRCh38
rs930279433 Chr.4: 3019700-3019700 on Build GRCh38
rs757952139 Chr.4: 3019608-3019608 on Build GRCh38
rs756580603 Chr.4: 3019624-3019624 on Build GRCh38
rs116468759 Chr.4: 3019953-3019953 on Build GRCh38
rs941500378 Chr.4: 3019576-3019576 on Build GRCh38
rs777672543 Chr.4: 3019860-3019860 on Build GRCh38
rs867838234 Chr.4: 3019691-3019691 on Build GRCh38
rs141637189 Chr.4: 3019745-3019745 on Build GRCh38
rs764549771 Chr.4: 3019723-3019723 on Build GRCh38
rs751081081 Chr.4: 3019710-3019710 on Build GRCh38
rs751026513 Chr.4: 3019611-3019611 on Build GRCh38
rs912766851 Chr.4: 3019894-3019894 on Build GRCh38
rs750046644 Chr.4: 3019601-3019601 on Build GRCh38
rs777712417 Chr.4: 3019785-3019785 on Build GRCh38
rs750885539 Chr.4: 3019743-3019743 on Build GRCh38
rs1051180667 Chr.4: 3019957-3019957 on Build GRCh38
rs201879521 Chr.4: 3019769-3019769 on Build GRCh38
rs545770050 Chr.4: 3019767-3019767 on Build GRCh38
rs375066699 Chr.4: 3019882-3019882 on Build GRCh38
rs747122424 Chr.4: 3019766-3019766 on Build GRCh38
rs769605390 Chr.4: 3019686-3019686 on Build GRCh38
rs575812899 Chr.4: 3019784-3019784 on Build GRCh38
rs770625534 Chr.4: 3019793-3019793 on Build GRCh38
rs113476947 Chr.4: 3019735-3019735 on Build GRCh38
rs779456235 Chr.4: 3019747-3019747 on Build GRCh38
rs534375159 Chr.4: 3019733-3019733 on Build GRCh38
rs1047310278 Chr.4: 3019538-3019538 on Build GRCh38
rs757612075 Chr.4: 3019627-3019627 on Build GRCh38

View Full Product Details