Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116373740-116374268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1A1
Entrez Gene ID
476
Gene Name
ATPase Na+/K+ transporting subunit alpha 1
Gene Aliases
-
UniGene
Hs.371889
Gene Chromosome Location
Chr.1: 116373173-116404774 on Build GRCh38
Transcripts
NM_000701.7 NM_001160233.1

SNP Detail

SNP ID Chromosome Location
rs569399389 Chr.1: 116374149-116374149 on Build GRCh38
rs368964935 Chr.1: 116373795-116373795 on Build GRCh38
rs992587877 Chr.1: 116373938-116373938 on Build GRCh38
rs895828368 Chr.1: 116374100-116374100 on Build GRCh38
rs958241530 Chr.1: 116374159-116374159 on Build GRCh38
rs749166380 Chr.1: 116374023-116374024 on Build GRCh38
rs568137425 Chr.1: 116373909-116373909 on Build GRCh38
rs1032612380 Chr.1: 116374139-116374139 on Build GRCh38
rs1001442767 Chr.1: 116374076-116374076 on Build GRCh38
rs908013299 Chr.1: 116374055-116374055 on Build GRCh38
rs905832874 Chr.1: 116373876-116373876 on Build GRCh38
rs925237598 Chr.1: 116373966-116373966 on Build GRCh38
rs569948355 Chr.1: 116374007-116374007 on Build GRCh38
rs1057147704 Chr.1: 116374091-116374091 on Build GRCh38
rs900519692 Chr.1: 116373768-116373768 on Build GRCh38
rs1000690709 Chr.1: 116374129-116374129 on Build GRCh38
rs974950030 Chr.1: 116374169-116374169 on Build GRCh38
rs1016109576 Chr.1: 116373862-116373862 on Build GRCh38
rs1021173995 Chr.1: 116373945-116373945 on Build GRCh38
rs1052322958 Chr.1: 116373805-116373805 on Build GRCh38
rs942227975 Chr.1: 116374067-116374067 on Build GRCh38
rs1004985643 Chr.1: 116374053-116374053 on Build GRCh38
rs906096885 Chr.1: 116374083-116374083 on Build GRCh38
rs943316663 Chr.1: 116373978-116373978 on Build GRCh38
rs760703206 Chr.1: 116374206-116374206 on Build GRCh38
rs1019967187 Chr.1: 116374116-116374116 on Build GRCh38
rs954397634 Chr.1: 116374176-116374176 on Build GRCh38
rs535580718 Chr.1: 116373940-116373940 on Build GRCh38
rs113873221 Chr.1: 116373791-116373791 on Build GRCh38
rs11289194 Chr.1: 116374024-116374024 on Build GRCh38
rs1010285622 Chr.1: 116374105-116374105 on Build GRCh38
rs867902778 Chr.1: 116373857-116373857 on Build GRCh38
rs540080561 Chr.1: 116374152-116374152 on Build GRCh38
rs951216753 Chr.1: 116373778-116373778 on Build GRCh38
rs551148528 Chr.1: 116374124-116374124 on Build GRCh38
rs960109832 Chr.1: 116373913-116373913 on Build GRCh38
rs1001826601 Chr.1: 116374087-116374087 on Build GRCh38
rs150267900 Chr.1: 116374229-116374229 on Build GRCh38
rs528107648 Chr.1: 116373877-116373877 on Build GRCh38
rs1005153699 Chr.1: 116373839-116373839 on Build GRCh38
rs376117429 Chr.1: 116374228-116374228 on Build GRCh38
rs774764129 Chr.1: 116374185-116374185 on Build GRCh38
rs745481832 Chr.1: 116373958-116373958 on Build GRCh38
rs112392332 Chr.1: 116374039-116374039 on Build GRCh38
rs865816205 Chr.1: 116373794-116373794 on Build GRCh38
rs941802169 Chr.1: 116374210-116374210 on Build GRCh38
rs922905482 Chr.1: 116374161-116374161 on Build GRCh38
rs907870037 Chr.1: 116374234-116374234 on Build GRCh38
rs964594182 Chr.1: 116374144-116374144 on Build GRCh38
rs1038306384 Chr.1: 116374074-116374074 on Build GRCh38
rs1020384445 Chr.1: 116374117-116374117 on Build GRCh38
rs966570726 Chr.1: 116373951-116373951 on Build GRCh38
rs886257444 Chr.1: 116373812-116373812 on Build GRCh38
rs974358825 Chr.1: 116374003-116374003 on Build GRCh38
rs974664181 Chr.1: 116374147-116374147 on Build GRCh38
rs895464202 Chr.1: 116374104-116374104 on Build GRCh38
rs1047478408 Chr.1: 116374044-116374044 on Build GRCh38
rs868183236 Chr.1: 116373957-116373957 on Build GRCh38
rs755822194 Chr.1: 116374200-116374200 on Build GRCh38
rs183880157 Chr.1: 116373893-116373893 on Build GRCh38
rs1032465247 Chr.1: 116374158-116374158 on Build GRCh38
rs933060940 Chr.1: 116374017-116374017 on Build GRCh38
rs936890803 Chr.1: 116374001-116374001 on Build GRCh38
rs370963656 Chr.1: 116374137-116374137 on Build GRCh38
rs1038972470 Chr.1: 116373759-116373759 on Build GRCh38
rs1034306902 Chr.1: 116374086-116374086 on Build GRCh38
rs963266096 Chr.1: 116373881-116373881 on Build GRCh38
rs922363554 Chr.1: 116373763-116373763 on Build GRCh38
rs1014364769 Chr.1: 116374115-116374115 on Build GRCh38

View Full Product Details