Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8292181-8292693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA3
Entrez Gene ID
94031
Gene Name
HtrA serine peptidase 3
Gene Aliases
Prsp, Tasp
UniGene
Hs.479119
Gene Chromosome Location
Chr.4: 8269757-8307111 on Build GRCh38
Transcripts
NM_001297559.1 NM_053044.4 XM_011513596.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683097 c.915C>T p.S305S Chr.4: 8292332-8292332 on Build GRCh38
6253755 c.904-4G>T p.? Chr.4: 8292317-8292317 on Build GRCh38
6192878 c.912C>T p.N304N Chr.4: 8292329-8292329 on Build GRCh38
4792312 c.918G>T p.G306G Chr.4: 8292335-8292335 on Build GRCh38
4792311 c.918G>T p.G306G Chr.4: 8292335-8292335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774930128 Chr.4: 8292395-8292395 on Build GRCh38
rs374106060 Chr.4: 8292612-8292612 on Build GRCh38
rs368909714 Chr.4: 8292416-8292416 on Build GRCh38
rs969543902 Chr.4: 8292519-8292519 on Build GRCh38
rs762165356 Chr.4: 8292398-8292398 on Build GRCh38
rs13130633 Chr.4: 8292469-8292469 on Build GRCh38
rs763597664 Chr.4: 8292300-8292300 on Build GRCh38
rs746306918 Chr.4: 8292545-8292545 on Build GRCh38
rs1013082603 Chr.4: 8292630-8292630 on Build GRCh38
rs1048355014 Chr.4: 8292572-8292572 on Build GRCh38
rs528802985 Chr.4: 8292560-8292560 on Build GRCh38
rs575767072 Chr.4: 8292490-8292490 on Build GRCh38
rs1035465921 Chr.4: 8292379-8292379 on Build GRCh38
rs558767790 Chr.4: 8292277-8292277 on Build GRCh38
rs749659601 Chr.4: 8292378-8292378 on Build GRCh38
rs1025182980 Chr.4: 8292503-8292503 on Build GRCh38
rs772919295 Chr.4: 8292318-8292318 on Build GRCh38
rs972450390 Chr.4: 8292571-8292571 on Build GRCh38
rs776049578 Chr.4: 8292369-8292369 on Build GRCh38
rs530688784 Chr.4: 8292614-8292614 on Build GRCh38
rs933365866 Chr.4: 8292581-8292581 on Build GRCh38
rs546085213 Chr.4: 8292524-8292524 on Build GRCh38
rs73083639 Chr.4: 8292443-8292443 on Build GRCh38
rs753805351 Chr.4: 8292491-8292491 on Build GRCh38
rs775961333 Chr.4: 8292405-8292405 on Build GRCh38
rs374869630 Chr.4: 8292363-8292363 on Build GRCh38
rs757372580 Chr.4: 8292353-8292353 on Build GRCh38
rs776907841 Chr.4: 8292281-8292281 on Build GRCh38
rs367887152 Chr.4: 8292323-8292323 on Build GRCh38
rs752457550 Chr.4: 8292344-8292344 on Build GRCh38
rs992713941 Chr.4: 8292221-8292221 on Build GRCh38
rs769357281 Chr.4: 8292382-8292382 on Build GRCh38
rs386671348 Chr.4: 8292560-8292561 on Build GRCh38
rs746020679 Chr.4: 8292282-8292282 on Build GRCh38
rs369251365 Chr.4: 8292381-8292381 on Build GRCh38
rs1006794503 Chr.4: 8292450-8292450 on Build GRCh38
rs775959556 Chr.4: 8292299-8292299 on Build GRCh38
rs770305800 Chr.4: 8292285-8292285 on Build GRCh38
rs556796165 Chr.4: 8292474-8292474 on Build GRCh38
rs770665456 Chr.4: 8292377-8292378 on Build GRCh38
rs919351274 Chr.4: 8292573-8292573 on Build GRCh38
rs745703718 Chr.4: 8292324-8292324 on Build GRCh38
rs564763218 Chr.4: 8292529-8292529 on Build GRCh38
rs1023782162 Chr.4: 8292631-8292631 on Build GRCh38
rs376097355 Chr.4: 8292295-8292295 on Build GRCh38
rs892123274 Chr.4: 8292660-8292660 on Build GRCh38
rs753538326 Chr.4: 8292332-8292332 on Build GRCh38
rs146222685 Chr.4: 8292606-8292606 on Build GRCh38
rs777674430 Chr.4: 8292350-8292350 on Build GRCh38
rs758378129 Chr.4: 8292345-8292345 on Build GRCh38
rs918177478 Chr.4: 8292245-8292245 on Build GRCh38
rs775804901 Chr.4: 8292395-8292397 on Build GRCh38
rs540232625 Chr.4: 8292561-8292561 on Build GRCh38
rs979643039 Chr.4: 8292651-8292651 on Build GRCh38
rs1003934744 Chr.4: 8292376-8292376 on Build GRCh38
rs912685170 Chr.4: 8292530-8292530 on Build GRCh38
rs4235258 Chr.4: 8292247-8292247 on Build GRCh38
rs981012506 Chr.4: 8292331-8292331 on Build GRCh38
rs943859880 Chr.4: 8292534-8292534 on Build GRCh38
rs548509936 Chr.4: 8292333-8292333 on Build GRCh38
rs765043929 Chr.4: 8292341-8292341 on Build GRCh38
rs200341931 Chr.4: 8292263-8292264 on Build GRCh38
rs138967430 Chr.4: 8292402-8292402 on Build GRCh38
rs558744259 Chr.4: 8292473-8292473 on Build GRCh38
rs969543230 Chr.4: 8292642-8292642 on Build GRCh38
rs909252021 Chr.4: 8292358-8292358 on Build GRCh38
rs747124417 Chr.4: 8292351-8292351 on Build GRCh38
rs199583748 Chr.4: 8292359-8292359 on Build GRCh38
rs374023395 Chr.4: 8292335-8292335 on Build GRCh38
rs188716305 Chr.4: 8292567-8292567 on Build GRCh38
rs141608184 Chr.4: 8292602-8292602 on Build GRCh38
rs570489280 Chr.4: 8292549-8292549 on Build GRCh38
rs886791495 Chr.4: 8292373-8292373 on Build GRCh38
rs1030770584 Chr.4: 8292481-8292481 on Build GRCh38
rs774323563 Chr.4: 8292254-8292254 on Build GRCh38

View Full Product Details