Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9781915-9782440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3607186 c.315T>C p.G105G Chr.4: 9782344-9782344 on Build GRCh38
5549174 c.65C>A p.A22E Chr.4: 9782094-9782094 on Build GRCh38
6649304 c.376G>A p.A126T Chr.4: 9782405-9782405 on Build GRCh38
4679377 c.181G>A p.V61M Chr.4: 9782210-9782210 on Build GRCh38
7427554 c.264T>C p.L88L Chr.4: 9782293-9782293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770087035 Chr.4: 9781999-9781999 on Build GRCh38
rs755213948 Chr.4: 9782059-9782059 on Build GRCh38
rs144460797 Chr.4: 9782168-9782168 on Build GRCh38
rs775440942 Chr.4: 9782336-9782336 on Build GRCh38
rs780448872 Chr.4: 9782000-9782000 on Build GRCh38
rs780561302 Chr.4: 9782233-9782233 on Build GRCh38
rs752541804 Chr.4: 9782206-9782206 on Build GRCh38
rs199687249 Chr.4: 9782191-9782191 on Build GRCh38
rs761016838 Chr.4: 9782420-9782420 on Build GRCh38
rs777868525 Chr.4: 9782107-9782107 on Build GRCh38
rs377175551 Chr.4: 9782204-9782204 on Build GRCh38
rs766183341 Chr.4: 9782097-9782097 on Build GRCh38
rs763661600 Chr.4: 9782083-9782083 on Build GRCh38
rs772783654 Chr.4: 9782190-9782190 on Build GRCh38
rs565360620 Chr.4: 9782056-9782056 on Build GRCh38
rs769659017 Chr.4: 9782335-9782335 on Build GRCh38
rs1011181846 Chr.4: 9782087-9782091 on Build GRCh38
rs1023663186 Chr.4: 9782115-9782115 on Build GRCh38
rs749506188 Chr.4: 9782050-9782050 on Build GRCh38
rs747990243 Chr.4: 9782406-9782406 on Build GRCh38
rs756513171 Chr.4: 9782283-9782283 on Build GRCh38
rs757198240 Chr.4: 9782034-9782034 on Build GRCh38
rs761830697 Chr.4: 9782300-9782300 on Build GRCh38
rs1014821510 Chr.4: 9782269-9782269 on Build GRCh38
rs750197120 Chr.4: 9782040-9782040 on Build GRCh38
rs765682568 Chr.4: 9782367-9782367 on Build GRCh38
rs746619432 Chr.4: 9782301-9782301 on Build GRCh38
rs563430281 Chr.4: 9782024-9782024 on Build GRCh38
rs990837435 Chr.4: 9781972-9781972 on Build GRCh38
rs764915017 Chr.4: 9782203-9782203 on Build GRCh38
rs2227840 Chr.4: 9782214-9782214 on Build GRCh38
rs773857136 Chr.4: 9782084-9782084 on Build GRCh38
rs772138219 Chr.4: 9782409-9782409 on Build GRCh38
rs985876615 Chr.4: 9782375-9782375 on Build GRCh38
rs781612446 Chr.4: 9782110-9782110 on Build GRCh38
rs758067205 Chr.4: 9782234-9782234 on Build GRCh38
rs769046341 Chr.4: 9782174-9782174 on Build GRCh38
rs534438513 Chr.4: 9782352-9782352 on Build GRCh38
rs756987933 Chr.4: 9782314-9782314 on Build GRCh38
rs756703093 Chr.4: 9782225-9782225 on Build GRCh38
rs780559451 Chr.4: 9782329-9782329 on Build GRCh38
rs776905009 Chr.4: 9782078-9782078 on Build GRCh38
rs6282 Chr.4: 9782292-9782292 on Build GRCh38
rs372681931 Chr.4: 9781935-9781935 on Build GRCh38
rs76288744 Chr.4: 9782293-9782293 on Build GRCh38
rs113695344 Chr.4: 9782347-9782347 on Build GRCh38
rs745775968 Chr.4: 9782252-9782252 on Build GRCh38
rs201023282 Chr.4: 9782251-9782251 on Build GRCh38
rs747790549 Chr.4: 9782184-9782184 on Build GRCh38
rs760170081 Chr.4: 9782365-9782365 on Build GRCh38
rs768309664 Chr.4: 9782124-9782124 on Build GRCh38
rs748181038 Chr.4: 9782060-9782060 on Build GRCh38
rs184288806 Chr.4: 9782353-9782353 on Build GRCh38
rs755160094 Chr.4: 9782099-9782099 on Build GRCh38
rs757728622 Chr.4: 9782166-9782166 on Build GRCh38
rs927943382 Chr.4: 9781947-9781947 on Build GRCh38
rs756368276 Chr.4: 9782186-9782186 on Build GRCh38
rs2227848 Chr.4: 9782281-9782281 on Build GRCh38
rs771111537 Chr.4: 9782421-9782421 on Build GRCh38
rs780517411 Chr.4: 9782179-9782179 on Build GRCh38
rs2227849 Chr.4: 9782358-9782358 on Build GRCh38
rs201926104 Chr.4: 9782178-9782178 on Build GRCh38
rs147038465 Chr.4: 9782330-9782330 on Build GRCh38
rs536904632 Chr.4: 9782158-9782158 on Build GRCh38
rs772510703 Chr.4: 9782074-9782074 on Build GRCh38
rs750875411 Chr.4: 9782218-9782218 on Build GRCh38
rs767483503 Chr.4: 9782278-9782278 on Build GRCh38
rs759557243 Chr.4: 9782200-9782200 on Build GRCh38
rs749266929 Chr.4: 9782338-9782338 on Build GRCh38
rs749213890 Chr.4: 9782397-9782397 on Build GRCh38
rs2227839 Chr.4: 9782192-9782192 on Build GRCh38
rs151282040 Chr.4: 9782253-9782253 on Build GRCh38
rs767679526 Chr.4: 9782217-9782217 on Build GRCh38
rs541886645 Chr.4: 9782369-9782369 on Build GRCh38
rs762147902 Chr.4: 9782277-9782277 on Build GRCh38
rs748676694 Chr.4: 9782020-9782020 on Build GRCh38
rs780035670 Chr.4: 9782041-9782041 on Build GRCh38
rs542146372 Chr.4: 9782009-9782009 on Build GRCh38
rs776463741 Chr.4: 9782199-9782199 on Build GRCh38
rs960657255 Chr.4: 9781970-9781970 on Build GRCh38
rs748208599 Chr.4: 9782103-9782103 on Build GRCh38
rs567268264 Chr.4: 9782129-9782129 on Build GRCh38
rs200812406 Chr.4: 9782025-9782025 on Build GRCh38
rs772633407 Chr.4: 9782023-9782023 on Build GRCh38
rs749289106 Chr.4: 9782008-9782008 on Build GRCh38
rs777590080 Chr.4: 9782243-9782243 on Build GRCh38
rs761938234 Chr.4: 9782351-9782351 on Build GRCh38
rs773728722 Chr.4: 9782075-9782075 on Build GRCh38
rs766233243 Chr.4: 9782137-9782137 on Build GRCh38
rs753609229 Chr.4: 9782138-9782138 on Build GRCh38
rs374043983 Chr.4: 9782374-9782374 on Build GRCh38
rs1012374541 Chr.4: 9782246-9782246 on Build GRCh38
rs565962667 Chr.4: 9782125-9782125 on Build GRCh38
rs755593107 Chr.4: 9782377-9782377 on Build GRCh38
rs570059380 Chr.4: 9782215-9782215 on Build GRCh38
rs370317852 Chr.4: 9782368-9782368 on Build GRCh38
rs148402761 Chr.4: 9782291-9782291 on Build GRCh38
rs984757325 Chr.4: 9782122-9782122 on Build GRCh38
rs746485772 Chr.4: 9782245-9782245 on Build GRCh38
rs201570391 Chr.4: 9782361-9782361 on Build GRCh38
rs746235418 Chr.4: 9782112-9782112 on Build GRCh38
rs756170920 Chr.4: 9782096-9782096 on Build GRCh38
rs772917283 Chr.4: 9782130-9782130 on Build GRCh38
rs768648199 Chr.4: 9782402-9782402 on Build GRCh38
rs758502580 Chr.4: 9782104-9782104 on Build GRCh38
rs775197050 Chr.4: 9782256-9782256 on Build GRCh38
rs755423087 Chr.4: 9782295-9782295 on Build GRCh38
rs773513412 Chr.4: 9782416-9782416 on Build GRCh38
rs768752880 Chr.4: 9782013-9782013 on Build GRCh38
rs764302520 Chr.4: 9782212-9782212 on Build GRCh38
rs372277553 Chr.4: 9782372-9782372 on Build GRCh38
rs774529086 Chr.4: 9782262-9782262 on Build GRCh38
rs779016797 Chr.4: 9782101-9782101 on Build GRCh38
rs751702807 Chr.4: 9782164-9782164 on Build GRCh38
rs371107353 Chr.4: 9782081-9782081 on Build GRCh38
rs761432622 Chr.4: 9782091-9782091 on Build GRCh38
rs1023630531 Chr.4: 9782250-9782250 on Build GRCh38
rs768215154 Chr.4: 9782260-9782260 on Build GRCh38
rs774344700 Chr.4: 9782014-9782014 on Build GRCh38
rs1049349176 Chr.4: 9782001-9782001 on Build GRCh38
rs767525523 Chr.4: 9782092-9782092 on Build GRCh38
rs145405809 Chr.4: 9782194-9782194 on Build GRCh38
rs751086015 Chr.4: 9782027-9782027 on Build GRCh38
rs2227841 Chr.4: 9782404-9782404 on Build GRCh38
rs368710755 Chr.4: 9781998-9781998 on Build GRCh38
rs761601511 Chr.4: 9782126-9782126 on Build GRCh38
rs201330876 Chr.4: 9782151-9782151 on Build GRCh38
rs771252628 Chr.4: 9782076-9782076 on Build GRCh38
rs747489876 Chr.4: 9782108-9782108 on Build GRCh38
rs191825696 Chr.4: 9782095-9782095 on Build GRCh38
rs764263788 Chr.4: 9782261-9782261 on Build GRCh38
rs952051787 Chr.4: 9782121-9782121 on Build GRCh38
rs975877349 Chr.4: 9782296-9782296 on Build GRCh38
rs765459092 Chr.4: 9782152-9782152 on Build GRCh38
rs771481750 Chr.4: 9782109-9782109 on Build GRCh38
rs777815501 Chr.4: 9782161-9782161 on Build GRCh38
rs113223975 Chr.4: 9782210-9782210 on Build GRCh38
rs767580702 Chr.4: 9782035-9782035 on Build GRCh38
rs559099689 Chr.4: 9781980-9781980 on Build GRCh38
rs771687760 Chr.4: 9782127-9782127 on Build GRCh38
rs753644106 Chr.4: 9782098-9782098 on Build GRCh38
rs368348754 Chr.4: 9782026-9782026 on Build GRCh38
rs779592809 Chr.4: 9782380-9782380 on Build GRCh38
rs147655139 Chr.4: 9782345-9782345 on Build GRCh38

View Full Product Details