Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9782257-9782765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4572350 c.719T>G p.I240S Chr.4: 9782748-9782748 on Build GRCh38
3607186 c.315T>C p.G105G Chr.4: 9782344-9782344 on Build GRCh38
6649304 c.376G>A p.A126T Chr.4: 9782405-9782405 on Build GRCh38
7427554 c.264T>C p.L88L Chr.4: 9782293-9782293 on Build GRCh38
3607188 c.381C>T p.S127S Chr.4: 9782410-9782410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775440942 Chr.4: 9782336-9782336 on Build GRCh38
rs745901421 Chr.4: 9782583-9782583 on Build GRCh38
rs761016838 Chr.4: 9782420-9782420 on Build GRCh38
rs556526603 Chr.4: 9782493-9782493 on Build GRCh38
rs769659017 Chr.4: 9782335-9782335 on Build GRCh38
rs532610655 Chr.4: 9782557-9782557 on Build GRCh38
rs765976922 Chr.4: 9782614-9782614 on Build GRCh38
rs935592815 Chr.4: 9782451-9782451 on Build GRCh38
rs756513171 Chr.4: 9782283-9782283 on Build GRCh38
rs747990243 Chr.4: 9782406-9782406 on Build GRCh38
rs531122680 Chr.4: 9782704-9782704 on Build GRCh38
rs761830697 Chr.4: 9782300-9782300 on Build GRCh38
rs375459639 Chr.4: 9782556-9782556 on Build GRCh38
rs777620276 Chr.4: 9782575-9782575 on Build GRCh38
rs769898539 Chr.4: 9782592-9782592 on Build GRCh38
rs765682568 Chr.4: 9782367-9782367 on Build GRCh38
rs779016278 Chr.4: 9782742-9782742 on Build GRCh38
rs746619432 Chr.4: 9782301-9782301 on Build GRCh38
rs769335952 Chr.4: 9782696-9782696 on Build GRCh38
rs760112914 Chr.4: 9782433-9782433 on Build GRCh38
rs368255650 Chr.4: 9782443-9782443 on Build GRCh38
rs746801776 Chr.4: 9782576-9782576 on Build GRCh38
rs2227846 Chr.4: 9782659-9782659 on Build GRCh38
rs137945596 Chr.4: 9782660-9782660 on Build GRCh38
rs757493976 Chr.4: 9782579-9782579 on Build GRCh38
rs149304627 Chr.4: 9782539-9782539 on Build GRCh38
rs772138219 Chr.4: 9782409-9782409 on Build GRCh38
rs771867152 Chr.4: 9782499-9782499 on Build GRCh38
rs985876615 Chr.4: 9782375-9782375 on Build GRCh38
rs935559976 Chr.4: 9782709-9782709 on Build GRCh38
rs200733038 Chr.4: 9782553-9782553 on Build GRCh38
rs1030113370 Chr.4: 9782586-9782586 on Build GRCh38
rs534438513 Chr.4: 9782352-9782352 on Build GRCh38
rs756987933 Chr.4: 9782314-9782314 on Build GRCh38
rs538751369 Chr.4: 9782502-9782502 on Build GRCh38
rs780559451 Chr.4: 9782329-9782329 on Build GRCh38
rs6282 Chr.4: 9782292-9782292 on Build GRCh38
rs764080313 Chr.4: 9782711-9782711 on Build GRCh38
rs76288744 Chr.4: 9782293-9782293 on Build GRCh38
rs113695344 Chr.4: 9782347-9782347 on Build GRCh38
rs868700197 Chr.4: 9782463-9782463 on Build GRCh38
rs752655773 Chr.4: 9782565-9782565 on Build GRCh38
rs781585108 Chr.4: 9782580-9782580 on Build GRCh38
rs753915673 Chr.4: 9782740-9782740 on Build GRCh38
rs760170081 Chr.4: 9782365-9782365 on Build GRCh38
rs753351371 Chr.4: 9782485-9782485 on Build GRCh38
rs184288806 Chr.4: 9782353-9782353 on Build GRCh38
rs2227848 Chr.4: 9782281-9782281 on Build GRCh38
rs771111537 Chr.4: 9782421-9782421 on Build GRCh38
rs763394332 Chr.4: 9782450-9782450 on Build GRCh38
rs2227849 Chr.4: 9782358-9782358 on Build GRCh38
rs147038465 Chr.4: 9782330-9782330 on Build GRCh38
rs1015371380 Chr.4: 9782552-9782552 on Build GRCh38
rs774641147 Chr.4: 9782603-9782603 on Build GRCh38
rs764692136 Chr.4: 9782563-9782563 on Build GRCh38
rs749266929 Chr.4: 9782338-9782338 on Build GRCh38
rs749213890 Chr.4: 9782397-9782397 on Build GRCh38
rs776780535 Chr.4: 9782524-9782524 on Build GRCh38
rs754515760 Chr.4: 9782560-9782560 on Build GRCh38
rs368793452 Chr.4: 9782644-9782644 on Build GRCh38
rs199843999 Chr.4: 9782731-9782731 on Build GRCh38
rs781170757 Chr.4: 9782630-9782630 on Build GRCh38
rs541886645 Chr.4: 9782369-9782369 on Build GRCh38
rs759697803 Chr.4: 9782538-9782538 on Build GRCh38
rs759158462 Chr.4: 9782615-9782615 on Build GRCh38
rs773622534 Chr.4: 9782673-9782673 on Build GRCh38
rs144132215 Chr.4: 9782597-9782597 on Build GRCh38
rs571550831 Chr.4: 9782694-9782694 on Build GRCh38
rs759221755 Chr.4: 9782685-9782685 on Build GRCh38
rs763423111 Chr.4: 9782548-9782548 on Build GRCh38
rs778933591 Chr.4: 9782494-9782494 on Build GRCh38
rs530028740 Chr.4: 9782627-9782627 on Build GRCh38
rs769157941 Chr.4: 9782656-9782656 on Build GRCh38
rs761938234 Chr.4: 9782351-9782351 on Build GRCh38
rs764442712 Chr.4: 9782617-9782617 on Build GRCh38
rs374043983 Chr.4: 9782374-9782374 on Build GRCh38
rs755593107 Chr.4: 9782377-9782377 on Build GRCh38
rs370317852 Chr.4: 9782368-9782368 on Build GRCh38
rs765839800 Chr.4: 9782441-9782441 on Build GRCh38
rs148402761 Chr.4: 9782291-9782291 on Build GRCh38
rs201570391 Chr.4: 9782361-9782361 on Build GRCh38
rs371771975 Chr.4: 9782735-9782735 on Build GRCh38
rs761248882 Chr.4: 9782566-9782566 on Build GRCh38
rs777476781 Chr.4: 9782503-9782503 on Build GRCh38
rs575640830 Chr.4: 9782474-9782474 on Build GRCh38
rs761902155 Chr.4: 9782725-9782725 on Build GRCh38
rs768648199 Chr.4: 9782402-9782402 on Build GRCh38
rs187886787 Chr.4: 9782622-9782622 on Build GRCh38
rs569998475 Chr.4: 9782662-9782662 on Build GRCh38
rs755423087 Chr.4: 9782295-9782295 on Build GRCh38
rs372770621 Chr.4: 9782618-9782618 on Build GRCh38
rs773513412 Chr.4: 9782416-9782416 on Build GRCh38
rs747220811 Chr.4: 9782506-9782506 on Build GRCh38
rs762388093 Chr.4: 9782705-9782705 on Build GRCh38
rs372277553 Chr.4: 9782372-9782372 on Build GRCh38
rs769051986 Chr.4: 9782601-9782601 on Build GRCh38
rs2227852 Chr.4: 9782741-9782741 on Build GRCh38
rs751346520 Chr.4: 9782626-9782626 on Build GRCh38
rs776907638 Chr.4: 9782426-9782426 on Build GRCh38
rs751508044 Chr.4: 9782719-9782719 on Build GRCh38
rs546007578 Chr.4: 9782477-9782477 on Build GRCh38
rs865994568 Chr.4: 9782526-9782526 on Build GRCh38
rs760254927 Chr.4: 9782555-9782555 on Build GRCh38
rs369087783 Chr.4: 9782595-9782595 on Build GRCh38
rs763947953 Chr.4: 9782623-9782623 on Build GRCh38
rs2227841 Chr.4: 9782404-9782404 on Build GRCh38
rs762189023 Chr.4: 9782604-9782604 on Build GRCh38
rs773390586 Chr.4: 9782442-9782442 on Build GRCh38
rs143535200 Chr.4: 9782540-9782540 on Build GRCh38
rs2227843 Chr.4: 9782727-9782727 on Build GRCh38
rs2227845 Chr.4: 9782648-9782648 on Build GRCh38
rs975877349 Chr.4: 9782296-9782296 on Build GRCh38
rs774650319 Chr.4: 9782554-9782554 on Build GRCh38
rs771346209 Chr.4: 9782513-9782513 on Build GRCh38
rs750070766 Chr.4: 9782464-9782464 on Build GRCh38
rs780118849 Chr.4: 9782639-9782639 on Build GRCh38
rs746077383 Chr.4: 9782632-9782632 on Build GRCh38
rs150176923 Chr.4: 9782452-9782452 on Build GRCh38
rs896687934 Chr.4: 9782469-9782469 on Build GRCh38
rs749801481 Chr.4: 9782598-9782598 on Build GRCh38
rs779592809 Chr.4: 9782380-9782380 on Build GRCh38
rs147655139 Chr.4: 9782345-9782345 on Build GRCh38
rs750193946 Chr.4: 9782730-9782730 on Build GRCh38

View Full Product Details