Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9783305-9783813 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6168211 c.1291G>T p.E431* Chr.4: 9783320-9783320 on Build GRCh38
3826471 c.1300T>A p.F434I Chr.4: 9783329-9783329 on Build GRCh38
1059126 c.1284C>T p.D428D Chr.4: 9783313-9783313 on Build GRCh38
6168212 c.1281C>T p.N427N Chr.4: 9783310-9783310 on Build GRCh38
1059127 c.1306C>T p.R436C Chr.4: 9783335-9783335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1064138 Chr.4: 9783394-9783394 on Build GRCh38
rs775745746 Chr.4: 9783550-9783550 on Build GRCh38
rs867973828 Chr.4: 9783336-9783336 on Build GRCh38
rs943389257 Chr.4: 9783353-9783353 on Build GRCh38
rs1967551 Chr.4: 9783510-9783510 on Build GRCh38
rs778254921 Chr.4: 9783484-9783484 on Build GRCh38
rs775082292 Chr.4: 9783496-9783496 on Build GRCh38
rs910596367 Chr.4: 9783766-9783766 on Build GRCh38
rs934620937 Chr.4: 9783614-9783614 on Build GRCh38
rs972186825 Chr.4: 9783558-9783558 on Build GRCh38
rs34514924 Chr.4: 9783731-9783731 on Build GRCh38
rs1016905856 Chr.4: 9783539-9783539 on Build GRCh38
rs534114386 Chr.4: 9783664-9783664 on Build GRCh38
rs150352708 Chr.4: 9783342-9783342 on Build GRCh38
rs745499859 Chr.4: 9783490-9783490 on Build GRCh38
rs964851120 Chr.4: 9783767-9783767 on Build GRCh38
rs1014106880 Chr.4: 9783635-9783635 on Build GRCh38
rs528816164 Chr.4: 9783524-9783524 on Build GRCh38
rs371426510 Chr.4: 9783774-9783774 on Build GRCh38
rs879087241 Chr.4: 9783358-9783358 on Build GRCh38
rs919464744 Chr.4: 9783562-9783563 on Build GRCh38
rs569016288 Chr.4: 9783625-9783625 on Build GRCh38
rs753931613 Chr.4: 9783453-9783453 on Build GRCh38
rs750699062 Chr.4: 9783331-9783331 on Build GRCh38
rs905631544 Chr.4: 9783645-9783645 on Build GRCh38
rs1064137 Chr.4: 9783382-9783382 on Build GRCh38
rs755432095 Chr.4: 9783459-9783459 on Build GRCh38
rs540731190 Chr.4: 9783410-9783410 on Build GRCh38
rs748536594 Chr.4: 9783368-9783368 on Build GRCh38
rs762916143 Chr.4: 9783409-9783409 on Build GRCh38
rs954785080 Chr.4: 9783567-9783567 on Build GRCh38
rs780239142 Chr.4: 9783343-9783343 on Build GRCh38
rs779431218 Chr.4: 9783460-9783460 on Build GRCh38
rs564604787 Chr.4: 9783520-9783520 on Build GRCh38
rs1005601977 Chr.4: 9783655-9783655 on Build GRCh38
rs563301478 Chr.4: 9783467-9783467 on Build GRCh38
rs189072747 Chr.4: 9783583-9783583 on Build GRCh38
rs748394404 Chr.4: 9783543-9783543 on Build GRCh38
rs372890003 Chr.4: 9783514-9783514 on Build GRCh38
rs748827680 Chr.4: 9783499-9783499 on Build GRCh38
rs575474654 Chr.4: 9783347-9783350 on Build GRCh38
rs562881679 Chr.4: 9783722-9783722 on Build GRCh38
rs142559173 Chr.4: 9783424-9783424 on Build GRCh38
rs555093968 Chr.4: 9783698-9783698 on Build GRCh38
rs374749357 Chr.4: 9783482-9783482 on Build GRCh38
rs113892809 Chr.4: 9783530-9783530 on Build GRCh38
rs768069474 Chr.4: 9783414-9783414 on Build GRCh38
rs766615396 Chr.4: 9783515-9783515 on Build GRCh38
rs1052940088 Chr.4: 9783616-9783616 on Build GRCh38
rs533730716 Chr.4: 9783352-9783352 on Build GRCh38
rs536502048 Chr.4: 9783730-9783731 on Build GRCh38
rs575223156 Chr.4: 9783506-9783506 on Build GRCh38
rs954834968 Chr.4: 9783750-9783750 on Build GRCh38
rs868125937 Chr.4: 9783494-9783494 on Build GRCh38
rs767395524 Chr.4: 9783415-9783415 on Build GRCh38
rs777036084 Chr.4: 9783743-9783743 on Build GRCh38
rs35127018 Chr.4: 9783543-9783544 on Build GRCh38
rs759278863 Chr.4: 9783388-9783388 on Build GRCh38
rs751348585 Chr.4: 9783411-9783411 on Build GRCh38
rs186756507 Chr.4: 9783612-9783612 on Build GRCh38
rs766382205 Chr.4: 9783434-9783434 on Build GRCh38
rs768674099 Chr.4: 9783501-9783501 on Build GRCh38
rs539061507 Chr.4: 9783632-9783632 on Build GRCh38
rs573510948 Chr.4: 9783740-9783740 on Build GRCh38
rs191141694 Chr.4: 9783658-9783658 on Build GRCh38
rs993744101 Chr.4: 9783739-9783739 on Build GRCh38
rs776114395 Chr.4: 9783387-9783387 on Build GRCh38
rs779829910 Chr.4: 9783437-9783437 on Build GRCh38
rs775427531 Chr.4: 9783457-9783457 on Build GRCh38
rs761482714 Chr.4: 9783414-9783414 on Build GRCh38
rs770575054 Chr.4: 9783386-9783386 on Build GRCh38
rs769490992 Chr.4: 9783495-9783495 on Build GRCh38
rs1016630366 Chr.4: 9783673-9783673 on Build GRCh38
rs759989601 Chr.4: 9783751-9783751 on Build GRCh38
rs987483311 Chr.4: 9783574-9783574 on Build GRCh38
rs377000033 Chr.4: 9783771-9783771 on Build GRCh38
rs781290478 Chr.4: 9783327-9783327 on Build GRCh38
rs1967550 Chr.4: 9783641-9783641 on Build GRCh38
rs181467183 Chr.4: 9783589-9783589 on Build GRCh38
rs879610499 Chr.4: 9783613-9783613 on Build GRCh38
rs376052395 Chr.4: 9783366-9783366 on Build GRCh38
rs775206734 Chr.4: 9783406-9783406 on Build GRCh38
rs750598038 Chr.4: 9783423-9783423 on Build GRCh38
rs546845104 Chr.4: 9783544-9783544 on Build GRCh38

View Full Product Details