Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 9783700-9784199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD5
Entrez Gene ID
1816
Gene Name
dopamine receptor D5
Gene Aliases
DBDR, DRD1B, DRD1L2
UniGene
Hs.380681
Gene Chromosome Location
Chr.4: 9781634-9784009 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

Gene Symbol
SLC2A9
Entrez Gene ID
56606
Gene Name
solute carrier family 2 member 9
Gene Aliases
GLUT9, GLUTX, UAQTL2, URATv1
UniGene
Hs.444612
Gene Chromosome Location
Chr.4: 9771125-10040248 on Build GRCh38
Transcripts
NM_000798.4 XM_006713968.3 XM_011513856.2 XM_011513857.1 XM_011513858.1 XM_011513862.2 XM_011513863.2 XM_011513864.2 XM_011513867.2 XM_017008457.1 XM_017008459.1

SNP Detail

SNP ID Chromosome Location
rs905597612 Chr.4: 9783954-9783954 on Build GRCh38
rs767060497 Chr.4: 9784040-9784040 on Build GRCh38
rs978905098 Chr.4: 9784157-9784157 on Build GRCh38
rs367668848 Chr.4: 9784099-9784099 on Build GRCh38
rs560656317 Chr.4: 9783991-9783991 on Build GRCh38
rs113678207 Chr.4: 9784143-9784143 on Build GRCh38
rs1017793686 Chr.4: 9784144-9784144 on Build GRCh38
rs536502048 Chr.4: 9783730-9783731 on Build GRCh38
rs954834968 Chr.4: 9783750-9783750 on Build GRCh38
rs200396084 Chr.4: 9783797-9783797 on Build GRCh38
rs370580507 Chr.4: 9784128-9784128 on Build GRCh38
rs910596367 Chr.4: 9783766-9783766 on Build GRCh38
rs926016707 Chr.4: 9783839-9783839 on Build GRCh38
rs777036084 Chr.4: 9783743-9783743 on Build GRCh38
rs1038435965 Chr.4: 9783982-9783982 on Build GRCh38
rs34514924 Chr.4: 9783731-9783731 on Build GRCh38
rs762245203 Chr.4: 9783822-9783822 on Build GRCh38
rs779761982 Chr.4: 9783824-9783824 on Build GRCh38
rs185764800 Chr.4: 9783934-9783934 on Build GRCh38
rs964851120 Chr.4: 9783767-9783767 on Build GRCh38
rs371426510 Chr.4: 9783774-9783774 on Build GRCh38
rs573510948 Chr.4: 9783740-9783740 on Build GRCh38
rs993744101 Chr.4: 9783739-9783739 on Build GRCh38
rs373824597 Chr.4: 9784068-9784068 on Build GRCh38
rs183259207 Chr.4: 9784172-9784172 on Build GRCh38
rs1026498406 Chr.4: 9784079-9784079 on Build GRCh38
rs759989601 Chr.4: 9783751-9783751 on Build GRCh38
rs748892950 Chr.4: 9783841-9783841 on Build GRCh38
rs1052992321 Chr.4: 9783847-9783847 on Build GRCh38
rs377000033 Chr.4: 9783771-9783771 on Build GRCh38
rs978718960 Chr.4: 9783836-9783848 on Build GRCh38
rs150796194 Chr.4: 9783960-9783960 on Build GRCh38
rs557031730 Chr.4: 9783843-9783843 on Build GRCh38
rs374019749 Chr.4: 9784137-9784137 on Build GRCh38
rs954969914 Chr.4: 9784105-9784105 on Build GRCh38
rs141292564 Chr.4: 9783897-9783897 on Build GRCh38
rs768004687 Chr.4: 9783896-9783896 on Build GRCh38
rs151074574 Chr.4: 9783842-9783843 on Build GRCh38
rs1033520642 Chr.4: 9784169-9784169 on Build GRCh38
rs965156496 Chr.4: 9784156-9784156 on Build GRCh38
rs768341533 Chr.4: 9783884-9783884 on Build GRCh38
rs533440848 Chr.4: 9784154-9784154 on Build GRCh38
rs896783445 Chr.4: 9784029-9784029 on Build GRCh38
rs1044264334 Chr.4: 9783952-9783952 on Build GRCh38
rs191110325 Chr.4: 9784164-9784164 on Build GRCh38
rs180936488 Chr.4: 9783925-9783925 on Build GRCh38
rs562226202 Chr.4: 9784069-9784069 on Build GRCh38

View Full Product Details