Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 24539934-24540434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX15
Entrez Gene ID
1665
Gene Name
DEAH-box helicase 15
Gene Aliases
DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43p
UniGene
Hs.696074
Gene Chromosome Location
Chr.4: 24527464-24584603 on Build GRCh38
Transcripts
NM_001358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4124156 c.1741T>C p.Y581H Chr.4: 24540153-24540153 on Build GRCh38
5451769 c.1610T>C p.M537T Chr.4: 24540284-24540284 on Build GRCh38
733666 c.1636C>A p.L546M Chr.4: 24540258-24540258 on Build GRCh38
6578620 c.1609A>C p.M537L Chr.4: 24540285-24540285 on Build GRCh38
300278 c.1671A>G p.E557E Chr.4: 24540223-24540223 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139327915 Chr.4: 24540295-24540295 on Build GRCh38
rs747413812 Chr.4: 24539962-24539962 on Build GRCh38
rs1038250774 Chr.4: 24539969-24539969 on Build GRCh38
rs936461561 Chr.4: 24540005-24540005 on Build GRCh38
rs766232609 Chr.4: 24540099-24540102 on Build GRCh38
rs372466605 Chr.4: 24540132-24540132 on Build GRCh38
rs752876212 Chr.4: 24540280-24540280 on Build GRCh38
rs955475850 Chr.4: 24540050-24540050 on Build GRCh38
rs3749588 Chr.4: 24540000-24540000 on Build GRCh38
rs777075399 Chr.4: 24540092-24540092 on Build GRCh38
rs202140718 Chr.4: 24540353-24540354 on Build GRCh38
rs769974659 Chr.4: 24540065-24540066 on Build GRCh38
rs956171570 Chr.4: 24540094-24540094 on Build GRCh38
rs748150312 Chr.4: 24540062-24540062 on Build GRCh38
rs200166548 Chr.4: 24540232-24540232 on Build GRCh38
rs1028724980 Chr.4: 24540026-24540026 on Build GRCh38
rs187334769 Chr.4: 24540328-24540328 on Build GRCh38
rs768968970 Chr.4: 24539999-24539999 on Build GRCh38
rs747470843 Chr.4: 24540079-24540079 on Build GRCh38
rs538894078 Chr.4: 24540322-24540322 on Build GRCh38
rs945046748 Chr.4: 24539980-24539980 on Build GRCh38
rs763072154 Chr.4: 24540348-24540348 on Build GRCh38
rs762725804 Chr.4: 24540094-24540094 on Build GRCh38
rs370256081 Chr.4: 24540338-24540338 on Build GRCh38
rs773574555 Chr.4: 24540072-24540072 on Build GRCh38
rs567060089 Chr.4: 24540320-24540320 on Build GRCh38
rs1034922294 Chr.4: 24540339-24540339 on Build GRCh38
rs761216650 Chr.4: 24540193-24540193 on Build GRCh38
rs530882545 Chr.4: 24540404-24540409 on Build GRCh38
rs767558812 Chr.4: 24540199-24540199 on Build GRCh38
rs200810199 Chr.4: 24540256-24540256 on Build GRCh38
rs769919262 Chr.4: 24540235-24540235 on Build GRCh38
rs113961055 Chr.4: 24540205-24540205 on Build GRCh38
rs147661530 Chr.4: 24540174-24540174 on Build GRCh38
rs199863119 Chr.4: 24540228-24540228 on Build GRCh38
rs141880755 Chr.4: 24540133-24540133 on Build GRCh38
rs774024235 Chr.4: 24540351-24540352 on Build GRCh38
rs543233923 Chr.4: 24540378-24540378 on Build GRCh38
rs1050005401 Chr.4: 24540017-24540017 on Build GRCh38
rs571629920 Chr.4: 24540080-24540081 on Build GRCh38
rs771424052 Chr.4: 24540080-24540080 on Build GRCh38
rs1040977788 Chr.4: 24540126-24540126 on Build GRCh38
rs760551356 Chr.4: 24540217-24540217 on Build GRCh38
rs36050518 Chr.4: 24540266-24540266 on Build GRCh38
rs990723218 Chr.4: 24540103-24540103 on Build GRCh38
rs970699878 Chr.4: 24540170-24540170 on Build GRCh38
rs772679930 Chr.4: 24540067-24540067 on Build GRCh38

View Full Product Details