Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 24542801-24543348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX15
Entrez Gene ID
1665
Gene Name
DEAH-box helicase 15
Gene Aliases
DBP1, DDX15, HRH2, PRP43, PRPF43, PrPp43p
UniGene
Hs.696074
Gene Chromosome Location
Chr.4: 24527464-24584603 on Build GRCh38
Transcripts
NM_001358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5866221 c.1273G>A p.E425K Chr.4: 24543002-24543002 on Build GRCh38
1429218 c.1258T>C p.S420P Chr.4: 24543017-24543017 on Build GRCh38
220437 c.1327A>G p.K443E Chr.4: 24542948-24542948 on Build GRCh38
4464383 c.1330C>T p.Q444* Chr.4: 24542945-24542945 on Build GRCh38
6643404 c.1319G>A p.G440E Chr.4: 24542956-24542956 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764281638 Chr.4: 24543040-24543040 on Build GRCh38
rs554481852 Chr.4: 24542911-24542911 on Build GRCh38
rs558579529 Chr.4: 24542842-24542842 on Build GRCh38
rs780316420 Chr.4: 24542892-24542892 on Build GRCh38
rs775190098 Chr.4: 24543029-24543029 on Build GRCh38
rs540157418 Chr.4: 24543047-24543047 on Build GRCh38
rs894282213 Chr.4: 24543259-24543259 on Build GRCh38
rs1014149393 Chr.4: 24542871-24542871 on Build GRCh38
rs372681345 Chr.4: 24543120-24543120 on Build GRCh38
rs866522443 Chr.4: 24543320-24543320 on Build GRCh38
rs893022661 Chr.4: 24543287-24543287 on Build GRCh38
rs1045720452 Chr.4: 24543269-24543269 on Build GRCh38
rs753203519 Chr.4: 24543089-24543089 on Build GRCh38
rs925747800 Chr.4: 24543232-24543232 on Build GRCh38
rs867420919 Chr.4: 24543234-24543234 on Build GRCh38
rs957364592 Chr.4: 24542827-24542828 on Build GRCh38
rs768048912 Chr.4: 24542875-24542875 on Build GRCh38
rs538687418 Chr.4: 24542835-24542835 on Build GRCh38
rs137864122 Chr.4: 24543262-24543262 on Build GRCh38
rs766884225 Chr.4: 24543059-24543059 on Build GRCh38
rs774718539 Chr.4: 24543046-24543046 on Build GRCh38
rs146220881 Chr.4: 24543056-24543056 on Build GRCh38
rs191764928 Chr.4: 24543217-24543217 on Build GRCh38
rs755370388 Chr.4: 24543068-24543068 on Build GRCh38
rs745817057 Chr.4: 24542997-24542997 on Build GRCh38
rs1038427133 Chr.4: 24543066-24543066 on Build GRCh38
rs894413122 Chr.4: 24542846-24542846 on Build GRCh38
rs779186523 Chr.4: 24543069-24543069 on Build GRCh38
rs778136351 Chr.4: 24543301-24543301 on Build GRCh38
rs776064489 Chr.4: 24542988-24542988 on Build GRCh38
rs201977832 Chr.4: 24543052-24543052 on Build GRCh38
rs762787646 Chr.4: 24543038-24543038 on Build GRCh38
rs113889895 Chr.4: 24543158-24543158 on Build GRCh38
rs990543035 Chr.4: 24542828-24542831 on Build GRCh38
rs962033240 Chr.4: 24542882-24542882 on Build GRCh38
rs1011529874 Chr.4: 24543310-24543310 on Build GRCh38
rs746592647 Chr.4: 24542935-24542935 on Build GRCh38
rs575447718 Chr.4: 24542880-24542880 on Build GRCh38
rs370609421 Chr.4: 24543060-24543060 on Build GRCh38
rs749631187 Chr.4: 24542903-24542903 on Build GRCh38
rs1055646921 Chr.4: 24543257-24543257 on Build GRCh38
rs1052910562 Chr.4: 24543280-24543280 on Build GRCh38
rs187424781 Chr.4: 24543003-24543003 on Build GRCh38
rs531736245 Chr.4: 24543159-24543159 on Build GRCh38
rs758032651 Chr.4: 24542909-24542909 on Build GRCh38
rs537915513 Chr.4: 24542888-24542888 on Build GRCh38
rs562520102 Chr.4: 24543140-24543140 on Build GRCh38
rs770435424 Chr.4: 24542949-24542949 on Build GRCh38
rs945765172 Chr.4: 24543121-24543121 on Build GRCh38

View Full Product Details