Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 37438680-37439178 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NWD2
Entrez Gene ID
57495
Gene Name
NACHT and WD repeat domain containing 2
Gene Aliases
KIAA1239
UniGene
Hs.4280
Gene Chromosome Location
Chr.4: 37245068-37449465 on Build GRCh38
Transcripts
NM_001144990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4609064 c.1009C>A p.H337N Chr.4: 37439103-37439103 on Build GRCh38
3825783 c.1017C>A p.I339I Chr.4: 37439111-37439111 on Build GRCh38
5941903 c.748C>T p.P250S Chr.4: 37438842-37438842 on Build GRCh38
6877396 c.789T>C p.I263I Chr.4: 37438883-37438883 on Build GRCh38
5931504 c.779T>C p.I260T Chr.4: 37438873-37438873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs865858430 Chr.4: 37439053-37439053 on Build GRCh38
rs1046841965 Chr.4: 37438844-37438844 on Build GRCh38
rs994905407 Chr.4: 37438722-37438722 on Build GRCh38
rs912879238 Chr.4: 37438951-37438951 on Build GRCh38
rs1009969498 Chr.4: 37439103-37439103 on Build GRCh38
rs773963313 Chr.4: 37439095-37439095 on Build GRCh38
rs769720546 Chr.4: 37438971-37438971 on Build GRCh38
rs778171840 Chr.4: 37438947-37438947 on Build GRCh38
rs779117767 Chr.4: 37438787-37438787 on Build GRCh38
rs1003808643 Chr.4: 37438952-37438952 on Build GRCh38
rs34189147 Chr.4: 37438998-37438999 on Build GRCh38
rs1022418879 Chr.4: 37439144-37439144 on Build GRCh38
rs139138063 Chr.4: 37438780-37438780 on Build GRCh38
rs1035658261 Chr.4: 37438955-37438955 on Build GRCh38
rs527765753 Chr.4: 37438944-37438944 on Build GRCh38
rs761255574 Chr.4: 37439096-37439096 on Build GRCh38
rs752838933 Chr.4: 37438907-37438907 on Build GRCh38
rs897941844 Chr.4: 37438718-37438718 on Build GRCh38
rs1055967603 Chr.4: 37439151-37439151 on Build GRCh38
rs35241919 Chr.4: 37438921-37438922 on Build GRCh38
rs780123645 Chr.4: 37438978-37438978 on Build GRCh38
rs976413647 Chr.4: 37438934-37438934 on Build GRCh38
rs1045258902 Chr.4: 37438896-37438896 on Build GRCh38
rs978496812 Chr.4: 37439031-37439031 on Build GRCh38
rs532827655 Chr.4: 37439111-37439111 on Build GRCh38
rs189191557 Chr.4: 37438894-37438894 on Build GRCh38
rs768021995 Chr.4: 37439048-37439048 on Build GRCh38
rs892491181 Chr.4: 37438974-37438974 on Build GRCh38
rs547623268 Chr.4: 37439042-37439042 on Build GRCh38
rs762332151 Chr.4: 37438781-37438783 on Build GRCh38
rs929615038 Chr.4: 37438820-37438820 on Build GRCh38
rs1027718707 Chr.4: 37438759-37438759 on Build GRCh38
rs937511376 Chr.4: 37439117-37439117 on Build GRCh38
rs142483670 Chr.4: 37439150-37439150 on Build GRCh38
rs567644799 Chr.4: 37439148-37439148 on Build GRCh38
rs888186534 Chr.4: 37438890-37438890 on Build GRCh38
rs531504870 Chr.4: 37438716-37438716 on Build GRCh38
rs760469448 Chr.4: 37438962-37438962 on Build GRCh38
rs771676203 Chr.4: 37439109-37439109 on Build GRCh38

View Full Product Details