Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 44174161-44174675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD8
Entrez Gene ID
386617
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
UniGene
Hs.479644
Gene Chromosome Location
Chr.4: 44173903-44448885 on Build GRCh38
Transcripts
NM_198353.2 XM_011513690.2

SNP Detail

SNP ID Chromosome Location
rs1046546005 Chr.4: 44174534-44174534 on Build GRCh38
rs988191171 Chr.4: 44174339-44174339 on Build GRCh38
rs553954911 Chr.4: 44174536-44174536 on Build GRCh38
rs535342070 Chr.4: 44174478-44174478 on Build GRCh38
rs551946222 Chr.4: 44174232-44174232 on Build GRCh38
rs956696408 Chr.4: 44174326-44174326 on Build GRCh38
rs970844005 Chr.4: 44174381-44174381 on Build GRCh38
rs570429904 Chr.4: 44174233-44174233 on Build GRCh38
rs762574766 Chr.4: 44174503-44174503 on Build GRCh38
rs112572952 Chr.4: 44174584-44174584 on Build GRCh38
rs1007797635 Chr.4: 44174252-44174252 on Build GRCh38
rs919610322 Chr.4: 44174437-44174437 on Build GRCh38
rs182174248 Chr.4: 44174242-44174242 on Build GRCh38
rs200535947 Chr.4: 44174629-44174631 on Build GRCh38
rs555729860 Chr.4: 44174250-44174250 on Build GRCh38
rs987068562 Chr.4: 44174606-44174606 on Build GRCh38
rs1007331354 Chr.4: 44174586-44174586 on Build GRCh38
rs981329861 Chr.4: 44174391-44174391 on Build GRCh38
rs781210690 Chr.4: 44174598-44174598 on Build GRCh38
rs113112956 Chr.4: 44174221-44174222 on Build GRCh38
rs557917427 Chr.4: 44174642-44174642 on Build GRCh38
rs1037760326 Chr.4: 44174190-44174190 on Build GRCh38
rs371434545 Chr.4: 44174587-44174587 on Build GRCh38
rs958002132 Chr.4: 44174223-44174223 on Build GRCh38
rs202057347 Chr.4: 44174233-44174233 on Build GRCh38
rs912321960 Chr.4: 44174280-44174280 on Build GRCh38
rs928980729 Chr.4: 44174525-44174525 on Build GRCh38
rs969020240 Chr.4: 44174289-44174289 on Build GRCh38
rs572110410 Chr.4: 44174576-44174576 on Build GRCh38
rs1009561623 Chr.4: 44174573-44174573 on Build GRCh38
rs904368413 Chr.4: 44174613-44174613 on Build GRCh38
rs918001276 Chr.4: 44174365-44174365 on Build GRCh38
rs967653069 Chr.4: 44174245-44174245 on Build GRCh38
rs374685371 Chr.4: 44174248-44174248 on Build GRCh38
rs967305384 Chr.4: 44174605-44174605 on Build GRCh38
rs781124641 Chr.4: 44174543-44174543 on Build GRCh38
rs978728641 Chr.4: 44174305-44174305 on Build GRCh38
rs186307781 Chr.4: 44174285-44174285 on Build GRCh38
rs918883880 Chr.4: 44174520-44174520 on Build GRCh38
rs897510472 Chr.4: 44174208-44174208 on Build GRCh38
rs890200778 Chr.4: 44174567-44174567 on Build GRCh38
rs1032387237 Chr.4: 44174562-44174562 on Build GRCh38
rs368998012 Chr.4: 44174473-44174473 on Build GRCh38
rs1039830312 Chr.4: 44174367-44174367 on Build GRCh38
rs778311574 Chr.4: 44174597-44174598 on Build GRCh38
rs1032313542 Chr.4: 44174307-44174307 on Build GRCh38
rs527398694 Chr.4: 44174231-44174231 on Build GRCh38
rs990070579 Chr.4: 44174232-44174233 on Build GRCh38
rs754739787 Chr.4: 44174474-44174474 on Build GRCh38
rs1032357777 Chr.4: 44174221-44174221 on Build GRCh38

View Full Product Details