Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47515291-47515786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP10D
Entrez Gene ID
57205
Gene Name
ATPase phospholipid transporting 10D (putative)
Gene Aliases
ATPVD
UniGene
Hs.437241
Gene Chromosome Location
Chr.4: 47485261-47593486 on Build GRCh38
Transcripts
NM_020453.3 XM_005248119.4 XM_005248120.4 XM_011513722.2 XM_017008472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1618868 c.412G>A p.D138N Chr.4: 47515597-47515597 on Build GRCh38
7255296 c.430C>A p.R144R Chr.4: 47515615-47515615 on Build GRCh38
6167543 c.360G>T p.K120N Chr.4: 47515545-47515545 on Build GRCh38
4491267 c.377C>T p.P126L Chr.4: 47515562-47515562 on Build GRCh38
4940972 c.331T>C p.W111R Chr.4: 47515516-47515516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769698698 Chr.4: 47515641-47515641 on Build GRCh38
rs1025644928 Chr.4: 47515327-47515327 on Build GRCh38
rs756442186 Chr.4: 47515563-47515563 on Build GRCh38
rs762570178 Chr.4: 47515500-47515500 on Build GRCh38
rs748267873 Chr.4: 47515644-47515648 on Build GRCh38
rs750910886 Chr.4: 47515713-47515713 on Build GRCh38
rs767056951 Chr.4: 47515701-47515701 on Build GRCh38
rs769312328 Chr.4: 47515631-47515634 on Build GRCh38
rs980392547 Chr.4: 47515744-47515744 on Build GRCh38
rs757700600 Chr.4: 47515514-47515514 on Build GRCh38
rs773859110 Chr.4: 47515699-47515699 on Build GRCh38
rs116597196 Chr.4: 47515354-47515354 on Build GRCh38
rs562388135 Chr.4: 47515365-47515365 on Build GRCh38
rs376785558 Chr.4: 47515710-47515710 on Build GRCh38
rs889046802 Chr.4: 47515664-47515664 on Build GRCh38
rs112932074 Chr.4: 47515352-47515352 on Build GRCh38
rs893455576 Chr.4: 47515492-47515492 on Build GRCh38
rs770997081 Chr.4: 47515462-47515462 on Build GRCh38
rs916854873 Chr.4: 47515423-47515423 on Build GRCh38
rs935079532 Chr.4: 47515446-47515446 on Build GRCh38
rs747359900 Chr.4: 47515578-47515579 on Build GRCh38
rs762595611 Chr.4: 47515676-47515676 on Build GRCh38
rs566390429 Chr.4: 47515580-47515580 on Build GRCh38
rs956122430 Chr.4: 47515662-47515662 on Build GRCh38
rs776676294 Chr.4: 47515620-47515620 on Build GRCh38
rs775214799 Chr.4: 47515657-47515657 on Build GRCh38
rs757823453 Chr.4: 47515425-47515425 on Build GRCh38
rs756675230 Chr.4: 47515717-47515717 on Build GRCh38
rs751767426 Chr.4: 47515722-47515722 on Build GRCh38
rs368259495 Chr.4: 47515467-47515467 on Build GRCh38
rs758422740 Chr.4: 47515355-47515355 on Build GRCh38
rs201079510 Chr.4: 47515497-47515497 on Build GRCh38
rs375086728 Chr.4: 47515451-47515451 on Build GRCh38
rs778984994 Chr.4: 47515613-47515613 on Build GRCh38
rs780288846 Chr.4: 47515440-47515440 on Build GRCh38
rs368560299 Chr.4: 47515721-47515721 on Build GRCh38
rs763651667 Chr.4: 47515505-47515505 on Build GRCh38
rs551926806 Chr.4: 47515684-47515684 on Build GRCh38
rs781751759 Chr.4: 47515641-47515641 on Build GRCh38
rs780602025 Chr.4: 47515532-47515533 on Build GRCh38
rs745848496 Chr.4: 47515638-47515638 on Build GRCh38
rs371064535 Chr.4: 47515507-47515507 on Build GRCh38
rs768763145 Chr.4: 47515443-47515443 on Build GRCh38
rs950239648 Chr.4: 47515712-47515712 on Build GRCh38
rs370900814 Chr.4: 47515434-47515434 on Build GRCh38
rs984351740 Chr.4: 47515402-47515402 on Build GRCh38
rs897689594 Chr.4: 47515735-47515735 on Build GRCh38
rs184019410 Chr.4: 47515487-47515487 on Build GRCh38
rs188403608 Chr.4: 47515668-47515668 on Build GRCh38
rs760658088 Chr.4: 47515459-47515459 on Build GRCh38
rs770945581 Chr.4: 47515616-47515616 on Build GRCh38
rs750749360 Chr.4: 47515534-47515534 on Build GRCh38
rs749516753 Chr.4: 47515441-47515441 on Build GRCh38
rs768032388 Chr.4: 47515527-47515527 on Build GRCh38
rs761342317 Chr.4: 47515700-47515700 on Build GRCh38
rs748191553 Chr.4: 47515615-47515615 on Build GRCh38
rs755207032 Chr.4: 47515723-47515723 on Build GRCh38
rs781465717 Chr.4: 47515427-47515427 on Build GRCh38
rs1018778244 Chr.4: 47515734-47515734 on Build GRCh38
rs989419261 Chr.4: 47515693-47515693 on Build GRCh38
rs759429598 Chr.4: 47515473-47515473 on Build GRCh38
rs749516555 Chr.4: 47515587-47515587 on Build GRCh38
rs755222321 Chr.4: 47515588-47515588 on Build GRCh38
rs763663606 Chr.4: 47515680-47515680 on Build GRCh38
rs926064307 Chr.4: 47515651-47515651 on Build GRCh38
rs769952411 Chr.4: 47515437-47515437 on Build GRCh38

View Full Product Details