Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47594866-47595410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORIN
Entrez Gene ID
10699
Gene Name
corin, serine peptidase
Gene Aliases
ATC2, CRN, Lrp4, PEE5, TMPRSS10
UniGene
Hs.518618
Gene Chromosome Location
Chr.4: 47593998-47838106 on Build GRCh38
Transcripts
NM_001278585.1 NM_006587.3

SNP Detail

SNP ID Chromosome Location
rs964010637 Chr.4: 47595310-47595310 on Build GRCh38
rs566291451 Chr.4: 47594895-47594895 on Build GRCh38
rs968937643 Chr.4: 47595231-47595231 on Build GRCh38
rs954721240 Chr.4: 47595054-47595054 on Build GRCh38
rs191428358 Chr.4: 47595200-47595200 on Build GRCh38
rs975383406 Chr.4: 47595119-47595119 on Build GRCh38
rs1015969264 Chr.4: 47594902-47594902 on Build GRCh38
rs767014812 Chr.4: 47595111-47595111 on Build GRCh38
rs113705654 Chr.4: 47595131-47595131 on Build GRCh38
rs963287372 Chr.4: 47595035-47595035 on Build GRCh38
rs1035862821 Chr.4: 47595172-47595172 on Build GRCh38
rs879875591 Chr.4: 47595164-47595164 on Build GRCh38
rs569945121 Chr.4: 47595215-47595215 on Build GRCh38
rs576072352 Chr.4: 47595031-47595031 on Build GRCh38
rs1026653322 Chr.4: 47595344-47595344 on Build GRCh38
rs61758584 Chr.4: 47595073-47595073 on Build GRCh38
rs1006700531 Chr.4: 47595159-47595159 on Build GRCh38
rs141880583 Chr.4: 47595165-47595165 on Build GRCh38
rs61758583 Chr.4: 47595040-47595040 on Build GRCh38
rs533252345 Chr.4: 47595190-47595190 on Build GRCh38
rs1035978497 Chr.4: 47594900-47594900 on Build GRCh38
rs904933549 Chr.4: 47595225-47595225 on Build GRCh38
rs61759668 Chr.4: 47595284-47595284 on Build GRCh38
rs993399071 Chr.4: 47595336-47595336 on Build GRCh38
rs986865125 Chr.4: 47595067-47595067 on Build GRCh38
rs912949066 Chr.4: 47594934-47594934 on Build GRCh38
rs543116779 Chr.4: 47595043-47595043 on Build GRCh38
rs61758585 Chr.4: 47595191-47595191 on Build GRCh38
rs551529792 Chr.4: 47595248-47595248 on Build GRCh38
rs964131403 Chr.4: 47595089-47595089 on Build GRCh38
rs555997509 Chr.4: 47595298-47595298 on Build GRCh38
rs936913272 Chr.4: 47595132-47595132 on Build GRCh38
rs942416548 Chr.4: 47595055-47595055 on Build GRCh38
rs922516644 Chr.4: 47595079-47595079 on Build GRCh38
rs564056328 Chr.4: 47594987-47594987 on Build GRCh38
rs1037351436 Chr.4: 47595312-47595312 on Build GRCh38
rs868839305 Chr.4: 47595268-47595268 on Build GRCh38
rs968452076 Chr.4: 47595019-47595019 on Build GRCh38
rs976453211 Chr.4: 47595335-47595335 on Build GRCh38
rs1001580083 Chr.4: 47595166-47595166 on Build GRCh38
rs988939504 Chr.4: 47594933-47594934 on Build GRCh38
rs1037816976 Chr.4: 47595162-47595162 on Build GRCh38
rs546377271 Chr.4: 47594944-47594944 on Build GRCh38
rs898860786 Chr.4: 47595333-47595333 on Build GRCh38
rs61759667 Chr.4: 47595240-47595240 on Build GRCh38
rs1046111360 Chr.4: 47595224-47595224 on Build GRCh38
rs1026134227 Chr.4: 47595051-47595051 on Build GRCh38
rs960192877 Chr.4: 47594911-47594911 on Build GRCh38
rs763368719 Chr.4: 47594952-47594952 on Build GRCh38
rs533003304 Chr.4: 47595241-47595241 on Build GRCh38

View Full Product Details