Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47595295-47595804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORIN
Entrez Gene ID
10699
Gene Name
corin, serine peptidase
Gene Aliases
ATC2, CRN, Lrp4, PEE5, TMPRSS10
UniGene
Hs.518618
Gene Chromosome Location
Chr.4: 47593998-47838106 on Build GRCh38
Transcripts
NM_001278585.1 NM_006587.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4800528 c.3055G>A p.V1019I Chr.4: 47595795-47595795 on Build GRCh38
6529220 c.3109C>G p.Q1037E Chr.4: 47595741-47595741 on Build GRCh38
1694340 c.3082G>A p.E1028K Chr.4: 47595768-47595768 on Build GRCh38
258647 c.3071C>T p.S1024L Chr.4: 47595779-47595779 on Build GRCh38
734314 c.3127T>A p.*1043K Chr.4: 47595723-47595723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540619947 Chr.4: 47595772-47595772 on Build GRCh38
rs949694652 Chr.4: 47595580-47595580 on Build GRCh38
rs756754101 Chr.4: 47595778-47595778 on Build GRCh38
rs775759080 Chr.4: 47595702-47595702 on Build GRCh38
rs781612859 Chr.4: 47595682-47595682 on Build GRCh38
rs568476346 Chr.4: 47595516-47595516 on Build GRCh38
rs780623218 Chr.4: 47595781-47595781 on Build GRCh38
rs1017432074 Chr.4: 47595431-47595431 on Build GRCh38
rs767774967 Chr.4: 47595735-47595735 on Build GRCh38
rs751217895 Chr.4: 47595773-47595773 on Build GRCh38
rs982722013 Chr.4: 47595387-47595387 on Build GRCh38
rs767487727 Chr.4: 47595393-47595393 on Build GRCh38
rs900359528 Chr.4: 47595752-47595752 on Build GRCh38
rs377153350 Chr.4: 47595696-47595696 on Build GRCh38
rs964080794 Chr.4: 47595453-47595453 on Build GRCh38
rs1020522075 Chr.4: 47595558-47595559 on Build GRCh38
rs756263714 Chr.4: 47595490-47595490 on Build GRCh38
rs753019214 Chr.4: 47595451-47595451 on Build GRCh38
rs1026653322 Chr.4: 47595344-47595344 on Build GRCh38
rs765048638 Chr.4: 47595768-47595768 on Build GRCh38
rs776620819 Chr.4: 47595737-47595737 on Build GRCh38
rs114481233 Chr.4: 47595698-47595698 on Build GRCh38
rs112447911 Chr.4: 47595388-47595388 on Build GRCh38
rs758207729 Chr.4: 47595771-47595771 on Build GRCh38
rs768952132 Chr.4: 47595723-47595723 on Build GRCh38
rs139348561 Chr.4: 47595557-47595557 on Build GRCh38
rs750478010 Chr.4: 47595711-47595711 on Build GRCh38
rs770174493 Chr.4: 47595697-47595697 on Build GRCh38
rs946594565 Chr.4: 47595464-47595464 on Build GRCh38
rs576532968 Chr.4: 47595661-47595661 on Build GRCh38
rs572239466 Chr.4: 47595619-47595619 on Build GRCh38
rs993399071 Chr.4: 47595336-47595336 on Build GRCh38
rs963844136 Chr.4: 47595635-47595636 on Build GRCh38
rs568031235 Chr.4: 47595394-47595394 on Build GRCh38
rs746356819 Chr.4: 47595782-47595782 on Build GRCh38
rs1038187969 Chr.4: 47595436-47595436 on Build GRCh38
rs76700540 Chr.4: 47595732-47595732 on Build GRCh38
rs759437195 Chr.4: 47595748-47595748 on Build GRCh38
rs898808736 Chr.4: 47595655-47595655 on Build GRCh38
rs376673462 Chr.4: 47595769-47595769 on Build GRCh38
rs758144314 Chr.4: 47595603-47595603 on Build GRCh38
rs1029315985 Chr.4: 47595640-47595640 on Build GRCh38
rs370891199 Chr.4: 47595703-47595703 on Build GRCh38
rs774730839 Chr.4: 47595726-47595726 on Build GRCh38
rs3749584 Chr.4: 47595637-47595637 on Build GRCh38
rs976453211 Chr.4: 47595335-47595335 on Build GRCh38
rs560872385 Chr.4: 47595435-47595435 on Build GRCh38
rs183601558 Chr.4: 47595437-47595437 on Build GRCh38
rs758638403 Chr.4: 47595750-47595753 on Build GRCh38
rs3749585 Chr.4: 47595573-47595573 on Build GRCh38
rs892744192 Chr.4: 47595747-47595747 on Build GRCh38
rs867695381 Chr.4: 47595764-47595764 on Build GRCh38
rs898860786 Chr.4: 47595333-47595333 on Build GRCh38
rs570134607 Chr.4: 47595480-47595481 on Build GRCh38
rs549962865 Chr.4: 47595509-47595509 on Build GRCh38
rs535247396 Chr.4: 47595530-47595530 on Build GRCh38
rs928973108 Chr.4: 47595673-47595673 on Build GRCh38
rs982684262 Chr.4: 47595602-47595602 on Build GRCh38
rs868207934 Chr.4: 47595779-47595779 on Build GRCh38
rs543507992 Chr.4: 47595663-47595663 on Build GRCh38
rs1000372249 Chr.4: 47595597-47595597 on Build GRCh38
rs749695065 Chr.4: 47595711-47595711 on Build GRCh38
rs1009139334 Chr.4: 47595428-47595428 on Build GRCh38
rs958052020 Chr.4: 47595535-47595535 on Build GRCh38
rs539679919 Chr.4: 47595620-47595620 on Build GRCh38
rs756772633 Chr.4: 47595677-47595677 on Build GRCh38

View Full Product Details